Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64224013-64224522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPT1
Entrez Gene ID
83707
Gene Name
tRNA phosphotransferase 1
Gene Aliases
-
UniGene
Hs.326586
Gene Chromosome Location
Chr.11: 64223799-64226254 on Build GRCh38
Transcripts
NM_001033678.3 NM_001160389.1 NM_001160390.1 NM_001160392.1 NM_001160393.1 NM_031472.3 XM_005274345.4 XM_005274346.4 XM_005274347.4 XM_006718706.3 XM_006718707.3 XM_017018400.1 XM_017018401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7277228 c.671C>T p.P224L Chr.11: 64224105-64224105 on Build GRCh38
7228313 c.667C>T p.R223C Chr.11: 64224109-64224109 on Build GRCh38
3998532 c.515A>G p.H172R Chr.11: 64224329-64224329 on Build GRCh38
7322760 c.591G>T p.G197G Chr.11: 64224179-64224179 on Build GRCh38
7277229 c.665C>T p.P222L Chr.11: 64224105-64224105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773944976 Chr.11: 64224245-64224245 on Build GRCh38
rs779904553 Chr.11: 64224099-64224099 on Build GRCh38
rs573129191 Chr.11: 64224340-64224340 on Build GRCh38
rs777986459 Chr.11: 64224080-64224080 on Build GRCh38
rs771872650 Chr.11: 64224229-64224229 on Build GRCh38
rs764164975 Chr.11: 64224347-64224347 on Build GRCh38
rs754581611 Chr.11: 64224285-64224285 on Build GRCh38
rs764705523 Chr.11: 64224192-64224192 on Build GRCh38
rs774583512 Chr.11: 64224356-64224356 on Build GRCh38
rs569081074 Chr.11: 64224302-64224302 on Build GRCh38
rs779896703 Chr.11: 64224273-64224273 on Build GRCh38
rs754688179 Chr.11: 64224210-64224210 on Build GRCh38
rs755699847 Chr.11: 64224062-64224062 on Build GRCh38
rs1002898986 Chr.11: 64224472-64224472 on Build GRCh38
rs375635773 Chr.11: 64224269-64224269 on Build GRCh38
rs758829104 Chr.11: 64224187-64224187 on Build GRCh38
rs539086044 Chr.11: 64224047-64224047 on Build GRCh38
rs761149079 Chr.11: 64224492-64224492 on Build GRCh38
rs922898230 Chr.11: 64224335-64224335 on Build GRCh38
rs768095148 Chr.11: 64224097-64224098 on Build GRCh38
rs1029438279 Chr.11: 64224431-64224431 on Build GRCh38
rs35584599 Chr.11: 64224464-64224464 on Build GRCh38
rs200796315 Chr.11: 64224175-64224175 on Build GRCh38
rs775061103 Chr.11: 64224041-64224042 on Build GRCh38
rs766654216 Chr.11: 64224038-64224038 on Build GRCh38
rs770647901 Chr.11: 64224240-64224240 on Build GRCh38
rs1010457976 Chr.11: 64224414-64224414 on Build GRCh38
rs767931078 Chr.11: 64224280-64224280 on Build GRCh38
rs1020665250 Chr.11: 64224243-64224243 on Build GRCh38
rs757840258 Chr.11: 64224275-64224275 on Build GRCh38
rs189441925 Chr.11: 64224134-64224134 on Build GRCh38
rs762748800 Chr.11: 64224458-64224458 on Build GRCh38
rs1059440 Chr.11: 64224329-64224329 on Build GRCh38
rs373285281 Chr.11: 64224183-64224183 on Build GRCh38
rs768200879 Chr.11: 64224370-64224370 on Build GRCh38
rs751953482 Chr.11: 64224193-64224193 on Build GRCh38
rs373880889 Chr.11: 64224346-64224346 on Build GRCh38
rs997638800 Chr.11: 64224121-64224121 on Build GRCh38
rs147179194 Chr.11: 64224057-64224057 on Build GRCh38
rs776426065 Chr.11: 64224245-64224245 on Build GRCh38
rs748880595 Chr.11: 64224201-64224201 on Build GRCh38
rs370231160 Chr.11: 64224314-64224314 on Build GRCh38
rs372890333 Chr.11: 64224252-64224252 on Build GRCh38
rs1042433956 Chr.11: 64224247-64224247 on Build GRCh38
rs757650003 Chr.11: 64224195-64224195 on Build GRCh38
rs781712780 Chr.11: 64224200-64224200 on Build GRCh38
rs11549690 Chr.11: 64224109-64224109 on Build GRCh38
rs761362420 Chr.11: 64224384-64224385 on Build GRCh38
rs140410065 Chr.11: 64224375-64224375 on Build GRCh38
rs976531315 Chr.11: 64224421-64224421 on Build GRCh38
rs762392788 Chr.11: 64224360-64224360 on Build GRCh38
rs777154490 Chr.11: 64224307-64224307 on Build GRCh38
rs753080202 Chr.11: 64224062-64224062 on Build GRCh38
rs766564317 Chr.11: 64224150-64224150 on Build GRCh38
rs372630710 Chr.11: 64224097-64224097 on Build GRCh38
rs775405339 Chr.11: 64224249-64224249 on Build GRCh38
rs753882486 Chr.11: 64224046-64224046 on Build GRCh38
rs747760690 Chr.11: 64224218-64224218 on Build GRCh38
rs116905598 Chr.11: 64224336-64224336 on Build GRCh38
rs746969916 Chr.11: 64224235-64224235 on Build GRCh38
rs760502324 Chr.11: 64224135-64224135 on Build GRCh38
rs775735807 Chr.11: 64224344-64224344 on Build GRCh38
rs778269038 Chr.11: 64224216-64224216 on Build GRCh38
rs747477193 Chr.11: 64224092-64224092 on Build GRCh38
rs772104416 Chr.11: 64224128-64224128 on Build GRCh38
rs577123097 Chr.11: 64224328-64224328 on Build GRCh38
rs909063879 Chr.11: 64224289-64224289 on Build GRCh38
rs946703436 Chr.11: 64224461-64224461 on Build GRCh38
rs773913933 Chr.11: 64224108-64224108 on Build GRCh38
rs778666111 Chr.11: 64224300-64224300 on Build GRCh38
rs184646976 Chr.11: 64224454-64224454 on Build GRCh38
rs757323255 Chr.11: 64224378-64224378 on Build GRCh38
rs530408080 Chr.11: 64224071-64224071 on Build GRCh38
rs1050598191 Chr.11: 64224368-64224368 on Build GRCh38
rs369333740 Chr.11: 64224248-64224248 on Build GRCh38
rs754020655 Chr.11: 64224167-64224167 on Build GRCh38
rs747049298 Chr.11: 64224498-64224498 on Build GRCh38
rs538111971 Chr.11: 64224306-64224306 on Build GRCh38
rs374904463 Chr.11: 64224233-64224233 on Build GRCh38
rs375889630 Chr.11: 64224100-64224100 on Build GRCh38
rs146774292 Chr.11: 64224184-64224184 on Build GRCh38
rs367785158 Chr.11: 64224079-64224079 on Build GRCh38
rs376217986 Chr.11: 64224394-64224394 on Build GRCh38
rs750871550 Chr.11: 64224283-64224283 on Build GRCh38
rs879071737 Chr.11: 64224417-64224417 on Build GRCh38
rs369921555 Chr.11: 64224402-64224402 on Build GRCh38
rs575856954 Chr.11: 64224445-64224445 on Build GRCh38
rs752595262 Chr.11: 64224403-64224403 on Build GRCh38

View Full Product Details