Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64343101-64343592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC88B
Entrez Gene ID
283234
Gene Name
coiled-coil domain containing 88B
Gene Aliases
BRLZ, CCDC88, HKRP3, gipie
UniGene
Hs.98564
Gene Chromosome Location
Chr.11: 64340203-64357534 on Build GRCh38
Transcripts
NM_032251.5 XM_006718519.3 XM_011544957.2 XM_011544959.2 XM_011544960.2 XM_017017601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4602708 c.1166G>A p.R389H Chr.11: 64343282-64343282 on Build GRCh38
6323457 c.1202C>T p.A401V Chr.11: 64343318-64343318 on Build GRCh38
429536 c.1095G>C p.A365A Chr.11: 64343211-64343211 on Build GRCh38
6310832 c.1176G>T p.L392L Chr.11: 64343292-64343292 on Build GRCh38
930077 c.1292A>G p.E431G Chr.11: 64343589-64343589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970045954 Chr.11: 64343501-64343501 on Build GRCh38
rs763200470 Chr.11: 64343316-64343316 on Build GRCh38
rs548514744 Chr.11: 64343313-64343313 on Build GRCh38
rs35692796 Chr.11: 64343333-64343333 on Build GRCh38
rs777376317 Chr.11: 64343249-64343249 on Build GRCh38
rs479552 Chr.11: 64343211-64343211 on Build GRCh38
rs917354143 Chr.11: 64343192-64343192 on Build GRCh38
rs769613676 Chr.11: 64343310-64343310 on Build GRCh38
rs759849547 Chr.11: 64343175-64343175 on Build GRCh38
rs143193161 Chr.11: 64343457-64343457 on Build GRCh38
rs534974036 Chr.11: 64343126-64343126 on Build GRCh38
rs764207814 Chr.11: 64343188-64343188 on Build GRCh38
rs756785818 Chr.11: 64343477-64343477 on Build GRCh38
rs372368269 Chr.11: 64343362-64343362 on Build GRCh38
rs200238832 Chr.11: 64343366-64343366 on Build GRCh38
rs754664428 Chr.11: 64343219-64343219 on Build GRCh38
rs994402485 Chr.11: 64343235-64343235 on Build GRCh38
rs762997411 Chr.11: 64343186-64343186 on Build GRCh38
rs563252980 Chr.11: 64343164-64343164 on Build GRCh38
rs371409114 Chr.11: 64343296-64343296 on Build GRCh38
rs552535568 Chr.11: 64343420-64343420 on Build GRCh38
rs878941230 Chr.11: 64343374-64343374 on Build GRCh38
rs866861198 Chr.11: 64343505-64343505 on Build GRCh38
rs139453508 Chr.11: 64343217-64343217 on Build GRCh38
rs542984933 Chr.11: 64343158-64343158 on Build GRCh38
rs781561787 Chr.11: 64343199-64343199 on Build GRCh38
rs764447359 Chr.11: 64343515-64343516 on Build GRCh38
rs12808002 Chr.11: 64343460-64343460 on Build GRCh38
rs1019024898 Chr.11: 64343255-64343255 on Build GRCh38
rs201489781 Chr.11: 64343166-64343166 on Build GRCh38
rs771533199 Chr.11: 64343242-64343242 on Build GRCh38
rs141405442 Chr.11: 64343522-64343522 on Build GRCh38
rs994261991 Chr.11: 64343515-64343515 on Build GRCh38
rs765306023 Chr.11: 64343182-64343182 on Build GRCh38
rs574835 Chr.11: 64343196-64343196 on Build GRCh38
rs746963494 Chr.11: 64343270-64343270 on Build GRCh38
rs368605372 Chr.11: 64343361-64343361 on Build GRCh38
rs763435540 Chr.11: 64343482-64343483 on Build GRCh38
rs776570567 Chr.11: 64343304-64343304 on Build GRCh38
rs913117847 Chr.11: 64343418-64343418 on Build GRCh38
rs759312375 Chr.11: 64343307-64343307 on Build GRCh38
rs1025855391 Chr.11: 64343548-64343548 on Build GRCh38
rs762326013 Chr.11: 64343352-64343352 on Build GRCh38
rs777439008 Chr.11: 64343469-64343469 on Build GRCh38
rs776516949 Chr.11: 64343173-64343173 on Build GRCh38
rs550167204 Chr.11: 64343365-64343365 on Build GRCh38
rs770933713 Chr.11: 64343271-64343271 on Build GRCh38
rs760656906 Chr.11: 64343160-64343160 on Build GRCh38
rs757725251 Chr.11: 64343197-64343197 on Build GRCh38
rs950031518 Chr.11: 64343445-64343445 on Build GRCh38
rs751953110 Chr.11: 64343556-64343561 on Build GRCh38
rs1032330644 Chr.11: 64343146-64343146 on Build GRCh38
rs920643337 Chr.11: 64343564-64343564 on Build GRCh38
rs905849291 Chr.11: 64343458-64343458 on Build GRCh38
rs78042412 Chr.11: 64343142-64343142 on Build GRCh38
rs980217140 Chr.11: 64343536-64343536 on Build GRCh38
rs778499620 Chr.11: 64343230-64343230 on Build GRCh38
rs747825888 Chr.11: 64343239-64343239 on Build GRCh38
rs539116396 Chr.11: 64343414-64343414 on Build GRCh38
rs1024338362 Chr.11: 64343483-64343483 on Build GRCh38
rs752956217 Chr.11: 64343185-64343185 on Build GRCh38
rs77179073 Chr.11: 64343154-64343154 on Build GRCh38
rs61354326 Chr.11: 64343483-64343483 on Build GRCh38
rs561918725 Chr.11: 64343324-64343324 on Build GRCh38

View Full Product Details