Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64726828-64727324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASGRP2
Entrez Gene ID
10235
Gene Name
RAS guanyl releasing protein 2
Gene Aliases
CALDAG-GEFI, CDC25L
UniGene
Hs.99491
Gene Chromosome Location
Chr.11: 64726911-64745456 on Build GRCh38
Transcripts
NM_001098670.1 NM_001098671.1 NM_001318398.1 NM_153819.1 XM_005273707.3 XM_011544718.1 XM_011544720.1 XM_011544721.1 XM_011544722.1 XM_011544723.2 XM_011544725.2 XM_017017082.1 XM_017017083.1 XM_017017084.1 XM_017017085.1 XM_017017086.1

SNP Detail

SNP ID Chromosome Location
rs1022812006 Chr.11: 64727200-64727200 on Build GRCh38
rs778056414 Chr.11: 64727263-64727263 on Build GRCh38
rs377037439 Chr.11: 64727271-64727271 on Build GRCh38
rs910397654 Chr.11: 64727165-64727165 on Build GRCh38
rs944288710 Chr.11: 64727049-64727049 on Build GRCh38
rs757282816 Chr.11: 64727039-64727039 on Build GRCh38
rs1043510462 Chr.11: 64727079-64727079 on Build GRCh38
rs973533732 Chr.11: 64726944-64726944 on Build GRCh38
rs983005772 Chr.11: 64727012-64727012 on Build GRCh38
rs758881316 Chr.11: 64727257-64727257 on Build GRCh38
rs753238712 Chr.11: 64727253-64727253 on Build GRCh38
rs768323185 Chr.11: 64727282-64727282 on Build GRCh38
rs565015334 Chr.11: 64726989-64726989 on Build GRCh38
rs950466021 Chr.11: 64727248-64727248 on Build GRCh38
rs933188435 Chr.11: 64726890-64726890 on Build GRCh38
rs561028200 Chr.11: 64727147-64727147 on Build GRCh38
rs180722704 Chr.11: 64727205-64727205 on Build GRCh38
rs113681913 Chr.11: 64727295-64727295 on Build GRCh38
rs1026450622 Chr.11: 64727222-64727222 on Build GRCh38
rs377165249 Chr.11: 64727051-64727051 on Build GRCh38
rs963521440 Chr.11: 64727202-64727202 on Build GRCh38
rs984964711 Chr.11: 64727061-64727061 on Build GRCh38
rs188830656 Chr.11: 64726956-64726956 on Build GRCh38
rs773976577 Chr.11: 64727288-64727288 on Build GRCh38
rs895759272 Chr.11: 64727151-64727151 on Build GRCh38
rs748839933 Chr.11: 64727280-64727280 on Build GRCh38
rs555842364 Chr.11: 64726902-64726902 on Build GRCh38
rs71581751 Chr.11: 64727136-64727136 on Build GRCh38
rs965626194 Chr.11: 64727274-64727274 on Build GRCh38
rs1013052690 Chr.11: 64727024-64727024 on Build GRCh38
rs962040558 Chr.11: 64727080-64727080 on Build GRCh38
rs576132458 Chr.11: 64726990-64726990 on Build GRCh38
rs903185101 Chr.11: 64727086-64727086 on Build GRCh38
rs923220450 Chr.11: 64727090-64727091 on Build GRCh38
rs3725 Chr.11: 64727009-64727009 on Build GRCh38
rs998790166 Chr.11: 64726885-64726885 on Build GRCh38
rs899984559 Chr.11: 64727058-64727058 on Build GRCh38
rs549472324 Chr.11: 64727246-64727246 on Build GRCh38
rs896038959 Chr.11: 64727260-64727260 on Build GRCh38
rs940593596 Chr.11: 64726864-64726864 on Build GRCh38
rs376152344 Chr.11: 64727250-64727250 on Build GRCh38
rs542273409 Chr.11: 64726891-64726891 on Build GRCh38
rs2230415 Chr.11: 64727064-64727064 on Build GRCh38
rs963190390 Chr.11: 64726927-64726927 on Build GRCh38
rs765761509 Chr.11: 64727252-64727252 on Build GRCh38
rs184154066 Chr.11: 64726854-64726854 on Build GRCh38
rs748159147 Chr.11: 64727299-64727299 on Build GRCh38
rs960195047 Chr.11: 64727015-64727015 on Build GRCh38
rs998821572 Chr.11: 64727134-64727134 on Build GRCh38
rs780958060 Chr.11: 64727249-64727249 on Build GRCh38
rs747393947 Chr.11: 64727264-64727264 on Build GRCh38
rs1057146091 Chr.11: 64727149-64727149 on Build GRCh38
rs1036388435 Chr.11: 64727014-64727014 on Build GRCh38
rs1012827460 Chr.11: 64727169-64727169 on Build GRCh38
rs1050180862 Chr.11: 64727165-64727165 on Build GRCh38
rs545003733 Chr.11: 64726967-64726967 on Build GRCh38
rs551781055 Chr.11: 64727251-64727251 on Build GRCh38
rs991360721 Chr.11: 64726893-64726893 on Build GRCh38
rs1036709318 Chr.11: 64726884-64726885 on Build GRCh38
rs71581750 Chr.11: 64726910-64726910 on Build GRCh38
rs923113982 Chr.11: 64726888-64726888 on Build GRCh38
rs755725025 Chr.11: 64727266-64727266 on Build GRCh38
rs1023148356 Chr.11: 64727047-64727047 on Build GRCh38
rs563017028 Chr.11: 64727247-64727247 on Build GRCh38
rs1035565984 Chr.11: 64726886-64726886 on Build GRCh38
rs1023202531 Chr.11: 64726926-64726926 on Build GRCh38

View Full Product Details