Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64898183-64898691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG2A
Entrez Gene ID
23130
Gene Name
autophagy related 2A
Gene Aliases
-
UniGene
Hs.370671
Gene Chromosome Location
Chr.11: 64894532-64917250 on Build GRCh38
Transcripts
NM_015104.2 XM_005273849.3 XM_005273850.3 XM_011544863.2 XM_011544864.2 XM_011544865.2 XM_011544866.2 XM_011544868.2 NR_106808.1

Gene Symbol
MIR6750
Entrez Gene ID
102466192
Gene Name
microRNA 6750
Gene Aliases
hsa-mir-6750
UniGene
Gene Chromosome Location
Chr.11: 64898363-64898437 on Build GRCh38
Transcripts
NM_015104.2 XM_005273849.3 XM_005273850.3 XM_011544863.2 XM_011544864.2 XM_011544865.2 XM_011544866.2 XM_011544868.2 NR_106808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1676163 c.4772A>G p.Q1591R Chr.11: 64898262-64898262 on Build GRCh38
309327 c.4639G>A p.E1547K Chr.11: 64898668-64898668 on Build GRCh38
4420533 c.4719G>A p.G1573G Chr.11: 64898315-64898315 on Build GRCh38
5469708 c.4652G>A p.R1551Q Chr.11: 64898655-64898655 on Build GRCh38
147268 c.4737C>T p.R1579R Chr.11: 64898297-64898297 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560740512 Chr.11: 64898546-64898546 on Build GRCh38
rs895115573 Chr.11: 64898377-64898377 on Build GRCh38
rs569233396 Chr.11: 64898270-64898270 on Build GRCh38
rs375562657 Chr.11: 64898399-64898399 on Build GRCh38
rs183090470 Chr.11: 64898302-64898302 on Build GRCh38
rs754187523 Chr.11: 64898632-64898632 on Build GRCh38
rs564802579 Chr.11: 64898536-64898536 on Build GRCh38
rs765382915 Chr.11: 64898304-64898304 on Build GRCh38
rs754692280 Chr.11: 64898337-64898337 on Build GRCh38
rs767770099 Chr.11: 64898332-64898332 on Build GRCh38
rs1005035455 Chr.11: 64898430-64898430 on Build GRCh38
rs553911523 Chr.11: 64898329-64898329 on Build GRCh38
rs757534256 Chr.11: 64898241-64898241 on Build GRCh38
rs750841015 Chr.11: 64898440-64898440 on Build GRCh38
rs772991379 Chr.11: 64898278-64898278 on Build GRCh38
rs761904968 Chr.11: 64898620-64898620 on Build GRCh38
rs753145099 Chr.11: 64898569-64898569 on Build GRCh38
rs139612405 Chr.11: 64898334-64898334 on Build GRCh38
rs1015388518 Chr.11: 64898441-64898441 on Build GRCh38
rs776977388 Chr.11: 64898298-64898298 on Build GRCh38
rs569193120 Chr.11: 64898658-64898658 on Build GRCh38
rs144122454 Chr.11: 64898343-64898343 on Build GRCh38
rs1005360342 Chr.11: 64898571-64898571 on Build GRCh38
rs772736982 Chr.11: 64898622-64898622 on Build GRCh38
rs139990915 Chr.11: 64898652-64898652 on Build GRCh38
rs766081579 Chr.11: 64898561-64898561 on Build GRCh38
rs762975865 Chr.11: 64898315-64898315 on Build GRCh38
rs756507844 Chr.11: 64898446-64898446 on Build GRCh38
rs540779238 Chr.11: 64898545-64898545 on Build GRCh38
rs768349863 Chr.11: 64898248-64898248 on Build GRCh38
rs61745749 Chr.11: 64898664-64898664 on Build GRCh38
rs796211953 Chr.11: 64898568-64898568 on Build GRCh38
rs575968970 Chr.11: 64898484-64898484 on Build GRCh38
rs985796603 Chr.11: 64898478-64898478 on Build GRCh38
rs967878294 Chr.11: 64898492-64898492 on Build GRCh38
rs527504613 Chr.11: 64898543-64898543 on Build GRCh38
rs1016831841 Chr.11: 64898285-64898285 on Build GRCh38
rs758771691 Chr.11: 64898572-64898572 on Build GRCh38
rs764570565 Chr.11: 64898419-64898440 on Build GRCh38
rs770832715 Chr.11: 64898354-64898354 on Build GRCh38
rs768857516 Chr.11: 64898600-64898600 on Build GRCh38
rs769559069 Chr.11: 64898587-64898587 on Build GRCh38
rs752384483 Chr.11: 64898451-64898451 on Build GRCh38
rs151054074 Chr.11: 64898487-64898487 on Build GRCh38
rs763751721 Chr.11: 64898653-64898653 on Build GRCh38
rs778441446 Chr.11: 64898591-64898591 on Build GRCh38
rs377220222 Chr.11: 64898367-64898367 on Build GRCh38
rs772069402 Chr.11: 64898277-64898277 on Build GRCh38
rs535914611 Chr.11: 64898464-64898464 on Build GRCh38
rs11824823 Chr.11: 64898476-64898476 on Build GRCh38
rs780317244 Chr.11: 64898589-64898589 on Build GRCh38
rs1054865505 Chr.11: 64898565-64898565 on Build GRCh38
rs868851769 Chr.11: 64898309-64898309 on Build GRCh38
rs199922623 Chr.11: 64898253-64898253 on Build GRCh38
rs757160704 Chr.11: 64898477-64898477 on Build GRCh38
rs766844831 Chr.11: 64898628-64898628 on Build GRCh38
rs768011302 Chr.11: 64898437-64898437 on Build GRCh38
rs901968744 Chr.11: 64898580-64898580 on Build GRCh38
rs774310030 Chr.11: 64898604-64898604 on Build GRCh38
rs762232844 Chr.11: 64898419-64898419 on Build GRCh38
rs746914035 Chr.11: 64898349-64898349 on Build GRCh38
rs939441859 Chr.11: 64898544-64898544 on Build GRCh38
rs751989812 Chr.11: 64898236-64898236 on Build GRCh38
rs775661966 Chr.11: 64898396-64898396 on Build GRCh38
rs145890967 Chr.11: 64898247-64898247 on Build GRCh38
rs375781359 Chr.11: 64898579-64898579 on Build GRCh38
rs1049364464 Chr.11: 64898570-64898570 on Build GRCh38
rs1042043404 Chr.11: 64898539-64898539 on Build GRCh38
rs764070708 Chr.11: 64898403-64898403 on Build GRCh38
rs977892635 Chr.11: 64898531-64898531 on Build GRCh38
rs534364576 Chr.11: 64898314-64898314 on Build GRCh38
rs778660253 Chr.11: 64898340-64898340 on Build GRCh38
rs936170792 Chr.11: 64898445-64898445 on Build GRCh38
rs142830078 Chr.11: 64898297-64898297 on Build GRCh38
rs955367605 Chr.11: 64898642-64898642 on Build GRCh38
rs146480656 Chr.11: 64898345-64898345 on Build GRCh38
rs751059416 Chr.11: 64898655-64898655 on Build GRCh38
rs374841480 Chr.11: 64898210-64898210 on Build GRCh38
rs777150488 Chr.11: 64898466-64898466 on Build GRCh38
rs998363615 Chr.11: 64898584-64898584 on Build GRCh38
rs773833263 Chr.11: 64898625-64898625 on Build GRCh38
rs202080937 Chr.11: 64898657-64898657 on Build GRCh38
rs745609823 Chr.11: 64898373-64898373 on Build GRCh38
rs778089034 Chr.11: 64898231-64898231 on Build GRCh38
rs750271831 Chr.11: 64898416-64898417 on Build GRCh38
rs761178485 Chr.11: 64898627-64898627 on Build GRCh38
rs770030093 Chr.11: 64898374-64898374 on Build GRCh38
rs774991545 Chr.11: 64898407-64898407 on Build GRCh38
rs770376438 Chr.11: 64898411-64898433 on Build GRCh38
rs760323027 Chr.11: 64898382-64898400 on Build GRCh38
rs776420309 Chr.11: 64898360-64898360 on Build GRCh38
rs765030979 Chr.11: 64898448-64898448 on Build GRCh38
rs757700302 Chr.11: 64898331-64898331 on Build GRCh38
rs148923613 Chr.11: 64898291-64898291 on Build GRCh38
rs763460649 Chr.11: 64898417-64898417 on Build GRCh38
rs370100424 Chr.11: 64898590-64898590 on Build GRCh38

View Full Product Details