Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 63559254-63559814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L2
Entrez Gene ID
283417
Gene Name
dpy-19 like 2
Gene Aliases
SPATA34, SPGF9
UniGene
Hs.533644
Gene Chromosome Location
Chr.12: 63558913-63669201 on Build GRCh38
Transcripts
NM_173812.4 XM_006719352.2 XM_011538215.2 XM_011538218.2 XM_017019188.1 XM_017019189.1 XM_017019190.1 XM_017019191.1 XM_017019192.1 XM_017019193.1 XM_017019194.1 XM_017019195.1 XM_017019196.1 XM_017019197.1 XM_017019198.1 XM_017019199.1 XM_017019201.1 XM_017019202.1 XM_017019203.1 XM_017019204.1 XM_017019205.1

SNP Detail

SNP ID Chromosome Location
rs12299121 Chr.12: 63559334-63559334 on Build GRCh38
rs117438826 Chr.12: 63559603-63559603 on Build GRCh38
rs555269465 Chr.12: 63559305-63559305 on Build GRCh38
rs12581178 Chr.12: 63559630-63559630 on Build GRCh38
rs530726122 Chr.12: 63559342-63559342 on Build GRCh38
rs752603241 Chr.12: 63559624-63559624 on Build GRCh38
rs767199410 Chr.12: 63559617-63559617 on Build GRCh38
rs1022543875 Chr.12: 63559463-63559463 on Build GRCh38
rs1033446303 Chr.12: 63559563-63559563 on Build GRCh38
rs12319634 Chr.12: 63559401-63559401 on Build GRCh38
rs573328985 Chr.12: 63559284-63559284 on Build GRCh38
rs1002332211 Chr.12: 63559544-63559544 on Build GRCh38
rs919809659 Chr.12: 63559667-63559667 on Build GRCh38
rs35605516 Chr.12: 63559522-63559523 on Build GRCh38
rs570100515 Chr.12: 63559702-63559702 on Build GRCh38
rs188890633 Chr.12: 63559788-63559788 on Build GRCh38
rs1818018 Chr.12: 63559293-63559293 on Build GRCh38
rs185730719 Chr.12: 63559412-63559412 on Build GRCh38
rs866482629 Chr.12: 63559663-63559663 on Build GRCh38
rs374084677 Chr.12: 63559773-63559773 on Build GRCh38
rs974051897 Chr.12: 63559634-63559634 on Build GRCh38
rs545726812 Chr.12: 63559467-63559467 on Build GRCh38
rs530036831 Chr.12: 63559668-63559668 on Build GRCh38
rs528408708 Chr.12: 63559596-63559596 on Build GRCh38
rs992304166 Chr.12: 63559588-63559588 on Build GRCh38
rs960457998 Chr.12: 63559583-63559583 on Build GRCh38
rs531208029 Chr.12: 63559564-63559564 on Build GRCh38
rs910651628 Chr.12: 63559706-63559706 on Build GRCh38
rs35691214 Chr.12: 63559507-63559508 on Build GRCh38
rs968214934 Chr.12: 63559472-63559472 on Build GRCh38
rs4086185 Chr.12: 63559275-63559275 on Build GRCh38
rs11175046 Chr.12: 63559777-63559777 on Build GRCh38
rs549720684 Chr.12: 63559689-63559689 on Build GRCh38
rs892451348 Chr.12: 63559440-63559440 on Build GRCh38
rs932479740 Chr.12: 63559362-63559362 on Build GRCh38
rs939000932 Chr.12: 63559282-63559282 on Build GRCh38
rs1040349194 Chr.12: 63559418-63559418 on Build GRCh38
rs892320747 Chr.12: 63559741-63559741 on Build GRCh38
rs559475596 Chr.12: 63559511-63559511 on Build GRCh38
rs1038675556 Chr.12: 63559336-63559336 on Build GRCh38
rs3905824 Chr.12: 63559339-63559339 on Build GRCh38
rs1353758 Chr.12: 63559340-63559340 on Build GRCh38
rs908467323 Chr.12: 63559329-63559329 on Build GRCh38
rs1049528685 Chr.12: 63559673-63559673 on Build GRCh38
rs1009858440 Chr.12: 63559448-63559448 on Build GRCh38
rs563382459 Chr.12: 63559407-63559407 on Build GRCh38

View Full Product Details