Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75402586-75403051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIPR1L2
Entrez Gene ID
144321
Gene Name
GLI pathogenesis-related 1 like 2
Gene Aliases
-
UniGene
Hs.406728
Gene Chromosome Location
Chr.12: 75391070-75432688 on Build GRCh38
Transcripts
NM_001270396.1 NM_152436.2 NR_072995.1 XM_011537949.2 XM_011537950.2 XM_017018848.1 XM_017018849.1

SNP Detail

SNP ID Chromosome Location
rs553052802 Chr.12: 75402955-75402955 on Build GRCh38
rs541480837 Chr.12: 75402628-75402628 on Build GRCh38
rs1041623119 Chr.12: 75402986-75402986 on Build GRCh38
rs975887477 Chr.12: 75402786-75402786 on Build GRCh38
rs377553410 Chr.12: 75402872-75402872 on Build GRCh38
rs548427266 Chr.12: 75402859-75402859 on Build GRCh38
rs752834506 Chr.12: 75403030-75403030 on Build GRCh38
rs1053223081 Chr.12: 75402690-75402690 on Build GRCh38
rs188802945 Chr.12: 75402945-75402945 on Build GRCh38
rs919113269 Chr.12: 75402787-75402788 on Build GRCh38
rs954065635 Chr.12: 75402716-75402716 on Build GRCh38
rs1022158060 Chr.12: 75402715-75402715 on Build GRCh38
rs1010120545 Chr.12: 75402672-75402672 on Build GRCh38
rs200080067 Chr.12: 75403002-75403002 on Build GRCh38
rs889200717 Chr.12: 75403008-75403008 on Build GRCh38
rs574623505 Chr.12: 75402931-75402931 on Build GRCh38
rs1007343216 Chr.12: 75402717-75402717 on Build GRCh38
rs561247209 Chr.12: 75402930-75402930 on Build GRCh38
rs1016179175 Chr.12: 75402791-75402791 on Build GRCh38
rs970900107 Chr.12: 75402844-75402844 on Build GRCh38
rs754909486 Chr.12: 75402727-75402727 on Build GRCh38
rs78158439 Chr.12: 75403023-75403024 on Build GRCh38
rs945960419 Chr.12: 75402977-75402977 on Build GRCh38
rs1011599664 Chr.12: 75402699-75402699 on Build GRCh38
rs962288284 Chr.12: 75402805-75402805 on Build GRCh38
rs972313142 Chr.12: 75402823-75402823 on Build GRCh38
rs369011452 Chr.12: 75402991-75402991 on Build GRCh38
rs984762944 Chr.12: 75402767-75402767 on Build GRCh38
rs1029061573 Chr.12: 75402762-75402762 on Build GRCh38
rs953419560 Chr.12: 75402763-75402763 on Build GRCh38
rs760174844 Chr.12: 75402982-75402982 on Build GRCh38
rs766348374 Chr.12: 75402944-75402944 on Build GRCh38
rs950552788 Chr.12: 75402803-75402803 on Build GRCh38
rs375670352 Chr.12: 75402664-75402664 on Build GRCh38
rs757590403 Chr.12: 75403019-75403019 on Build GRCh38
rs1020557379 Chr.12: 75402698-75402698 on Build GRCh38
rs927057429 Chr.12: 75402864-75402864 on Build GRCh38
rs532515999 Chr.12: 75402667-75402667 on Build GRCh38
rs74865917 Chr.12: 75402739-75402739 on Build GRCh38
rs1054410476 Chr.12: 75402654-75402654 on Build GRCh38
rs948409046 Chr.12: 75402911-75402911 on Build GRCh38
rs937041239 Chr.12: 75402885-75402885 on Build GRCh38
rs34954143 Chr.12: 75402803-75402804 on Build GRCh38
rs571385726 Chr.12: 75402832-75402832 on Build GRCh38
rs1053191986 Chr.12: 75402983-75402983 on Build GRCh38
rs901697595 Chr.12: 75402988-75402988 on Build GRCh38
rs879240323 Chr.12: 75402954-75402954 on Build GRCh38
rs7135440 Chr.12: 75402941-75402941 on Build GRCh38

View Full Product Details