Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52717673-52718230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMC6
Entrez Gene ID
5706
Gene Name
proteasome 26S subunit, ATPase 6
Gene Aliases
CADP44, HEL-S-73, P44, SUG2, p42
UniGene
Hs.156171
Gene Chromosome Location
Chr.14: 52707163-52727998 on Build GRCh38
Transcripts
NM_002806.3 XM_017021472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4501434 c.588C>T p.L196L Chr.14: 52718097-52718097 on Build GRCh38
433132 c.587T>G p.L196* Chr.14: 52718138-52718138 on Build GRCh38
4501838 c.599C>T p.A200V Chr.14: 52718108-52718108 on Build GRCh38
1152748 c.575C>G p.T192R Chr.14: 52718084-52718084 on Build GRCh38
1152749 c.634G>T p.V212F Chr.14: 52718229-52718229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12896994 Chr.14: 52717727-52717727 on Build GRCh38
rs144477145 Chr.14: 52717760-52717760 on Build GRCh38
rs904179882 Chr.14: 52717900-52717900 on Build GRCh38
rs930927685 Chr.14: 52717991-52717991 on Build GRCh38
rs969069320 Chr.14: 52717842-52717843 on Build GRCh38
rs766010190 Chr.14: 52718169-52718169 on Build GRCh38
rs372154274 Chr.14: 52718185-52718185 on Build GRCh38
rs368897330 Chr.14: 52718182-52718182 on Build GRCh38
rs1029811810 Chr.14: 52717989-52717989 on Build GRCh38
rs939443226 Chr.14: 52717837-52717837 on Build GRCh38
rs1042054209 Chr.14: 52717899-52717899 on Build GRCh38
rs190027142 Chr.14: 52717949-52717949 on Build GRCh38
rs370480651 Chr.14: 52718058-52718058 on Build GRCh38
rs1039372377 Chr.14: 52717946-52717946 on Build GRCh38
rs771473061 Chr.14: 52718104-52718104 on Build GRCh38
rs756305820 Chr.14: 52718195-52718195 on Build GRCh38
rs776597512 Chr.14: 52718164-52718164 on Build GRCh38
rs781404109 Chr.14: 52718065-52718065 on Build GRCh38
rs997948446 Chr.14: 52717984-52717984 on Build GRCh38
rs749784314 Chr.14: 52718094-52718094 on Build GRCh38
rs558630259 Chr.14: 52717745-52717745 on Build GRCh38
rs756217809 Chr.14: 52718084-52718084 on Build GRCh38
rs962739861 Chr.14: 52718050-52718050 on Build GRCh38
rs998315751 Chr.14: 52717739-52717739 on Build GRCh38
rs142536155 Chr.14: 52717861-52717861 on Build GRCh38
rs748193585 Chr.14: 52718073-52718073 on Build GRCh38
rs1010083052 Chr.14: 52717806-52717806 on Build GRCh38
rs1033695279 Chr.14: 52718062-52718062 on Build GRCh38
rs1050615478 Chr.14: 52718025-52718025 on Build GRCh38
rs374707958 Chr.14: 52718172-52718172 on Build GRCh38
rs752927237 Chr.14: 52718188-52718188 on Build GRCh38
rs554149569 Chr.14: 52717725-52717725 on Build GRCh38
rs559414697 Chr.14: 52717929-52717929 on Build GRCh38
rs753733156 Chr.14: 52718037-52718037 on Build GRCh38
rs963063612 Chr.14: 52717822-52717822 on Build GRCh38
rs945945504 Chr.14: 52717881-52717881 on Build GRCh38
rs529470921 Chr.14: 52717888-52717888 on Build GRCh38
rs200921247 Chr.14: 52718158-52718158 on Build GRCh38
rs899993182 Chr.14: 52717699-52717699 on Build GRCh38
rs771629284 Chr.14: 52718151-52718151 on Build GRCh38
rs764040663 Chr.14: 52718034-52718034 on Build GRCh38
rs960751301 Chr.14: 52718122-52718122 on Build GRCh38
rs778654482 Chr.14: 52718056-52718056 on Build GRCh38
rs148464335 Chr.14: 52717821-52717821 on Build GRCh38
rs926740002 Chr.14: 52717828-52717828 on Build GRCh38
rs528609919 Chr.14: 52717942-52717942 on Build GRCh38
rs755146611 Chr.14: 52718061-52718061 on Build GRCh38
rs889287814 Chr.14: 52718029-52718029 on Build GRCh38
rs775158903 Chr.14: 52718153-52718153 on Build GRCh38
rs1016918504 Chr.14: 52717762-52717762 on Build GRCh38
rs542889777 Chr.14: 52717889-52717889 on Build GRCh38
rs1029844290 Chr.14: 52717740-52717740 on Build GRCh38
rs866595005 Chr.14: 52718129-52718129 on Build GRCh38
rs899546560 Chr.14: 52717951-52717951 on Build GRCh38
rs373990232 Chr.14: 52718085-52718085 on Build GRCh38
rs992588380 Chr.14: 52717847-52717847 on Build GRCh38
rs951579178 Chr.14: 52717754-52717754 on Build GRCh38
rs1004456454 Chr.14: 52718032-52718032 on Build GRCh38
rs139274031 Chr.14: 52718097-52718097 on Build GRCh38
rs75620447 Chr.14: 52718030-52718030 on Build GRCh38
rs992996377 Chr.14: 52717943-52717943 on Build GRCh38
rs757083155 Chr.14: 52718043-52718043 on Build GRCh38
rs745600813 Chr.14: 52718109-52718109 on Build GRCh38

View Full Product Details