Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46581249-46581821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGF
Entrez Gene ID
5281
Gene Name
phosphatidylinositol glycan anchor biosynthesis class F
Gene Aliases
-
UniGene
Hs.468415
Gene Chromosome Location
Chr.2: 46581274-46617112 on Build GRCh38
Transcripts
NM_002643.3 NM_173074.2 XM_011532908.2 NM_012249.3 XM_005264229.2 XM_011532726.2 XM_011532728.2 XM_017003716.1

Gene Symbol
RHOQ
Entrez Gene ID
23433
Gene Name
ras homolog family member Q
Gene Aliases
ARHQ, HEL-S-42, RASL7A, TC10, TC10A
UniGene
Hs.709193
Gene Chromosome Location
Chr.2: 46542409-46584688 on Build GRCh38
Transcripts
NM_002643.3 NM_173074.2 XM_011532908.2 NM_012249.3 XM_005264229.2 XM_011532726.2 XM_011532728.2 XM_017003716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4411565 c.638A>C p.Q213P Chr.2: 46581500-46581500 on Build GRCh38
2153144 c.605T>C p.I202T Chr.2: 46581533-46581533 on Build GRCh38
4094405 c.606T>C p.I202I Chr.2: 46581532-46581532 on Build GRCh38
6373068 c.617G>T p.W206L Chr.2: 46581521-46581521 on Build GRCh38
3743810 c.601G>T p.V201F Chr.2: 46581537-46581537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746656805 Chr.2: 46581564-46581564 on Build GRCh38
rs936670450 Chr.2: 46581376-46581376 on Build GRCh38
rs769733236 Chr.2: 46581575-46581575 on Build GRCh38
rs776499331 Chr.2: 46581492-46581492 on Build GRCh38
rs144395549 Chr.2: 46581663-46581663 on Build GRCh38
rs770383518 Chr.2: 46581377-46581378 on Build GRCh38
rs1033305664 Chr.2: 46581769-46581769 on Build GRCh38
rs202122445 Chr.2: 46581364-46581366 on Build GRCh38
rs376861542 Chr.2: 46581487-46581487 on Build GRCh38
rs769778617 Chr.2: 46581494-46581494 on Build GRCh38
rs889864427 Chr.2: 46581379-46581379 on Build GRCh38
rs531181845 Chr.2: 46581683-46581683 on Build GRCh38
rs554944769 Chr.2: 46581428-46581428 on Build GRCh38
rs375902379 Chr.2: 46581477-46581477 on Build GRCh38
rs945238006 Chr.2: 46581355-46581355 on Build GRCh38
rs559692231 Chr.2: 46581520-46581520 on Build GRCh38
rs549383238 Chr.2: 46581695-46581695 on Build GRCh38
rs3088236 Chr.2: 46581421-46581421 on Build GRCh38
rs1002200899 Chr.2: 46581747-46581747 on Build GRCh38
rs565349060 Chr.2: 46581482-46581482 on Build GRCh38
rs764343770 Chr.2: 46581513-46581513 on Build GRCh38
rs775502314 Chr.2: 46581499-46581499 on Build GRCh38
rs1804166 Chr.2: 46581427-46581427 on Build GRCh38
rs754510182 Chr.2: 46581469-46581469 on Build GRCh38
rs936625334 Chr.2: 46581546-46581546 on Build GRCh38
rs577280268 Chr.2: 46581509-46581509 on Build GRCh38
rs777616861 Chr.2: 46581563-46581563 on Build GRCh38
rs576489985 Chr.2: 46581447-46581447 on Build GRCh38
rs182222238 Chr.2: 46581540-46581540 on Build GRCh38
rs750895869 Chr.2: 46581526-46581526 on Build GRCh38
rs903826517 Chr.2: 46581375-46581375 on Build GRCh38
rs756038604 Chr.2: 46581782-46581782 on Build GRCh38
rs186205049 Chr.2: 46581571-46581571 on Build GRCh38
rs747815125 Chr.2: 46581478-46581478 on Build GRCh38
rs532112767 Chr.2: 46581791-46581791 on Build GRCh38
rs780585821 Chr.2: 46581481-46581481 on Build GRCh38
rs764673920 Chr.2: 46581543-46581543 on Build GRCh38
rs781594998 Chr.2: 46581452-46581452 on Build GRCh38
rs1008299856 Chr.2: 46581384-46581384 on Build GRCh38
rs1019706064 Chr.2: 46581385-46581386 on Build GRCh38
rs745868978 Chr.2: 46581627-46581628 on Build GRCh38
rs897331078 Chr.2: 46581402-46581402 on Build GRCh38
rs1049744739 Chr.2: 46581550-46581550 on Build GRCh38
rs917765102 Chr.2: 46581338-46581338 on Build GRCh38
rs757950528 Chr.2: 46581562-46581562 on Build GRCh38
rs889793758 Chr.2: 46581566-46581566 on Build GRCh38
rs372737276 Chr.2: 46581441-46581441 on Build GRCh38
rs768928813 Chr.2: 46581720-46581720 on Build GRCh38
rs749422038 Chr.2: 46581588-46581588 on Build GRCh38
rs746839063 Chr.2: 46581484-46581484 on Build GRCh38
rs746415852 Chr.2: 46581332-46581332 on Build GRCh38
rs571281202 Chr.2: 46581774-46581774 on Build GRCh38
rs549897439 Chr.2: 46581417-46581417 on Build GRCh38
rs780985258 Chr.2: 46581572-46581572 on Build GRCh38
rs764468225 Chr.2: 46581435-46581435 on Build GRCh38
rs113300657 Chr.2: 46581762-46581762 on Build GRCh38
rs752212928 Chr.2: 46581547-46581547 on Build GRCh38
rs778739438 Chr.2: 46581628-46581628 on Build GRCh38
rs774619008 Chr.2: 46581611-46581611 on Build GRCh38
rs775377768 Chr.2: 46581580-46581580 on Build GRCh38
rs768705470 Chr.2: 46581604-46581604 on Build GRCh38
rs138221049 Chr.2: 46581493-46581493 on Build GRCh38
rs991909326 Chr.2: 46581501-46581501 on Build GRCh38
rs888696360 Chr.2: 46581710-46581710 on Build GRCh38
rs994329281 Chr.2: 46581685-46581685 on Build GRCh38
rs541572382 Chr.2: 46581519-46581519 on Build GRCh38
rs978355621 Chr.2: 46581539-46581539 on Build GRCh38

View Full Product Details