Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130182030-130182576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MZT2B
Entrez Gene ID
80097
Gene Name
mitotic spindle organizing protein 2B
Gene Aliases
FAM128B, MOZART2B
UniGene
Hs.469925
Gene Chromosome Location
Chr.2: 130181562-130204793 on Build GRCh38
Transcripts
NM_025029.3 XM_005263795.4 XM_011511886.2 XM_011511887.2 XM_011511888.2 XM_011511889.2 XM_011511890.2 NR_033230.2

Gene Symbol
SMPD4
Entrez Gene ID
55627
Gene Name
sphingomyelin phosphodiesterase 4
Gene Aliases
NET13, NSMASE-3, NSMASE3
UniGene
Hs.516450
Gene Chromosome Location
Chr.2: 130151392-130182750 on Build GRCh38
Transcripts
NM_025029.3 XM_005263795.4 XM_011511886.2 XM_011511887.2 XM_011511888.2 XM_011511889.2 XM_011511890.2 NR_033230.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3694872 c.83G>A p.R28Q Chr.2: 130182365-130182365 on Build GRCh38
4837379 c.138G>A p.A46A Chr.2: 130182420-130182420 on Build GRCh38
5959701 c.28C>A p.P10T Chr.2: 130182310-130182310 on Build GRCh38
7185581 c.99G>T p.L33L Chr.2: 130182381-130182381 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs547973996 Chr.2: 130182174-130182174 on Build GRCh38
rs974116130 Chr.2: 130182138-130182138 on Build GRCh38
rs975665706 Chr.2: 130182512-130182512 on Build GRCh38
rs1002147837 Chr.2: 130182399-130182399 on Build GRCh38
rs759973768 Chr.2: 130182395-130182395 on Build GRCh38
rs532735406 Chr.2: 130182072-130182072 on Build GRCh38
rs762322582 Chr.2: 130182438-130182438 on Build GRCh38
rs958787716 Chr.2: 130182112-130182112 on Build GRCh38
rs779799298 Chr.2: 130182328-130182328 on Build GRCh38
rs184959654 Chr.2: 130182162-130182162 on Build GRCh38
rs555482859 Chr.2: 130182270-130182270 on Build GRCh38
rs373183158 Chr.2: 130182529-130182529 on Build GRCh38
rs951501884 Chr.2: 130182417-130182417 on Build GRCh38
rs1053439538 Chr.2: 130182189-130182189 on Build GRCh38
rs756951425 Chr.2: 130182290-130182290 on Build GRCh38
rs970992576 Chr.2: 130182078-130182078 on Build GRCh38
rs533602859 Chr.2: 130182263-130182263 on Build GRCh38
rs989600911 Chr.2: 130182542-130182542 on Build GRCh38
rs1046072524 Chr.2: 130182352-130182352 on Build GRCh38
rs1801954 Chr.2: 130182431-130182431 on Build GRCh38
rs544924845 Chr.2: 130182507-130182507 on Build GRCh38
rs537876745 Chr.2: 130182330-130182330 on Build GRCh38
rs1046785131 Chr.2: 130182204-130182204 on Build GRCh38
rs28709423 Chr.2: 130182151-130182151 on Build GRCh38
rs775986505 Chr.2: 130182409-130182409 on Build GRCh38
rs751489091 Chr.2: 130182463-130182463 on Build GRCh38
rs965659023 Chr.2: 130182286-130182286 on Build GRCh38
rs752484390 Chr.2: 130182489-130182489 on Build GRCh38
rs567150159 Chr.2: 130182288-130182288 on Build GRCh38
rs963185277 Chr.2: 130182232-130182232 on Build GRCh38
rs771041012 Chr.2: 130182312-130182312 on Build GRCh38
rs566281698 Chr.2: 130182213-130182213 on Build GRCh38
rs779097320 Chr.2: 130182363-130182363 on Build GRCh38
rs867821484 Chr.2: 130182271-130182271 on Build GRCh38
rs749256285 Chr.2: 130182367-130182367 on Build GRCh38
rs376233184 Chr.2: 130182482-130182482 on Build GRCh38
rs995606062 Chr.2: 130182249-130182250 on Build GRCh38
rs1009910925 Chr.2: 130182076-130182076 on Build GRCh38
rs754885687 Chr.2: 130182481-130182481 on Build GRCh38
rs758360579 Chr.2: 130182325-130182325 on Build GRCh38
rs376076371 Chr.2: 130182400-130182400 on Build GRCh38
rs983461661 Chr.2: 130182154-130182154 on Build GRCh38
rs543028546 Chr.2: 130182257-130182257 on Build GRCh38
rs892106548 Chr.2: 130182060-130182060 on Build GRCh38
rs958239867 Chr.2: 130182521-130182521 on Build GRCh38
rs1034322350 Chr.2: 130182058-130182058 on Build GRCh38
rs780932617 Chr.2: 130182342-130182342 on Build GRCh38
rs1039879877 Chr.2: 130182170-130182170 on Build GRCh38
rs982792739 Chr.2: 130182269-130182269 on Build GRCh38
rs892161865 Chr.2: 130182194-130182194 on Build GRCh38
rs17017149 Chr.2: 130182414-130182414 on Build GRCh38
rs1009218858 Chr.2: 130182201-130182201 on Build GRCh38
rs749964413 Chr.2: 130182435-130182435 on Build GRCh38
rs748417678 Chr.2: 130182343-130182343 on Build GRCh38
rs1013819826 Chr.2: 130182467-130182467 on Build GRCh38
rs1033518060 Chr.2: 130182231-130182231 on Build GRCh38
rs906154657 Chr.2: 130182378-130182378 on Build GRCh38
rs945151036 Chr.2: 130182553-130182553 on Build GRCh38
rs912809360 Chr.2: 130182160-130182160 on Build GRCh38
rs774668981 Chr.2: 130182377-130182377 on Build GRCh38
rs913652784 Chr.2: 130182336-130182336 on Build GRCh38
rs746845252 Chr.2: 130182329-130182329 on Build GRCh38
rs764902134 Chr.2: 130182303-130182303 on Build GRCh38
rs201954757 Chr.2: 130182444-130182444 on Build GRCh38
rs556026147 Chr.2: 130182346-130182346 on Build GRCh38
rs1024880056 Chr.2: 130182077-130182077 on Build GRCh38
rs771478812 Chr.2: 130182369-130182369 on Build GRCh38
rs1053492819 Chr.2: 130182326-130182326 on Build GRCh38
rs756329572 Chr.2: 130182503-130182503 on Build GRCh38
rs1034882780 Chr.2: 130182102-130182102 on Build GRCh38
rs749902042 Chr.2: 130182324-130182324 on Build GRCh38
rs541351058 Chr.2: 130182124-130182124 on Build GRCh38
rs951499574 Chr.2: 130182264-130182264 on Build GRCh38
rs1000536099 Chr.2: 130182056-130182056 on Build GRCh38
rs1968675 Chr.2: 130182517-130182517 on Build GRCh38
rs1023905606 Chr.2: 130182490-130182490 on Build GRCh38
rs914039012 Chr.2: 130182546-130182547 on Build GRCh38
rs1027104218 Chr.2: 130182415-130182415 on Build GRCh38
rs919945210 Chr.2: 130182140-130182142 on Build GRCh38
rs879176358 Chr.2: 130182221-130182221 on Build GRCh38
rs936442265 Chr.2: 130182187-130182187 on Build GRCh38
rs980628084 Chr.2: 130182086-130182086 on Build GRCh38
rs773409032 Chr.2: 130182358-130182358 on Build GRCh38
rs373338046 Chr.2: 130182368-130182368 on Build GRCh38

View Full Product Details