Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201620310-201620883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM237
Entrez Gene ID
65062
Gene Name
transmembrane protein 237
Gene Aliases
ALS2CR4, JBTS14
UniGene
Hs.12319
Gene Chromosome Location
Chr.2: 201620184-201643529 on Build GRCh38
Transcripts
NM_001044385.2 NM_152388.3

SNP Detail

SNP ID Chromosome Location
rs762112690 Chr.2: 201620837-201620837 on Build GRCh38
rs934378758 Chr.2: 201620643-201620643 on Build GRCh38
rs985605049 Chr.2: 201620414-201620414 on Build GRCh38
rs991355461 Chr.2: 201620598-201620598 on Build GRCh38
rs143218667 Chr.2: 201620743-201620743 on Build GRCh38
rs991696362 Chr.2: 201620792-201620792 on Build GRCh38
rs34097188 Chr.2: 201620541-201620542 on Build GRCh38
rs950004949 Chr.2: 201620629-201620629 on Build GRCh38
rs904581147 Chr.2: 201620751-201620751 on Build GRCh38
rs761856568 Chr.2: 201620558-201620558 on Build GRCh38
rs578251760 Chr.2: 201620618-201620618 on Build GRCh38
rs900078116 Chr.2: 201620720-201620720 on Build GRCh38
rs753823056 Chr.2: 201620841-201620841 on Build GRCh38
rs747771285 Chr.2: 201620370-201620370 on Build GRCh38
rs779673601 Chr.2: 201620470-201620472 on Build GRCh38
rs1029600326 Chr.2: 201620734-201620734 on Build GRCh38
rs555315551 Chr.2: 201620527-201620527 on Build GRCh38
rs1029654424 Chr.2: 201620543-201620543 on Build GRCh38
rs1010694232 Chr.2: 201620363-201620363 on Build GRCh38
rs545570415 Chr.2: 201620639-201620639 on Build GRCh38
rs1052303255 Chr.2: 201620687-201620687 on Build GRCh38
rs1044848896 Chr.2: 201620640-201620640 on Build GRCh38
rs998487230 Chr.2: 201620724-201620724 on Build GRCh38
rs754036495 Chr.2: 201620833-201620833 on Build GRCh38
rs1006990324 Chr.2: 201620801-201620801 on Build GRCh38
rs982853617 Chr.2: 201620546-201620546 on Build GRCh38
rs909598569 Chr.2: 201620559-201620559 on Build GRCh38
rs1045415426 Chr.2: 201620633-201620633 on Build GRCh38
rs892413784 Chr.2: 201620705-201620705 on Build GRCh38
rs563749617 Chr.2: 201620738-201620738 on Build GRCh38
rs769560427 Chr.2: 201620426-201620426 on Build GRCh38
rs946836407 Chr.2: 201620671-201620671 on Build GRCh38
rs1018920082 Chr.2: 201620444-201620445 on Build GRCh38
rs962834387 Chr.2: 201620568-201620568 on Build GRCh38
rs760789868 Chr.2: 201620749-201620749 on Build GRCh38
rs573709537 Chr.2: 201620454-201620454 on Build GRCh38
rs1054234265 Chr.2: 201620666-201620666 on Build GRCh38
rs1127982 Chr.2: 201620540-201620540 on Build GRCh38
rs187328136 Chr.2: 201620400-201620400 on Build GRCh38
rs1004184063 Chr.2: 201620742-201620742 on Build GRCh38
rs766334593 Chr.2: 201620623-201620623 on Build GRCh38
rs892906964 Chr.2: 201620667-201620667 on Build GRCh38
rs965588691 Chr.2: 201620431-201620431 on Build GRCh38
rs759959195 Chr.2: 201620549-201620549 on Build GRCh38
rs77771615 Chr.2: 201620802-201620802 on Build GRCh38
rs906364781 Chr.2: 201620641-201620641 on Build GRCh38
rs1021706586 Chr.2: 201620398-201620398 on Build GRCh38
rs964332397 Chr.2: 201620472-201620472 on Build GRCh38
rs180747501 Chr.2: 201620349-201620349 on Build GRCh38
rs939212469 Chr.2: 201620678-201620678 on Build GRCh38
rs774570708 Chr.2: 201620499-201620499 on Build GRCh38
rs764235944 Chr.2: 201620782-201620782 on Build GRCh38
rs553740095 Chr.2: 201620569-201620569 on Build GRCh38
rs984226773 Chr.2: 201620539-201620539 on Build GRCh38
rs886055436 Chr.2: 201620742-201620742 on Build GRCh38

View Full Product Details