Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232407824-232408371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPPL2
Entrez Gene ID
251
Gene Name
alkaline phosphatase, placental like 2
Gene Aliases
ALPG, ALPPL, GCAP
UniGene
Hs.333509
Gene Chromosome Location
Chr.2: 232406842-232410714 on Build GRCh38
Transcripts
NM_031313.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1244871 c.506G>C p.R169P Chr.2: 232407875-232407875 on Build GRCh38
5396476 c.506G>A p.R169Q Chr.2: 232407875-232407875 on Build GRCh38
720248 c.742G>T p.D248Y Chr.2: 232408360-232408360 on Build GRCh38
4092383 c.593G>A p.R198H Chr.2: 232407962-232407962 on Build GRCh38
6598882 c.499A>G p.T167A Chr.2: 232407868-232407868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199598336 Chr.2: 232407946-232407946 on Build GRCh38
rs764423561 Chr.2: 232408067-232408067 on Build GRCh38
rs41265119 Chr.2: 232407957-232407957 on Build GRCh38
rs777876889 Chr.2: 232408029-232408029 on Build GRCh38
rs76299057 Chr.2: 232408232-232408232 on Build GRCh38
rs763405335 Chr.2: 232408284-232408284 on Build GRCh38
rs781312811 Chr.2: 232408350-232408350 on Build GRCh38
rs369229942 Chr.2: 232408238-232408238 on Build GRCh38
rs758750585 Chr.2: 232407997-232407997 on Build GRCh38
rs145557709 Chr.2: 232408017-232408017 on Build GRCh38
rs753201676 Chr.2: 232408049-232408049 on Build GRCh38
rs779124895 Chr.2: 232408300-232408301 on Build GRCh38
rs766289475 Chr.2: 232407874-232407875 on Build GRCh38
rs980041019 Chr.2: 232408155-232408155 on Build GRCh38
rs74503304 Chr.2: 232408135-232408135 on Build GRCh38
rs1044917113 Chr.2: 232407882-232407882 on Build GRCh38
rs1048987 Chr.2: 232407987-232407987 on Build GRCh38
rs751480816 Chr.2: 232407933-232407935 on Build GRCh38
rs769557579 Chr.2: 232408019-232408019 on Build GRCh38
rs746637543 Chr.2: 232408007-232408007 on Build GRCh38
rs542023570 Chr.2: 232407962-232407962 on Build GRCh38
rs769658713 Chr.2: 232407985-232407985 on Build GRCh38
rs377074419 Chr.2: 232408241-232408241 on Build GRCh38
rs768537631 Chr.2: 232408028-232408028 on Build GRCh38
rs751728680 Chr.2: 232407910-232407910 on Build GRCh38
rs535365122 Chr.2: 232407918-232407918 on Build GRCh38
rs754266714 Chr.2: 232407958-232407958 on Build GRCh38
rs746584152 Chr.2: 232407967-232407967 on Build GRCh38
rs769836426 Chr.2: 232408275-232408275 on Build GRCh38
rs754390834 Chr.2: 232407991-232407991 on Build GRCh38
rs746199307 Chr.2: 232408301-232408302 on Build GRCh38
rs766512279 Chr.2: 232408032-232408032 on Build GRCh38
rs368457694 Chr.2: 232407943-232407943 on Build GRCh38
rs1039322093 Chr.2: 232408042-232408042 on Build GRCh38
rs563461413 Chr.2: 232408108-232408108 on Build GRCh38
rs549626071 Chr.2: 232407905-232407905 on Build GRCh38
rs370089099 Chr.2: 232408148-232408148 on Build GRCh38
rs576271845 Chr.2: 232407956-232407956 on Build GRCh38
rs374652374 Chr.2: 232407875-232407875 on Build GRCh38
rs758805699 Chr.2: 232408063-232408063 on Build GRCh38
rs758606866 Chr.2: 232407870-232407870 on Build GRCh38
rs926547595 Chr.2: 232408158-232408158 on Build GRCh38
rs751808520 Chr.2: 232408293-232408293 on Build GRCh38
rs529459697 Chr.2: 232407901-232407901 on Build GRCh38
rs559940812 Chr.2: 232408139-232408139 on Build GRCh38
rs374249849 Chr.2: 232407933-232407933 on Build GRCh38
rs200632383 Chr.2: 232407921-232407921 on Build GRCh38
rs759602594 Chr.2: 232408273-232408273 on Build GRCh38
rs141105098 Chr.2: 232408312-232408312 on Build GRCh38
rs771594260 Chr.2: 232407935-232407935 on Build GRCh38
rs375289027 Chr.2: 232407984-232407984 on Build GRCh38
rs374388513 Chr.2: 232407911-232407911 on Build GRCh38
rs559437356 Chr.2: 232408228-232408228 on Build GRCh38
rs182567124 Chr.2: 232408227-232408227 on Build GRCh38
rs200884165 Chr.2: 232407850-232407850 on Build GRCh38
rs764443675 Chr.2: 232408286-232408286 on Build GRCh38
rs775552786 Chr.2: 232408020-232408020 on Build GRCh38
rs764528787 Chr.2: 232408074-232408074 on Build GRCh38
rs533246706 Chr.2: 232407881-232407881 on Build GRCh38
rs761377449 Chr.2: 232407897-232407897 on Build GRCh38
rs374167777 Chr.2: 232408311-232408311 on Build GRCh38
rs553481936 Chr.2: 232407923-232407923 on Build GRCh38
rs751251153 Chr.2: 232408216-232408216 on Build GRCh38
rs375443589 Chr.2: 232408047-232408047 on Build GRCh38
rs564544259 Chr.2: 232407876-232407876 on Build GRCh38
rs370073633 Chr.2: 232407903-232407903 on Build GRCh38
rs757537353 Chr.2: 232408066-232408067 on Build GRCh38
rs539457044 Chr.2: 232408218-232408218 on Build GRCh38
rs375906740 Chr.2: 232408234-232408234 on Build GRCh38
rs749719597 Chr.2: 232407922-232407922 on Build GRCh38
rs762221650 Chr.2: 232408298-232408298 on Build GRCh38
rs766272646 Chr.2: 232407961-232407961 on Build GRCh38
rs145892470 Chr.2: 232407907-232407907 on Build GRCh38
rs775880810 Chr.2: 232407884-232407884 on Build GRCh38
rs367963870 Chr.2: 232407914-232407914 on Build GRCh38
rs747056715 Chr.2: 232407874-232407874 on Build GRCh38
rs748826261 Chr.2: 232408200-232408200 on Build GRCh38
rs549298350 Chr.2: 232408138-232408138 on Build GRCh38
rs755941363 Chr.2: 232407866-232407866 on Build GRCh38
rs185167361 Chr.2: 232407898-232407898 on Build GRCh38
rs370897967 Chr.2: 232408021-232408021 on Build GRCh38
rs2678518 Chr.2: 232408215-232408215 on Build GRCh38
rs529376144 Chr.2: 232408132-232408132 on Build GRCh38
rs752886217 Chr.2: 232407867-232407867 on Build GRCh38
rs770347716 Chr.2: 232407969-232407969 on Build GRCh38
rs760104321 Chr.2: 232408043-232408043 on Build GRCh38
rs993241826 Chr.2: 232407976-232407976 on Build GRCh38
rs1032187342 Chr.2: 232407953-232407953 on Build GRCh38
rs75080031 Chr.2: 232408308-232408308 on Build GRCh38
rs542674135 Chr.2: 232408149-232408149 on Build GRCh38
rs780524913 Chr.2: 232408010-232408010 on Build GRCh38
rs75920311 Chr.2: 232408301-232408301 on Build GRCh38
rs773616900 Chr.2: 232408029-232408029 on Build GRCh38
rs1014287293 Chr.2: 232408131-232408131 on Build GRCh38
rs377502379 Chr.2: 232407879-232407879 on Build GRCh38
rs767138507 Chr.2: 232408300-232408300 on Build GRCh38
rs776900149 Chr.2: 232408249-232408249 on Build GRCh38
rs772670586 Chr.2: 232407936-232407936 on Build GRCh38
rs369419190 Chr.2: 232408285-232408285 on Build GRCh38
rs776672251 Chr.2: 232407979-232407979 on Build GRCh38
rs756185433 Chr.2: 232408012-232408012 on Build GRCh38
rs753003550 Chr.2: 232407994-232407994 on Build GRCh38
rs776509295 Chr.2: 232408037-232408037 on Build GRCh38
rs754042839 Chr.2: 232408051-232408052 on Build GRCh38
rs973682863 Chr.2: 232408209-232408209 on Build GRCh38
rs776422889 Chr.2: 232407941-232407941 on Build GRCh38
rs1031802152 Chr.2: 232408098-232408098 on Build GRCh38
rs756844703 Chr.2: 232408009-232408009 on Build GRCh38
rs963285467 Chr.2: 232408166-232408166 on Build GRCh38
rs370845315 Chr.2: 232407916-232407916 on Build GRCh38
rs765590125 Chr.2: 232407992-232407992 on Build GRCh38
rs146780985 Chr.2: 232407906-232407906 on Build GRCh38
rs140999297 Chr.2: 232407891-232407891 on Build GRCh38
rs113898433 Chr.2: 232407885-232407885 on Build GRCh38
rs368869875 Chr.2: 232407989-232407989 on Build GRCh38
rs1048986 Chr.2: 232407939-232407939 on Build GRCh38
rs778665902 Chr.2: 232407926-232407926 on Build GRCh38
rs777278704 Chr.2: 232407965-232407965 on Build GRCh38
rs148210569 Chr.2: 232407942-232407942 on Build GRCh38
rs552055226 Chr.2: 232408206-232408206 on Build GRCh38
rs549828948 Chr.2: 232408219-232408219 on Build GRCh38
rs776029863 Chr.2: 232408276-232408276 on Build GRCh38
rs777723971 Chr.2: 232407872-232407872 on Build GRCh38
rs151276345 Chr.2: 232407990-232407990 on Build GRCh38
rs749214306 Chr.2: 232408233-232408233 on Build GRCh38
rs956359473 Chr.2: 232408106-232408106 on Build GRCh38
rs771001719 Chr.2: 232408245-232408245 on Build GRCh38

View Full Product Details