Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34957823-34958187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH7B
Entrez Gene ID
57644
Gene Name
myosin heavy chain 7B
Gene Aliases
MHC14, MYH14
UniGene
Hs.414122
Gene Chromosome Location
Chr.20: 34955835-35002437 on Build GRCh38
Transcripts
NM_020884.4

SNP Detail

SNP ID Chromosome Location
rs955050959 Chr.20: 34957885-34957885 on Build GRCh38
rs185911800 Chr.20: 34957920-34957920 on Build GRCh38
rs773392177 Chr.20: 34958145-34958145 on Build GRCh38
rs369389160 Chr.20: 34958006-34958006 on Build GRCh38
rs74516190 Chr.20: 34958115-34958115 on Build GRCh38
rs147461850 Chr.20: 34958104-34958104 on Build GRCh38
rs958263408 Chr.20: 34957863-34957863 on Build GRCh38
rs181026903 Chr.20: 34957871-34957871 on Build GRCh38
rs879249431 Chr.20: 34957964-34957964 on Build GRCh38
rs150702675 Chr.20: 34957870-34957870 on Build GRCh38
rs972796147 Chr.20: 34957925-34957925 on Build GRCh38
rs757981507 Chr.20: 34958085-34958085 on Build GRCh38
rs550322266 Chr.20: 34957884-34957884 on Build GRCh38
rs139992834 Chr.20: 34958059-34958059 on Build GRCh38
rs995889616 Chr.20: 34957846-34957846 on Build GRCh38
rs971881819 Chr.20: 34957992-34957992 on Build GRCh38
rs368027479 Chr.20: 34957901-34957901 on Build GRCh38
rs3761143 Chr.20: 34957942-34957942 on Build GRCh38
rs539025545 Chr.20: 34957929-34957929 on Build GRCh38
rs376669796 Chr.20: 34958034-34958034 on Build GRCh38
rs935914228 Chr.20: 34958037-34958037 on Build GRCh38
rs6120768 Chr.20: 34958138-34958138 on Build GRCh38
rs546772876 Chr.20: 34957859-34957859 on Build GRCh38
rs1017071286 Chr.20: 34957880-34957880 on Build GRCh38
rs3761142 Chr.20: 34958013-34958013 on Build GRCh38
rs144580687 Chr.20: 34958101-34958101 on Build GRCh38
rs556615369 Chr.20: 34958123-34958123 on Build GRCh38
rs913080227 Chr.20: 34958098-34958098 on Build GRCh38

View Full Product Details