Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112541691-112542245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG3
Entrez Gene ID
64422
Gene Name
autophagy related 3
Gene Aliases
APG3, APG3-LIKE, APG3L, PC3-96
UniGene
Hs.477126
Gene Chromosome Location
Chr.3: 112532507-112561963 on Build GRCh38
Transcripts
NM_001278712.1 NM_022488.4 XM_011513074.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4820259 c.439G>A p.E147K Chr.3: 112541839-112541839 on Build GRCh38
4835341 c.419C>T p.S140L Chr.3: 112541859-112541859 on Build GRCh38
6535097 c.439G>C p.E147Q Chr.3: 112541839-112541839 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs879781902 Chr.3: 112541971-112541971 on Build GRCh38
rs67129931 Chr.3: 112542128-112542129 on Build GRCh38
rs556488573 Chr.3: 112542118-112542118 on Build GRCh38
rs148750516 Chr.3: 112542127-112542128 on Build GRCh38
rs768616191 Chr.3: 112541829-112541829 on Build GRCh38
rs1022378033 Chr.3: 112542187-112542187 on Build GRCh38
rs532003344 Chr.3: 112542002-112542002 on Build GRCh38
rs1027501211 Chr.3: 112541797-112541797 on Build GRCh38
rs956781828 Chr.3: 112542148-112542148 on Build GRCh38
rs189016423 Chr.3: 112541957-112541957 on Build GRCh38
rs771338345 Chr.3: 112541851-112541851 on Build GRCh38
rs3736269 Chr.3: 112541724-112541724 on Build GRCh38
rs760008866 Chr.3: 112541864-112541864 on Build GRCh38
rs932896566 Chr.3: 112541955-112541955 on Build GRCh38
rs919603292 Chr.3: 112542193-112542193 on Build GRCh38
rs11547153 Chr.3: 112541845-112541845 on Build GRCh38
rs915420157 Chr.3: 112542005-112542005 on Build GRCh38
rs527512699 Chr.3: 112541766-112541766 on Build GRCh38
rs762216951 Chr.3: 112542125-112542125 on Build GRCh38
rs756701603 Chr.3: 112541929-112541929 on Build GRCh38
rs968131849 Chr.3: 112542105-112542105 on Build GRCh38
rs569564481 Chr.3: 112541991-112541991 on Build GRCh38
rs1044211316 Chr.3: 112542046-112542046 on Build GRCh38
rs780806164 Chr.3: 112541791-112541791 on Build GRCh38
rs989636222 Chr.3: 112541989-112541989 on Build GRCh38
rs773205906 Chr.3: 112541837-112541837 on Build GRCh38
rs571851460 Chr.3: 112542014-112542014 on Build GRCh38
rs954362370 Chr.3: 112542124-112542124 on Build GRCh38
rs759920822 Chr.3: 112541761-112541761 on Build GRCh38
rs115634667 Chr.3: 112542064-112542064 on Build GRCh38
rs753104505 Chr.3: 112541768-112541768 on Build GRCh38
rs769514914 Chr.3: 112541794-112541794 on Build GRCh38
rs948269740 Chr.3: 112542016-112542017 on Build GRCh38
rs987462556 Chr.3: 112541968-112541968 on Build GRCh38
rs534437607 Chr.3: 112542017-112542017 on Build GRCh38
rs896481451 Chr.3: 112542075-112542075 on Build GRCh38
rs776924183 Chr.3: 112541852-112541852 on Build GRCh38
rs777193209 Chr.3: 112541775-112541775 on Build GRCh38
rs769250954 Chr.3: 112541841-112541841 on Build GRCh38
rs1031067366 Chr.3: 112541945-112541945 on Build GRCh38
rs373183146 Chr.3: 112541958-112541958 on Build GRCh38
rs1021027742 Chr.3: 112541900-112541900 on Build GRCh38
rs757947590 Chr.3: 112541769-112541769 on Build GRCh38
rs772352883 Chr.3: 112542133-112542133 on Build GRCh38
rs970032106 Chr.3: 112541742-112541742 on Build GRCh38
rs181732985 Chr.3: 112542006-112542006 on Build GRCh38
rs1036023175 Chr.3: 112542066-112542066 on Build GRCh38
rs749134942 Chr.3: 112541819-112541819 on Build GRCh38
rs1020975461 Chr.3: 112542100-112542100 on Build GRCh38
rs1003662547 Chr.3: 112541751-112541751 on Build GRCh38
rs912199502 Chr.3: 112541986-112541986 on Build GRCh38
rs1007432250 Chr.3: 112542114-112542114 on Build GRCh38
rs939647364 Chr.3: 112542144-112542144 on Build GRCh38
rs760834509 Chr.3: 112541847-112541847 on Build GRCh38
rs779777927 Chr.3: 112541947-112541948 on Build GRCh38
rs989528917 Chr.3: 112542169-112542169 on Build GRCh38
rs931128743 Chr.3: 112542041-112542041 on Build GRCh38
rs972418829 Chr.3: 112542166-112542166 on Build GRCh38
rs77882834 Chr.3: 112541786-112541786 on Build GRCh38
rs775897386 Chr.3: 112541917-112541917 on Build GRCh38
rs1028452927 Chr.3: 112542119-112542119 on Build GRCh38
rs931057310 Chr.3: 112542217-112542217 on Build GRCh38
rs72940020 Chr.3: 112541777-112541777 on Build GRCh38
rs773415470 Chr.3: 112542204-112542204 on Build GRCh38
rs765470192 Chr.3: 112541914-112541914 on Build GRCh38
rs60800484 Chr.3: 112542130-112542131 on Build GRCh38
rs141140717 Chr.3: 112541878-112541878 on Build GRCh38
rs762242572 Chr.3: 112541824-112541829 on Build GRCh38
rs145149591 Chr.3: 112542053-112542053 on Build GRCh38
rs111838719 Chr.3: 112541733-112541733 on Build GRCh38
rs1009562014 Chr.3: 112542078-112542078 on Build GRCh38
rs780074991 Chr.3: 112541817-112541817 on Build GRCh38
rs1036454043 Chr.3: 112542047-112542047 on Build GRCh38
rs775043631 Chr.3: 112541863-112541863 on Build GRCh38
rs201402926 Chr.3: 112541922-112541922 on Build GRCh38
rs536357422 Chr.3: 112542089-112542089 on Build GRCh38
rs111364855 Chr.3: 112542073-112542073 on Build GRCh38
rs751086230 Chr.3: 112541926-112541926 on Build GRCh38
rs888949159 Chr.3: 112542113-112542113 on Build GRCh38
rs765842095 Chr.3: 112541763-112541763 on Build GRCh38
rs921501769 Chr.3: 112541938-112541938 on Build GRCh38
rs957026019 Chr.3: 112541946-112541946 on Build GRCh38

View Full Product Details