Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 114234589-114235187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF80
Entrez Gene ID
7634
Gene Name
zinc finger protein 80
Gene Aliases
pT17
UniGene
Hs.271079
Gene Chromosome Location
Chr.3: 114211035-114237578 on Build GRCh38
Transcripts
NM_007136.3 XM_017007132.1 XM_017007133.1 XM_017007134.1 XM_017007135.1 XM_017007136.1

SNP Detail

SNP ID Chromosome Location
rs1029444055 Chr.3: 114234902-114234902 on Build GRCh38
rs188285817 Chr.3: 114234933-114234933 on Build GRCh38
rs192210819 Chr.3: 114234811-114234811 on Build GRCh38
rs1021069772 Chr.3: 114234842-114234842 on Build GRCh38
rs9832338 Chr.3: 114235024-114235024 on Build GRCh38
rs962621814 Chr.3: 114234948-114234948 on Build GRCh38
rs938210568 Chr.3: 114234982-114234982 on Build GRCh38
rs972682906 Chr.3: 114234975-114234975 on Build GRCh38
rs140797356 Chr.3: 114235046-114235046 on Build GRCh38
rs993374184 Chr.3: 114234655-114234655 on Build GRCh38
rs569213006 Chr.3: 114234696-114234696 on Build GRCh38
rs187716421 Chr.3: 114234670-114234670 on Build GRCh38
rs967212915 Chr.3: 114234854-114234854 on Build GRCh38
rs915576686 Chr.3: 114235007-114235007 on Build GRCh38
rs758757939 Chr.3: 114235087-114235087 on Build GRCh38
rs941727541 Chr.3: 114235009-114235009 on Build GRCh38
rs929064208 Chr.3: 114235025-114235025 on Build GRCh38
rs1007432402 Chr.3: 114234689-114234689 on Build GRCh38
rs778545103 Chr.3: 114235097-114235097 on Build GRCh38
rs1010442066 Chr.3: 114235140-114235140 on Build GRCh38
rs73235932 Chr.3: 114234633-114234633 on Build GRCh38
rs1021394518 Chr.3: 114235121-114235121 on Build GRCh38
rs953988938 Chr.3: 114234930-114234930 on Build GRCh38
rs374705358 Chr.3: 114234694-114234694 on Build GRCh38
rs542687964 Chr.3: 114235116-114235116 on Build GRCh38
rs182930856 Chr.3: 114234831-114234831 on Build GRCh38
rs1055095225 Chr.3: 114234644-114234644 on Build GRCh38
rs559077851 Chr.3: 114234993-114234993 on Build GRCh38
rs909756379 Chr.3: 114234940-114234946 on Build GRCh38
rs548462109 Chr.3: 114234814-114234814 on Build GRCh38
rs958071702 Chr.3: 114234792-114234792 on Build GRCh38
rs890346784 Chr.3: 114234688-114234688 on Build GRCh38
rs1010497376 Chr.3: 114234733-114234733 on Build GRCh38
rs147279518 Chr.3: 114235014-114235014 on Build GRCh38
rs1017523378 Chr.3: 114234736-114234736 on Build GRCh38
rs928120145 Chr.3: 114234981-114234981 on Build GRCh38
rs890293967 Chr.3: 114235058-114235058 on Build GRCh38
rs1007422481 Chr.3: 114235068-114235068 on Build GRCh38
rs982279627 Chr.3: 114234980-114234980 on Build GRCh38
rs1047046 Chr.3: 114235005-114235005 on Build GRCh38
rs756642944 Chr.3: 114234894-114234894 on Build GRCh38
rs1047048 Chr.3: 114234875-114234875 on Build GRCh38
rs893788026 Chr.3: 114235089-114235089 on Build GRCh38
rs997884135 Chr.3: 114234880-114234880 on Build GRCh38
rs73235934 Chr.3: 114235080-114235080 on Build GRCh38

View Full Product Details