Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117203675-117204176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAGLN
Entrez Gene ID
6876
Gene Name
transgelin
Gene Aliases
SM22, SMCC, TAGLN1, WS3-10
UniGene
Hs.410977
Gene Chromosome Location
Chr.11: 117199324-117204792 on Build GRCh38
Transcripts
NM_001001522.1 NM_003186.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4018130 c.446C>T p.P149L Chr.11: 117203869-117203869 on Build GRCh38
5459874 c.360C>A p.G120G Chr.11: 117203783-117203783 on Build GRCh38
6796492 c.426T>C p.D142D Chr.11: 117203849-117203849 on Build GRCh38
6796493 c.373G>A p.A125T Chr.11: 117203796-117203796 on Build GRCh38
6361884 c.374C>T p.A125V Chr.11: 117203797-117203797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975267354 Chr.11: 117203808-117203808 on Build GRCh38
rs563850243 Chr.11: 117203833-117203833 on Build GRCh38
rs531321247 Chr.11: 117203766-117203766 on Build GRCh38
rs763372587 Chr.11: 117203893-117203893 on Build GRCh38
rs576790308 Chr.11: 117203859-117203859 on Build GRCh38
rs1017341010 Chr.11: 117204073-117204073 on Build GRCh38
rs112576509 Chr.11: 117203886-117203886 on Build GRCh38
rs530159694 Chr.11: 117204050-117204050 on Build GRCh38
rs113278237 Chr.11: 117204109-117204109 on Build GRCh38
rs995188221 Chr.11: 117203951-117203951 on Build GRCh38
rs370263391 Chr.11: 117203813-117203813 on Build GRCh38
rs572360016 Chr.11: 117203915-117203915 on Build GRCh38
rs761539047 Chr.11: 117203758-117203758 on Build GRCh38
rs760090961 Chr.11: 117204085-117204085 on Build GRCh38
rs1038337521 Chr.11: 117203952-117203952 on Build GRCh38
rs771398083 Chr.11: 117203912-117203912 on Build GRCh38
rs747751212 Chr.11: 117203884-117203884 on Build GRCh38
rs531348646 Chr.11: 117204098-117204099 on Build GRCh38
rs767035120 Chr.11: 117203779-117203779 on Build GRCh38
rs760369893 Chr.11: 117204062-117204062 on Build GRCh38
rs768643861 Chr.11: 117203809-117203809 on Build GRCh38
rs749248950 Chr.11: 117203709-117203709 on Build GRCh38
rs1052972000 Chr.11: 117203749-117203749 on Build GRCh38
rs7358337 Chr.11: 117203888-117203888 on Build GRCh38
rs774888711 Chr.11: 117203918-117203919 on Build GRCh38
rs773552572 Chr.11: 117204037-117204037 on Build GRCh38
rs147514901 Chr.11: 117204044-117204044 on Build GRCh38
rs1025334203 Chr.11: 117203997-117203997 on Build GRCh38
rs989160679 Chr.11: 117204140-117204140 on Build GRCh38
rs754966865 Chr.11: 117203797-117203797 on Build GRCh38
rs776097926 Chr.11: 117203751-117203751 on Build GRCh38
rs773681954 Chr.11: 117203892-117203892 on Build GRCh38
rs752884318 Chr.11: 117203932-117203932 on Build GRCh38
rs894716788 Chr.11: 117203878-117203878 on Build GRCh38
rs951092948 Chr.11: 117203711-117203711 on Build GRCh38
rs1021445594 Chr.11: 117204084-117204084 on Build GRCh38
rs780758296 Chr.11: 117203881-117203881 on Build GRCh38
rs778689195 Chr.11: 117203735-117203735 on Build GRCh38
rs772051996 Chr.11: 117203739-117203739 on Build GRCh38
rs765631160 Chr.11: 117203760-117203760 on Build GRCh38
rs774954222 Chr.11: 117203742-117203742 on Build GRCh38
rs745707933 Chr.11: 117203825-117203825 on Build GRCh38
rs774666734 Chr.11: 117203907-117203907 on Build GRCh38
rs758299291 Chr.11: 117203858-117203858 on Build GRCh38
rs995217611 Chr.11: 117203700-117203700 on Build GRCh38
rs374464235 Chr.11: 117203860-117203860 on Build GRCh38
rs964345638 Chr.11: 117203791-117203791 on Build GRCh38
rs938806424 Chr.11: 117204114-117204114 on Build GRCh38
rs771877408 Chr.11: 117203848-117203848 on Build GRCh38
rs994796638 Chr.11: 117203992-117203992 on Build GRCh38
rs886881594 Chr.11: 117204037-117204037 on Build GRCh38
rs1005565131 Chr.11: 117203947-117203947 on Build GRCh38
rs912461525 Chr.11: 117203756-117203756 on Build GRCh38
rs367640314 Chr.11: 117203872-117203872 on Build GRCh38
rs778662789 Chr.11: 117203818-117203818 on Build GRCh38
rs45508091 Chr.11: 117203810-117203810 on Build GRCh38
rs773771927 Chr.11: 117203766-117203766 on Build GRCh38
rs897422799 Chr.11: 117203975-117203975 on Build GRCh38
rs1025239944 Chr.11: 117203708-117203708 on Build GRCh38
rs199711558 Chr.11: 117203914-117203914 on Build GRCh38
rs975331581 Chr.11: 117204075-117204075 on Build GRCh38
rs200244971 Chr.11: 117203738-117203738 on Build GRCh38
rs992806417 Chr.11: 117204149-117204149 on Build GRCh38
rs749604981 Chr.11: 117204055-117204055 on Build GRCh38
rs905940055 Chr.11: 117203920-117203920 on Build GRCh38
rs978389056 Chr.11: 117203720-117203720 on Build GRCh38
rs763532557 Chr.11: 117203767-117203767 on Build GRCh38
rs138833123 Chr.11: 117203743-117203743 on Build GRCh38
rs963960066 Chr.11: 117204065-117204065 on Build GRCh38
rs1029898958 Chr.11: 117204022-117204022 on Build GRCh38
rs771446473 Chr.11: 117203901-117203901 on Build GRCh38
rs955720582 Chr.11: 117204132-117204132 on Build GRCh38
rs541077152 Chr.11: 117204034-117204034 on Build GRCh38
rs543191586 Chr.11: 117204094-117204094 on Build GRCh38
rs752140448 Chr.11: 117203796-117203796 on Build GRCh38
rs1007890261 Chr.11: 117204051-117204051 on Build GRCh38
rs866416189 Chr.11: 117204071-117204071 on Build GRCh38
rs190406453 Chr.11: 117204099-117204099 on Build GRCh38

View Full Product Details