Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117820517-117821016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD2
Entrez Gene ID
486
Gene Name
FXYD domain containing ion transport regulator 2
Gene Aliases
ATP1G1, HOMG2
UniGene
Hs.731865
Gene Chromosome Location
Chr.11: 117820075-117828092 on Build GRCh38
Transcripts
NM_001680.4 NM_021603.3 NM_001204268.1 NM_001243598.2

Gene Symbol
FXYD6-FXYD2
Entrez Gene ID
100533181
Gene Name
FXYD6-FXYD2 readthrough
Gene Aliases
-
UniGene
Hs.744850
Gene Chromosome Location
Chr.11: 117820075-117877031 on Build GRCh38
Transcripts
NM_001680.4 NM_021603.3 NM_001204268.1 NM_001243598.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1492453 c.162G>A p.K54K Chr.11: 117820867-117820867 on Build GRCh38
123285 c.144C>G p.F48L Chr.11: 117820885-117820885 on Build GRCh38
5837892 c.193_194CC>TT p.P65F Chr.11: 117820696-117820697 on Build GRCh38
5837891 c.171-1_171GG>AA p.? Chr.11: 117820696-117820697 on Build GRCh38
5999630 c.151C>A p.L51I Chr.11: 117820654-117820654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980846458 Chr.11: 117820803-117820803 on Build GRCh38
rs752608951 Chr.11: 117820625-117820625 on Build GRCh38
rs376646131 Chr.11: 117820782-117820782 on Build GRCh38
rs200472357 Chr.11: 117820856-117820856 on Build GRCh38
rs139816783 Chr.11: 117820859-117820859 on Build GRCh38
rs143620414 Chr.11: 117820875-117820875 on Build GRCh38
rs1044799074 Chr.11: 117820687-117820687 on Build GRCh38
rs548124426 Chr.11: 117820925-117820925 on Build GRCh38
rs570509942 Chr.11: 117820890-117820890 on Build GRCh38
rs760465825 Chr.11: 117820686-117820686 on Build GRCh38
rs747313056 Chr.11: 117820812-117820812 on Build GRCh38
rs755359872 Chr.11: 117820883-117820883 on Build GRCh38
rs370529634 Chr.11: 117820836-117820836 on Build GRCh38
rs897902495 Chr.11: 117820645-117820645 on Build GRCh38
rs371153018 Chr.11: 117820665-117820665 on Build GRCh38
rs1044857010 Chr.11: 117820903-117820903 on Build GRCh38
rs760155230 Chr.11: 117820583-117820583 on Build GRCh38
rs766075198 Chr.11: 117820874-117820874 on Build GRCh38
rs750019905 Chr.11: 117820867-117820867 on Build GRCh38
rs202175753 Chr.11: 117820640-117820640 on Build GRCh38
rs768603606 Chr.11: 117820675-117820675 on Build GRCh38
rs753277223 Chr.11: 117820638-117820638 on Build GRCh38
rs964084113 Chr.11: 117820616-117820616 on Build GRCh38
rs769735680 Chr.11: 117820837-117820837 on Build GRCh38
rs763081279 Chr.11: 117820841-117820841 on Build GRCh38
rs748585940 Chr.11: 117820889-117820889 on Build GRCh38
rs180728680 Chr.11: 117820666-117820666 on Build GRCh38
rs761860028 Chr.11: 117820862-117820862 on Build GRCh38
rs774518680 Chr.11: 117820929-117820929 on Build GRCh38
rs764643788 Chr.11: 117820863-117820863 on Build GRCh38
rs369137285 Chr.11: 117820805-117820805 on Build GRCh38
rs768418823 Chr.11: 117820947-117820947 on Build GRCh38
rs114805473 Chr.11: 117820548-117820548 on Build GRCh38
rs771019695 Chr.11: 117820926-117820926 on Build GRCh38
rs758131389 Chr.11: 117820709-117820709 on Build GRCh38
rs761748059 Chr.11: 117820676-117820676 on Build GRCh38
rs11827585 Chr.11: 117820660-117820660 on Build GRCh38
rs201566468 Chr.11: 117820605-117820605 on Build GRCh38
rs912669306 Chr.11: 117820590-117820590 on Build GRCh38
rs536273709 Chr.11: 117820558-117820558 on Build GRCh38
rs149353193 Chr.11: 117820675-117820675 on Build GRCh38
rs779775215 Chr.11: 117820636-117820637 on Build GRCh38
rs548244596 Chr.11: 117820780-117820780 on Build GRCh38
rs781036200 Chr.11: 117820677-117820678 on Build GRCh38
rs756100911 Chr.11: 117820630-117820630 on Build GRCh38
rs760414736 Chr.11: 117820854-117820854 on Build GRCh38
rs749331252 Chr.11: 117820917-117820917 on Build GRCh38
rs1047662375 Chr.11: 117820948-117820948 on Build GRCh38
rs565529111 Chr.11: 117820684-117820684 on Build GRCh38
rs1045293871 Chr.11: 117820609-117820609 on Build GRCh38
rs996071296 Chr.11: 117820719-117820719 on Build GRCh38
rs1008452019 Chr.11: 117820982-117820982 on Build GRCh38
rs368170451 Chr.11: 117820853-117820853 on Build GRCh38
rs371615390 Chr.11: 117820688-117820688 on Build GRCh38
rs778383106 Chr.11: 117820651-117820651 on Build GRCh38
rs118032332 Chr.11: 117820637-117820637 on Build GRCh38
rs373698796 Chr.11: 117820852-117820852 on Build GRCh38
rs914604787 Chr.11: 117820807-117820807 on Build GRCh38
rs746130582 Chr.11: 117820938-117820938 on Build GRCh38
rs1019585185 Chr.11: 117820763-117820763 on Build GRCh38
rs74443383 Chr.11: 117820892-117820892 on Build GRCh38
rs909464998 Chr.11: 117820634-117820634 on Build GRCh38
rs1026283974 Chr.11: 117820569-117820569 on Build GRCh38
rs951698484 Chr.11: 117820748-117820748 on Build GRCh38
rs780565124 Chr.11: 117820728-117820728 on Build GRCh38
rs370215240 Chr.11: 117820681-117820681 on Build GRCh38
rs200063683 Chr.11: 117820884-117820884 on Build GRCh38
rs951447976 Chr.11: 117820570-117820570 on Build GRCh38
rs552097922 Chr.11: 117820680-117820680 on Build GRCh38
rs773271804 Chr.11: 117820679-117820679 on Build GRCh38
rs746837735 Chr.11: 117820670-117820672 on Build GRCh38
rs559192490 Chr.11: 117820565-117820565 on Build GRCh38
rs748243358 Chr.11: 117820678-117820679 on Build GRCh38
rs376994807 Chr.11: 117820788-117820788 on Build GRCh38
rs969148284 Chr.11: 117820981-117820981 on Build GRCh38
rs1025934997 Chr.11: 117820735-117820735 on Build GRCh38
rs201669990 Chr.11: 117820690-117820690 on Build GRCh38
rs77988721 Chr.11: 117820966-117820966 on Build GRCh38
rs1024640567 Chr.11: 117820978-117820978 on Build GRCh38
rs887254090 Chr.11: 117820964-117820964 on Build GRCh38
rs769162778 Chr.11: 117820827-117820827 on Build GRCh38
rs778525146 Chr.11: 117820702-117820702 on Build GRCh38
rs750026725 Chr.11: 117820707-117820707 on Build GRCh38
rs947743282 Chr.11: 117820582-117820582 on Build GRCh38
rs756684075 Chr.11: 117820700-117820700 on Build GRCh38
rs1008211630 Chr.11: 117820752-117820752 on Build GRCh38
rs995776616 Chr.11: 117820560-117820560 on Build GRCh38
rs144519777 Chr.11: 117820678-117820678 on Build GRCh38
rs942308194 Chr.11: 117820808-117820808 on Build GRCh38
rs869789 Chr.11: 117820671-117820671 on Build GRCh38
rs892270070 Chr.11: 117820942-117820942 on Build GRCh38
rs768330106 Chr.11: 117820561-117820561 on Build GRCh38
rs979671121 Chr.11: 117820829-117820829 on Build GRCh38
rs374310766 Chr.11: 117820872-117820872 on Build GRCh38

View Full Product Details