Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118589950-118590464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARCN1
Entrez Gene ID
372
Gene Name
archain 1
Gene Aliases
COPD
UniGene
Hs.33642
Gene Chromosome Location
Chr.11: 118572363-118603033 on Build GRCh38
Transcripts
NM_001142281.1 NM_001655.4 XM_005271542.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7186424 c.826A>G p.M276V Chr.11: 118590348-118590348 on Build GRCh38
6251727 c.931G>T p.G311C Chr.11: 118590453-118590453 on Build GRCh38
6602821 c.935G>A p.R312Q Chr.11: 118590457-118590457 on Build GRCh38
1245301 c.895G>A p.G299S Chr.11: 118590417-118590417 on Build GRCh38
6842075 c.893A>G p.H298R Chr.11: 118590415-118590415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573036183 Chr.11: 118590183-118590183 on Build GRCh38
rs182062396 Chr.11: 118590291-118590291 on Build GRCh38
rs782438302 Chr.11: 118589980-118589980 on Build GRCh38
rs782036972 Chr.11: 118590378-118590378 on Build GRCh38
rs781854385 Chr.11: 118590431-118590432 on Build GRCh38
rs782790694 Chr.11: 118590377-118590377 on Build GRCh38
rs1038435797 Chr.11: 118590040-118590040 on Build GRCh38
rs929752119 Chr.11: 118590280-118590280 on Build GRCh38
rs748079967 Chr.11: 118590415-118590415 on Build GRCh38
rs576568798 Chr.11: 118590097-118590097 on Build GRCh38
rs147118477 Chr.11: 118590390-118590390 on Build GRCh38
rs558433334 Chr.11: 118590248-118590248 on Build GRCh38
rs955180491 Chr.11: 118590092-118590092 on Build GRCh38
rs782223696 Chr.11: 118590337-118590337 on Build GRCh38
rs916870191 Chr.11: 118590197-118590197 on Build GRCh38
rs530117599 Chr.11: 118590313-118590313 on Build GRCh38
rs1006779798 Chr.11: 118590098-118590098 on Build GRCh38
rs149105341 Chr.11: 118590146-118590146 on Build GRCh38
rs782384548 Chr.11: 118590330-118590330 on Build GRCh38
rs782701863 Chr.11: 118590243-118590243 on Build GRCh38
rs375565238 Chr.11: 118590425-118590425 on Build GRCh38
rs11604765 Chr.11: 118590282-118590282 on Build GRCh38
rs941589257 Chr.11: 118590215-118590215 on Build GRCh38
rs45600633 Chr.11: 118590242-118590242 on Build GRCh38
rs568843209 Chr.11: 118590121-118590121 on Build GRCh38
rs928088209 Chr.11: 118589979-118589979 on Build GRCh38
rs782351933 Chr.11: 118590112-118590112 on Build GRCh38
rs939526119 Chr.11: 118590153-118590153 on Build GRCh38
rs782777693 Chr.11: 118590385-118590385 on Build GRCh38
rs782587618 Chr.11: 118590308-118590308 on Build GRCh38
rs576801697 Chr.11: 118590275-118590275 on Build GRCh38
rs781923327 Chr.11: 118590346-118590346 on Build GRCh38
rs1051263015 Chr.11: 118590286-118590286 on Build GRCh38
rs782665421 Chr.11: 118590435-118590436 on Build GRCh38
rs187397714 Chr.11: 118589982-118589982 on Build GRCh38
rs990974824 Chr.11: 118590182-118590182 on Build GRCh38
rs966828130 Chr.11: 118590137-118590137 on Build GRCh38
rs782367570 Chr.11: 118590342-118590342 on Build GRCh38
rs782687261 Chr.11: 118590290-118590290 on Build GRCh38
rs782140700 Chr.11: 118590088-118590088 on Build GRCh38
rs782009352 Chr.11: 118590333-118590333 on Build GRCh38
rs782424970 Chr.11: 118590307-118590307 on Build GRCh38
rs554465593 Chr.11: 118590171-118590171 on Build GRCh38
rs782109325 Chr.11: 118590382-118590382 on Build GRCh38
rs782156810 Chr.11: 118590419-118590419 on Build GRCh38
rs782601824 Chr.11: 118590300-118590300 on Build GRCh38
rs36123609 Chr.11: 118590266-118590267 on Build GRCh38
rs371011063 Chr.11: 118590389-118590389 on Build GRCh38
rs896940963 Chr.11: 118590046-118590046 on Build GRCh38
rs782225025 Chr.11: 118590306-118590306 on Build GRCh38
rs562936680 Chr.11: 118590295-118590295 on Build GRCh38
rs781905158 Chr.11: 118590438-118590439 on Build GRCh38
rs190936162 Chr.11: 118590030-118590030 on Build GRCh38
rs895384392 Chr.11: 118590004-118590004 on Build GRCh38
rs781883417 Chr.11: 118590270-118590270 on Build GRCh38
rs928138067 Chr.11: 118590152-118590152 on Build GRCh38
rs994281200 Chr.11: 118590087-118590087 on Build GRCh38
rs782775362 Chr.11: 118590426-118590426 on Build GRCh38
rs782723334 Chr.11: 118590430-118590431 on Build GRCh38
rs782520198 Chr.11: 118590433-118590434 on Build GRCh38
rs557058820 Chr.11: 118590106-118590106 on Build GRCh38

View Full Product Details