Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118894900-118895408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCR5
Entrez Gene ID
643
Gene Name
C-X-C motif chemokine receptor 5
Gene Aliases
BLR1, CD185, MDR15
UniGene
Hs.113916
Gene Chromosome Location
Chr.11: 118883766-118896271 on Build GRCh38
Transcripts
NM_001716.4 NM_032966.2

SNP Detail

SNP ID Chromosome Location
rs897564682 Chr.11: 118894926-118894926 on Build GRCh38
rs909463805 Chr.11: 118895132-118895132 on Build GRCh38
rs564032177 Chr.11: 118895379-118895379 on Build GRCh38
rs947327029 Chr.11: 118895019-118895019 on Build GRCh38
rs897264229 Chr.11: 118895136-118895136 on Build GRCh38
rs576432190 Chr.11: 118895180-118895180 on Build GRCh38
rs139379344 Chr.11: 118894931-118894931 on Build GRCh38
rs761464449 Chr.11: 118895074-118895074 on Build GRCh38
rs1002149963 Chr.11: 118895332-118895332 on Build GRCh38
rs888759522 Chr.11: 118894939-118894939 on Build GRCh38
rs559969564 Chr.11: 118895276-118895276 on Build GRCh38
rs145657278 Chr.11: 118895064-118895064 on Build GRCh38
rs1025911486 Chr.11: 118895239-118895239 on Build GRCh38
rs766377467 Chr.11: 118895310-118895310 on Build GRCh38
rs975099764 Chr.11: 118895301-118895301 on Build GRCh38
rs955031194 Chr.11: 118895377-118895377 on Build GRCh38
rs942322502 Chr.11: 118894970-118894970 on Build GRCh38
rs1051482896 Chr.11: 118895174-118895174 on Build GRCh38
rs529973297 Chr.11: 118895157-118895157 on Build GRCh38
rs3741331 Chr.11: 118895347-118895347 on Build GRCh38
rs919380756 Chr.11: 118895166-118895166 on Build GRCh38
rs969334125 Chr.11: 118895081-118895081 on Build GRCh38
rs896590757 Chr.11: 118895161-118895161 on Build GRCh38
rs972700770 Chr.11: 118895355-118895355 on Build GRCh38
rs942456576 Chr.11: 118895293-118895293 on Build GRCh38
rs773120545 Chr.11: 118895256-118895256 on Build GRCh38
rs185448133 Chr.11: 118895117-118895117 on Build GRCh38
rs770156751 Chr.11: 118895012-118895012 on Build GRCh38
rs902983468 Chr.11: 118895124-118895124 on Build GRCh38
rs974693159 Chr.11: 118895200-118895200 on Build GRCh38
rs981020202 Chr.11: 118895128-118895128 on Build GRCh38
rs1039800772 Chr.11: 118894974-118894974 on Build GRCh38
rs910247993 Chr.11: 118895179-118895179 on Build GRCh38
rs676925 Chr.11: 118894979-118894979 on Build GRCh38
rs919445499 Chr.11: 118895358-118895358 on Build GRCh38
rs56115103 Chr.11: 118895079-118895080 on Build GRCh38
rs557977987 Chr.11: 118895167-118895167 on Build GRCh38
rs541774674 Chr.11: 118895242-118895242 on Build GRCh38
rs193171551 Chr.11: 118894923-118894923 on Build GRCh38
rs1016182835 Chr.11: 118895343-118895343 on Build GRCh38
rs1053881 Chr.11: 118894951-118894951 on Build GRCh38
rs1002668499 Chr.11: 118895133-118895133 on Build GRCh38
rs535403087 Chr.11: 118895044-118895044 on Build GRCh38
rs527714704 Chr.11: 118894940-118894940 on Build GRCh38
rs111605944 Chr.11: 118895184-118895184 on Build GRCh38
rs986515855 Chr.11: 118894930-118894930 on Build GRCh38
rs999145658 Chr.11: 118895244-118895244 on Build GRCh38
rs768177800 Chr.11: 118895009-118895009 on Build GRCh38
rs776062385 Chr.11: 118895013-118895013 on Build GRCh38
rs986988066 Chr.11: 118895258-118895258 on Build GRCh38
rs569165734 Chr.11: 118895287-118895287 on Build GRCh38
rs373474342 Chr.11: 118895171-118895172 on Build GRCh38
rs567913719 Chr.11: 118895032-118895032 on Build GRCh38
rs45627033 Chr.11: 118895309-118895309 on Build GRCh38
rs56387466 Chr.11: 118895181-118895181 on Build GRCh38
rs904381267 Chr.11: 118895000-118895000 on Build GRCh38
rs1035039152 Chr.11: 118895158-118895158 on Build GRCh38
rs914263137 Chr.11: 118895302-118895302 on Build GRCh38
rs539783663 Chr.11: 118895119-118895119 on Build GRCh38
rs574268285 Chr.11: 118895237-118895237 on Build GRCh38
rs933028969 Chr.11: 118894926-118894926 on Build GRCh38

View Full Product Details