Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 120660222-120660714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIK4
Entrez Gene ID
2900
Gene Name
glutamate ionotropic receptor kainate type subunit 4
Gene Aliases
EAA1, GRIK, GluK4, KA1
UniGene
Hs.538738
Gene Chromosome Location
Chr.11: 120511746-120988805 on Build GRCh38
Transcripts
NM_001282470.2 NM_001282473.2 NM_014619.4 XM_011542784.1 XM_017017621.1 XM_017017622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
320707 c.58T>G p.C20G Chr.11: 120660376-120660376 on Build GRCh38
6067838 c.35C>T p.P12L Chr.11: 120660353-120660353 on Build GRCh38
6482474 c.69C>A p.H23Q Chr.11: 120660387-120660387 on Build GRCh38
4018632 c.8G>A p.R3H Chr.11: 120660326-120660326 on Build GRCh38
3791267 c.14C>T p.S5L Chr.11: 120660332-120660332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138617238 Chr.11: 120660332-120660332 on Build GRCh38
rs912321087 Chr.11: 120660593-120660593 on Build GRCh38
rs549685234 Chr.11: 120660607-120660607 on Build GRCh38
rs921997242 Chr.11: 120660247-120660247 on Build GRCh38
rs771779942 Chr.11: 120660280-120660280 on Build GRCh38
rs369721630 Chr.11: 120660400-120660400 on Build GRCh38
rs548988998 Chr.11: 120660290-120660290 on Build GRCh38
rs778805683 Chr.11: 120660274-120660274 on Build GRCh38
rs563510929 Chr.11: 120660539-120660539 on Build GRCh38
rs372852246 Chr.11: 120660257-120660257 on Build GRCh38
rs6589829 Chr.11: 120660264-120660264 on Build GRCh38
rs758306234 Chr.11: 120660437-120660437 on Build GRCh38
rs987522612 Chr.11: 120660415-120660415 on Build GRCh38
rs145979642 Chr.11: 120660366-120660366 on Build GRCh38
rs1046791697 Chr.11: 120660686-120660687 on Build GRCh38
rs370031223 Chr.11: 120660356-120660356 on Build GRCh38
rs754252446 Chr.11: 120660453-120660453 on Build GRCh38
rs200560956 Chr.11: 120660413-120660413 on Build GRCh38
rs201814702 Chr.11: 120660389-120660389 on Build GRCh38
rs953755036 Chr.11: 120660566-120660566 on Build GRCh38
rs1043088053 Chr.11: 120660553-120660553 on Build GRCh38
rs970190258 Chr.11: 120660344-120660344 on Build GRCh38
rs1001652982 Chr.11: 120660582-120660582 on Build GRCh38
rs943870139 Chr.11: 120660501-120660501 on Build GRCh38
rs936962192 Chr.11: 120660384-120660384 on Build GRCh38
rs116098730 Chr.11: 120660387-120660387 on Build GRCh38
rs905536147 Chr.11: 120660561-120660561 on Build GRCh38
rs573619024 Chr.11: 120660472-120660472 on Build GRCh38
rs1035437745 Chr.11: 120660599-120660599 on Build GRCh38
rs777233245 Chr.11: 120660313-120660313 on Build GRCh38
rs768044489 Chr.11: 120660293-120660293 on Build GRCh38
rs977807190 Chr.11: 120660609-120660609 on Build GRCh38
rs750379066 Chr.11: 120660425-120660425 on Build GRCh38
rs147411205 Chr.11: 120660246-120660246 on Build GRCh38
rs766063805 Chr.11: 120660442-120660442 on Build GRCh38
rs916791878 Chr.11: 120660654-120660654 on Build GRCh38
rs111487154 Chr.11: 120660333-120660333 on Build GRCh38
rs200538196 Chr.11: 120660364-120660364 on Build GRCh38
rs766648062 Chr.11: 120660328-120660328 on Build GRCh38
rs1015476691 Chr.11: 120660512-120660515 on Build GRCh38
rs947344842 Chr.11: 120660543-120660543 on Build GRCh38
rs559401746 Chr.11: 120660507-120660507 on Build GRCh38
rs372689614 Chr.11: 120660357-120660357 on Build GRCh38
rs745543714 Chr.11: 120660277-120660277 on Build GRCh38
rs890969036 Chr.11: 120660307-120660307 on Build GRCh38
rs564050342 Chr.11: 120660621-120660621 on Build GRCh38
rs545212051 Chr.11: 120660523-120660523 on Build GRCh38
rs112467423 Chr.11: 120660373-120660373 on Build GRCh38
rs780766871 Chr.11: 120660451-120660451 on Build GRCh38
rs372859300 Chr.11: 120660374-120660374 on Build GRCh38
rs142433514 Chr.11: 120660363-120660363 on Build GRCh38
rs182062965 Chr.11: 120660679-120660679 on Build GRCh38
rs150199548 Chr.11: 120660325-120660325 on Build GRCh38
rs761851204 Chr.11: 120660410-120660410 on Build GRCh38
rs771171484 Chr.11: 120660383-120660383 on Build GRCh38
rs768955265 Chr.11: 120660684-120660684 on Build GRCh38
rs753839952 Chr.11: 120660335-120660335 on Build GRCh38
rs1015123466 Chr.11: 120660644-120660644 on Build GRCh38
rs936826648 Chr.11: 120660625-120660625 on Build GRCh38
rs1052913739 Chr.11: 120660531-120660531 on Build GRCh38
rs989768761 Chr.11: 120660642-120660642 on Build GRCh38
rs201300237 Chr.11: 120660327-120660327 on Build GRCh38
rs973792630 Chr.11: 120660376-120660376 on Build GRCh38
rs1012640351 Chr.11: 120660411-120660411 on Build GRCh38
rs568567238 Chr.11: 120660304-120660304 on Build GRCh38
rs983659814 Chr.11: 120660367-120660367 on Build GRCh38
rs773499648 Chr.11: 120660326-120660326 on Build GRCh38
rs1053672881 Chr.11: 120660258-120660258 on Build GRCh38
rs142046109 Chr.11: 120660399-120660399 on Build GRCh38
rs138074057 Chr.11: 120660340-120660340 on Build GRCh38
rs867044276 Chr.11: 120660254-120660254 on Build GRCh38
rs758251026 Chr.11: 120660349-120660349 on Build GRCh38
rs1029372453 Chr.11: 120660686-120660686 on Build GRCh38
rs770327575 Chr.11: 120660338-120660338 on Build GRCh38
rs760499343 Chr.11: 120660650-120660650 on Build GRCh38
rs554168665 Chr.11: 120660687-120660687 on Build GRCh38
rs759541565 Chr.11: 120660450-120660451 on Build GRCh38
rs891697437 Chr.11: 120660537-120660537 on Build GRCh38
rs530988553 Chr.11: 120660581-120660581 on Build GRCh38
rs146239873 Chr.11: 120660337-120660337 on Build GRCh38
rs555672975 Chr.11: 120660475-120660475 on Build GRCh38
rs779441087 Chr.11: 120660285-120660285 on Build GRCh38
rs546632616 Chr.11: 120660630-120660630 on Build GRCh38
rs532822079 Chr.11: 120660512-120660512 on Build GRCh38
rs942024782 Chr.11: 120660244-120660244 on Build GRCh38
rs567855361 Chr.11: 120660608-120660608 on Build GRCh38
rs370863491 Chr.11: 120660444-120660444 on Build GRCh38
rs963940409 Chr.11: 120660645-120660645 on Build GRCh38
rs1012364247 Chr.11: 120660265-120660265 on Build GRCh38
rs540664984 Chr.11: 120660473-120660473 on Build GRCh38
rs770228738 Chr.11: 120660372-120660372 on Build GRCh38
rs750180890 Chr.11: 120660348-120660348 on Build GRCh38

View Full Product Details