Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 121469916-121470412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SORL1
Entrez Gene ID
6653
Gene Name
sortilin related receptor 1
Gene Aliases
C11orf32, LR11, LRP9, SORLA, SorLA-1, gp250
UniGene
Hs.368592
Gene Chromosome Location
Chr.11: 121452203-121633762 on Build GRCh38
Transcripts
NM_003105.5 XM_011542963.2 XM_017018169.1 XM_017018171.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5539551 c.348G>A p.V116V Chr.11: 121470069-121470069 on Build GRCh38
7201169 c.329C>G p.A110G Chr.11: 121470050-121470050 on Build GRCh38
1704707 c.298G>A p.D100N Chr.11: 121470019-121470019 on Build GRCh38
924037 c.328G>A p.A110T Chr.11: 121470049-121470049 on Build GRCh38
4424240 c.286-3C>T p.? Chr.11: 121470004-121470004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577587443 Chr.11: 121470104-121470104 on Build GRCh38
rs962594365 Chr.11: 121470072-121470072 on Build GRCh38
rs765759259 Chr.11: 121470151-121470151 on Build GRCh38
rs767065500 Chr.11: 121470198-121470198 on Build GRCh38
rs781496618 Chr.11: 121470044-121470044 on Build GRCh38
rs750011509 Chr.11: 121469998-121469998 on Build GRCh38
rs761691535 Chr.11: 121470118-121470118 on Build GRCh38
rs762595967 Chr.11: 121470383-121470383 on Build GRCh38
rs372499441 Chr.11: 121469996-121469996 on Build GRCh38
rs945678306 Chr.11: 121470340-121470340 on Build GRCh38
rs78397535 Chr.11: 121470066-121470066 on Build GRCh38
rs542707843 Chr.11: 121470260-121470260 on Build GRCh38
rs564006388 Chr.11: 121470067-121470067 on Build GRCh38
rs1041826702 Chr.11: 121470278-121470278 on Build GRCh38
rs772116245 Chr.11: 121469987-121469987 on Build GRCh38
rs1043768149 Chr.11: 121470341-121470341 on Build GRCh38
rs914148576 Chr.11: 121470334-121470334 on Build GRCh38
rs1002334920 Chr.11: 121470353-121470353 on Build GRCh38
rs748481255 Chr.11: 121470064-121470064 on Build GRCh38
rs752093771 Chr.11: 121470158-121470158 on Build GRCh38
rs578243838 Chr.11: 121470030-121470030 on Build GRCh38
rs771659216 Chr.11: 121470326-121470326 on Build GRCh38
rs766916904 Chr.11: 121470017-121470017 on Build GRCh38
rs527552540 Chr.11: 121470343-121470343 on Build GRCh38
rs772201413 Chr.11: 121470105-121470105 on Build GRCh38
rs775517202 Chr.11: 121470085-121470085 on Build GRCh38
rs982581946 Chr.11: 121470035-121470035 on Build GRCh38
rs374968242 Chr.11: 121469968-121469968 on Build GRCh38
rs753573584 Chr.11: 121469994-121469994 on Build GRCh38
rs1004264534 Chr.11: 121470357-121470357 on Build GRCh38
rs1016699318 Chr.11: 121470374-121470374 on Build GRCh38
rs772676860 Chr.11: 121470148-121470148 on Build GRCh38
rs769893383 Chr.11: 121469973-121469973 on Build GRCh38
rs958914185 Chr.11: 121469954-121469954 on Build GRCh38
rs910689441 Chr.11: 121470300-121470300 on Build GRCh38
rs142933415 Chr.11: 121470028-121470028 on Build GRCh38
rs972463538 Chr.11: 121470179-121470179 on Build GRCh38
rs775226673 Chr.11: 121469990-121469990 on Build GRCh38
rs749389644 Chr.11: 121470073-121470073 on Build GRCh38
rs183191029 Chr.11: 121469974-121469974 on Build GRCh38
rs762540405 Chr.11: 121470149-121470149 on Build GRCh38
rs768680314 Chr.11: 121470086-121470086 on Build GRCh38
rs763929406 Chr.11: 121469993-121469993 on Build GRCh38
rs770856759 Chr.11: 121470083-121470083 on Build GRCh38
rs116967561 Chr.11: 121470161-121470161 on Build GRCh38
rs760558412 Chr.11: 121469992-121469992 on Build GRCh38
rs767989786 Chr.11: 121470165-121470165 on Build GRCh38
rs765010178 Chr.11: 121470139-121470139 on Build GRCh38
rs368229087 Chr.11: 121470169-121470169 on Build GRCh38
rs867156559 Chr.11: 121470142-121470142 on Build GRCh38
rs777852927 Chr.11: 121470068-121470068 on Build GRCh38
rs768940438 Chr.11: 121469956-121469956 on Build GRCh38
rs751955751 Chr.11: 121470026-121470026 on Build GRCh38
rs757844871 Chr.11: 121470007-121470007 on Build GRCh38
rs561342774 Chr.11: 121470331-121470331 on Build GRCh38
rs147008552 Chr.11: 121470219-121470219 on Build GRCh38
rs868417203 Chr.11: 121470242-121470242 on Build GRCh38
rs909605764 Chr.11: 121470047-121470047 on Build GRCh38
rs370172030 Chr.11: 121469957-121469957 on Build GRCh38
rs960544735 Chr.11: 121470096-121470096 on Build GRCh38
rs762929621 Chr.11: 121469978-121469978 on Build GRCh38

View Full Product Details