Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 32795805-32796304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKP2
Entrez Gene ID
5318
Gene Name
plakophilin 2
Gene Aliases
ARVD9
UniGene
Hs.164384
Gene Chromosome Location
Chr.12: 32790746-32896846 on Build GRCh38
Transcripts
NM_001005242.2 NM_004572.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1476506 c.2306A>G p.E769G Chr.12: 32796292-32796292 on Build GRCh38
5375045 c.2364C>T p.L788L Chr.12: 32796234-32796234 on Build GRCh38
6000224 c.2384C>T p.A795V Chr.12: 32796214-32796214 on Build GRCh38
6462553 c.2443A>C p.N815H Chr.12: 32796155-32796155 on Build GRCh38
5840878 c.2458C>T p.Q820* Chr.12: 32796140-32796140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768677218 Chr.12: 32796182-32796182 on Build GRCh38
rs758834771 Chr.12: 32796250-32796250 on Build GRCh38
rs786204395 Chr.12: 32796150-32796155 on Build GRCh38
rs757565158 Chr.12: 32796263-32796263 on Build GRCh38
rs786204394 Chr.12: 32796264-32796269 on Build GRCh38
rs779956129 Chr.12: 32796170-32796170 on Build GRCh38
rs550309210 Chr.12: 32795977-32795977 on Build GRCh38
rs139258347 Chr.12: 32796117-32796117 on Build GRCh38
rs1046718916 Chr.12: 32795901-32795901 on Build GRCh38
rs551045165 Chr.12: 32796233-32796233 on Build GRCh38
rs560524894 Chr.12: 32795852-32795852 on Build GRCh38
rs1057518582 Chr.12: 32796109-32796109 on Build GRCh38
rs371917866 Chr.12: 32796158-32796158 on Build GRCh38
rs563242567 Chr.12: 32795952-32795952 on Build GRCh38
rs546725168 Chr.12: 32796042-32796042 on Build GRCh38
rs779066548 Chr.12: 32796064-32796064 on Build GRCh38
rs397517022 Chr.12: 32796260-32796264 on Build GRCh38
rs542942946 Chr.12: 32795950-32795950 on Build GRCh38
rs149714351 Chr.12: 32796017-32796017 on Build GRCh38
rs77991187 Chr.12: 32796036-32796036 on Build GRCh38
rs765938307 Chr.12: 32796234-32796234 on Build GRCh38
rs369837002 Chr.12: 32796150-32796150 on Build GRCh38
rs777940850 Chr.12: 32796075-32796075 on Build GRCh38
rs200947767 Chr.12: 32796164-32796164 on Build GRCh38
rs888178847 Chr.12: 32795939-32795939 on Build GRCh38
rs914237314 Chr.12: 32795973-32795973 on Build GRCh38
rs574235048 Chr.12: 32795914-32795914 on Build GRCh38
rs566555785 Chr.12: 32796046-32796046 on Build GRCh38
rs945591105 Chr.12: 32795873-32795873 on Build GRCh38
rs947158587 Chr.12: 32795981-32795981 on Build GRCh38
rs989585118 Chr.12: 32796062-32796062 on Build GRCh38
rs749907181 Chr.12: 32796269-32796269 on Build GRCh38
rs61927769 Chr.12: 32796037-32796037 on Build GRCh38
rs200272388 Chr.12: 32796186-32796186 on Build GRCh38
rs71447623 Chr.12: 32796036-32796036 on Build GRCh38
rs772348657 Chr.12: 32796071-32796071 on Build GRCh38
rs541654778 Chr.12: 32795839-32795839 on Build GRCh38
rs764172059 Chr.12: 32796261-32796261 on Build GRCh38
rs201594162 Chr.12: 32796093-32796094 on Build GRCh38
rs770999774 Chr.12: 32796168-32796168 on Build GRCh38
rs928113183 Chr.12: 32795903-32795903 on Build GRCh38
rs774849612 Chr.12: 32796035-32796036 on Build GRCh38
rs151264959 Chr.12: 32796113-32796113 on Build GRCh38
rs147438087 Chr.12: 32796027-32796027 on Build GRCh38
rs149968852 Chr.12: 32796095-32796096 on Build GRCh38
rs12228193 Chr.12: 32796000-32796000 on Build GRCh38
rs796631247 Chr.12: 32796096-32796097 on Build GRCh38
rs727504950 Chr.12: 32796220-32796220 on Build GRCh38
rs535677786 Chr.12: 32796088-32796088 on Build GRCh38
rs794729131 Chr.12: 32796274-32796274 on Build GRCh38
rs755375729 Chr.12: 32796063-32796063 on Build GRCh38
rs377139489 Chr.12: 32796072-32796072 on Build GRCh38
rs766775778 Chr.12: 32796149-32796149 on Build GRCh38
rs976756692 Chr.12: 32795913-32795913 on Build GRCh38
rs953031265 Chr.12: 32795919-32795919 on Build GRCh38
rs139734328 Chr.12: 32796167-32796167 on Build GRCh38
rs35962192 Chr.12: 32795836-32795837 on Build GRCh38
rs866332621 Chr.12: 32795980-32795980 on Build GRCh38
rs1055703192 Chr.12: 32795931-32795931 on Build GRCh38
rs777122614 Chr.12: 32796237-32796237 on Build GRCh38
rs751577129 Chr.12: 32796262-32796262 on Build GRCh38
rs770411816 Chr.12: 32796007-32796007 on Build GRCh38
rs267603441 Chr.12: 32796201-32796201 on Build GRCh38
rs548988161 Chr.12: 32796090-32796090 on Build GRCh38
rs962215610 Chr.12: 32795905-32795905 on Build GRCh38
rs774405490 Chr.12: 32796183-32796183 on Build GRCh38
rs794729118 Chr.12: 32796213-32796213 on Build GRCh38
rs765782274 Chr.12: 32796245-32796245 on Build GRCh38
rs556563706 Chr.12: 32796266-32796266 on Build GRCh38
rs543233804 Chr.12: 32796232-32796232 on Build GRCh38
rs938157755 Chr.12: 32795922-32795922 on Build GRCh38
rs759790276 Chr.12: 32796240-32796240 on Build GRCh38
rs142362933 Chr.12: 32796111-32796111 on Build GRCh38
rs768463319 Chr.12: 32796110-32796110 on Build GRCh38
rs1006306311 Chr.12: 32795862-32795862 on Build GRCh38
rs748215197 Chr.12: 32796066-32796066 on Build GRCh38
rs112592855 Chr.12: 32796206-32796206 on Build GRCh38
rs759959611 Chr.12: 32796037-32796037 on Build GRCh38
rs111517471 Chr.12: 32796108-32796108 on Build GRCh38
rs998057265 Chr.12: 32796004-32796004 on Build GRCh38
rs369166666 Chr.12: 32796165-32796165 on Build GRCh38
rs147653624 Chr.12: 32796143-32796143 on Build GRCh38
rs745712193 Chr.12: 32796087-32796087 on Build GRCh38
rs913899503 Chr.12: 32796069-32796069 on Build GRCh38
rs753226330 Chr.12: 32796249-32796249 on Build GRCh38
rs868569243 Chr.12: 32796267-32796267 on Build GRCh38
rs876657564 Chr.12: 32796198-32796198 on Build GRCh38
rs1045086472 Chr.12: 32796048-32796048 on Build GRCh38
rs750119363 Chr.12: 32796116-32796116 on Build GRCh38
rs727504509 Chr.12: 32796114-32796114 on Build GRCh38
rs794729098 Chr.12: 32796212-32796212 on Build GRCh38
rs533884071 Chr.12: 32796166-32796166 on Build GRCh38
rs386761907 Chr.12: 32796036-32796037 on Build GRCh38
rs749029420 Chr.12: 32796176-32796176 on Build GRCh38
rs145324631 Chr.12: 32796134-32796134 on Build GRCh38
rs145553222 Chr.12: 32796238-32796238 on Build GRCh38
rs886049320 Chr.12: 32796276-32796276 on Build GRCh38

View Full Product Details