Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 42085786-42086332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GXYLT1
Entrez Gene ID
283464
Gene Name
glucoside xylosyltransferase 1
Gene Aliases
GLT8D3
UniGene
Hs.259347
Gene Chromosome Location
Chr.12: 42081845-42144871 on Build GRCh38
Transcripts
NM_001099650.1 NM_173601.1 XM_017019211.1

SNP Detail

SNP ID Chromosome Location
rs777894938 Chr.12: 42085856-42085856 on Build GRCh38
rs796975592 Chr.12: 42086139-42086139 on Build GRCh38
rs980987093 Chr.12: 42085919-42085919 on Build GRCh38
rs199716489 Chr.12: 42085948-42085948 on Build GRCh38
rs563653894 Chr.12: 42086104-42086104 on Build GRCh38
rs985561321 Chr.12: 42086122-42086122 on Build GRCh38
rs987796837 Chr.12: 42085893-42085893 on Build GRCh38
rs1053272048 Chr.12: 42086299-42086299 on Build GRCh38
rs796387601 Chr.12: 42086252-42086252 on Build GRCh38
rs186634060 Chr.12: 42086129-42086129 on Build GRCh38
rs150603027 Chr.12: 42086031-42086031 on Build GRCh38
rs191227206 Chr.12: 42086162-42086162 on Build GRCh38
rs956363674 Chr.12: 42085852-42085852 on Build GRCh38
rs796135378 Chr.12: 42085859-42085859 on Build GRCh38
rs13377699 Chr.12: 42085895-42085895 on Build GRCh38
rs552325328 Chr.12: 42085845-42085845 on Build GRCh38
rs183706632 Chr.12: 42086276-42086276 on Build GRCh38
rs890327710 Chr.12: 42086239-42086239 on Build GRCh38
rs893154228 Chr.12: 42085926-42085926 on Build GRCh38
rs778393709 Chr.12: 42086071-42086071 on Build GRCh38
rs531343727 Chr.12: 42085855-42085855 on Build GRCh38
rs1019147434 Chr.12: 42085830-42085830 on Build GRCh38
rs796176051 Chr.12: 42085872-42085872 on Build GRCh38
rs994928925 Chr.12: 42086272-42086272 on Build GRCh38
rs796578024 Chr.12: 42086123-42086123 on Build GRCh38
rs192718771 Chr.12: 42086175-42086175 on Build GRCh38
rs767470384 Chr.12: 42085978-42085979 on Build GRCh38
rs527462606 Chr.12: 42085843-42085843 on Build GRCh38
rs898801385 Chr.12: 42086271-42086271 on Build GRCh38
rs768952660 Chr.12: 42086209-42086209 on Build GRCh38
rs796367664 Chr.12: 42086137-42086137 on Build GRCh38
rs879859258 Chr.12: 42085839-42085840 on Build GRCh38
rs905809929 Chr.12: 42086003-42086003 on Build GRCh38
rs796741674 Chr.12: 42085866-42085866 on Build GRCh38
rs182258993 Chr.12: 42086004-42086004 on Build GRCh38
rs77710749 Chr.12: 42085894-42085894 on Build GRCh38
rs946656641 Chr.12: 42085904-42085904 on Build GRCh38
rs139704807 Chr.12: 42086067-42086067 on Build GRCh38
rs749771136 Chr.12: 42086070-42086073 on Build GRCh38
rs140063640 Chr.12: 42086254-42086254 on Build GRCh38
rs1053164651 Chr.12: 42085923-42085923 on Build GRCh38
rs796230494 Chr.12: 42086125-42086125 on Build GRCh38
rs368162491 Chr.12: 42086013-42086013 on Build GRCh38
rs546311303 Chr.12: 42086230-42086230 on Build GRCh38
rs914670245 Chr.12: 42086301-42086301 on Build GRCh38
rs557843241 Chr.12: 42086227-42086227 on Build GRCh38
rs1022952915 Chr.12: 42085956-42085956 on Build GRCh38
rs539615286 Chr.12: 42086184-42086184 on Build GRCh38
rs536184635 Chr.12: 42086079-42086079 on Build GRCh38
rs952479606 Chr.12: 42086086-42086086 on Build GRCh38
rs943623344 Chr.12: 42085861-42085861 on Build GRCh38
rs77241271 Chr.12: 42085935-42085935 on Build GRCh38
rs1035491260 Chr.12: 42086022-42086022 on Build GRCh38
rs781267950 Chr.12: 42085898-42085898 on Build GRCh38
rs934739632 Chr.12: 42085897-42085897 on Build GRCh38
rs111538565 Chr.12: 42086182-42086182 on Build GRCh38
rs1014878078 Chr.12: 42086265-42086265 on Build GRCh38
rs76595571 Chr.12: 42085917-42085917 on Build GRCh38
rs796994102 Chr.12: 42085867-42085867 on Build GRCh38
rs79627623 Chr.12: 42085907-42085907 on Build GRCh38
rs1002743828 Chr.12: 42086007-42086007 on Build GRCh38
rs748722732 Chr.12: 42085944-42085944 on Build GRCh38
rs570528389 Chr.12: 42085854-42085854 on Build GRCh38

View Full Product Details