Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 44508735-44509224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NELL2
Entrez Gene ID
4753
Gene Name
neural EGFL like 2
Gene Aliases
NRP2
UniGene
Hs.505326
Gene Chromosome Location
Chr.12: 44508275-44921848 on Build GRCh38
Transcripts
NM_001145107.1 NM_001145108.1 NM_001145109.1 NM_001145110.1 NM_006159.2 XM_005268905.3 XM_011538396.1 XM_017019341.1 XM_017019342.1 XM_017019343.1 XM_017019344.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7379383 c.2404C>T p.H802Y Chr.12: 44508978-44508978 on Build GRCh38
6724649 c.2409C>A p.I803I Chr.12: 44508973-44508973 on Build GRCh38
1361637 c.2431C>T p.Q811* Chr.12: 44508954-44508954 on Build GRCh38
3460888 c.2405G>A p.G802D Chr.12: 44508980-44508980 on Build GRCh38
6724647 c.2562C>A p.I854I Chr.12: 44508973-44508973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764074101 Chr.12: 44509136-44509137 on Build GRCh38
rs753742958 Chr.12: 44509009-44509009 on Build GRCh38
rs1031803857 Chr.12: 44509182-44509182 on Build GRCh38
rs1008802244 Chr.12: 44509175-44509175 on Build GRCh38
rs753857415 Chr.12: 44509160-44509160 on Build GRCh38
rs374447353 Chr.12: 44508914-44508914 on Build GRCh38
rs558534370 Chr.12: 44509057-44509057 on Build GRCh38
rs773739627 Chr.12: 44508925-44508926 on Build GRCh38
rs556621137 Chr.12: 44508872-44508872 on Build GRCh38
rs957895016 Chr.12: 44508969-44508969 on Build GRCh38
rs924852603 Chr.12: 44508838-44508838 on Build GRCh38
rs1028516078 Chr.12: 44508779-44508779 on Build GRCh38
rs747677751 Chr.12: 44508895-44508895 on Build GRCh38
rs201448813 Chr.12: 44509012-44509012 on Build GRCh38
rs778579462 Chr.12: 44508900-44508900 on Build GRCh38
rs770264871 Chr.12: 44508888-44508889 on Build GRCh38
rs747742756 Chr.12: 44508901-44508901 on Build GRCh38
rs1015173514 Chr.12: 44508862-44508862 on Build GRCh38
rs535457634 Chr.12: 44509108-44509108 on Build GRCh38
rs771414147 Chr.12: 44508925-44508925 on Build GRCh38
rs568173323 Chr.12: 44508853-44508853 on Build GRCh38
rs762046983 Chr.12: 44508993-44508993 on Build GRCh38
rs370226647 Chr.12: 44508868-44508868 on Build GRCh38
rs940610838 Chr.12: 44509149-44509149 on Build GRCh38
rs553270096 Chr.12: 44508797-44508797 on Build GRCh38
rs766223221 Chr.12: 44509003-44509003 on Build GRCh38
rs1377002 Chr.12: 44508790-44508790 on Build GRCh38
rs73096324 Chr.12: 44509128-44509128 on Build GRCh38
rs887720785 Chr.12: 44508825-44508825 on Build GRCh38
rs746361889 Chr.12: 44508943-44508943 on Build GRCh38
rs191798719 Chr.12: 44509030-44509030 on Build GRCh38
rs567456603 Chr.12: 44508767-44508767 on Build GRCh38
rs1048750196 Chr.12: 44509088-44509088 on Build GRCh38
rs370452957 Chr.12: 44508897-44508897 on Build GRCh38
rs187562309 Chr.12: 44508887-44508887 on Build GRCh38
rs746570224 Chr.12: 44508918-44508918 on Build GRCh38
rs900147637 Chr.12: 44509095-44509095 on Build GRCh38
rs1803372 Chr.12: 44508892-44508892 on Build GRCh38
rs577562350 Chr.12: 44508806-44508806 on Build GRCh38
rs775736399 Chr.12: 44508952-44508952 on Build GRCh38
rs765034594 Chr.12: 44509020-44509020 on Build GRCh38
rs1037778121 Chr.12: 44509084-44509084 on Build GRCh38
rs199836199 Chr.12: 44508978-44508978 on Build GRCh38
rs781754699 Chr.12: 44508938-44508938 on Build GRCh38
rs368039851 Chr.12: 44509033-44509033 on Build GRCh38
rs775267725 Chr.12: 44508992-44508993 on Build GRCh38
rs367712742 Chr.12: 44508953-44508953 on Build GRCh38
rs759159833 Chr.12: 44508990-44508990 on Build GRCh38
rs375378527 Chr.12: 44508857-44508857 on Build GRCh38
rs1030075890 Chr.12: 44509164-44509164 on Build GRCh38
rs956097534 Chr.12: 44508848-44508848 on Build GRCh38
rs769257185 Chr.12: 44509067-44509067 on Build GRCh38
rs769042415 Chr.12: 44508960-44508960 on Build GRCh38
rs759408161 Chr.12: 44509013-44509013 on Build GRCh38
rs955562099 Chr.12: 44509165-44509165 on Build GRCh38
rs777231713 Chr.12: 44508916-44508916 on Build GRCh38
rs541352315 Chr.12: 44509134-44509134 on Build GRCh38
rs267603468 Chr.12: 44508942-44508942 on Build GRCh38
rs766391845 Chr.12: 44508883-44508883 on Build GRCh38
rs886348476 Chr.12: 44509053-44509053 on Build GRCh38
rs574958799 Chr.12: 44508875-44508875 on Build GRCh38
rs770135751 Chr.12: 44508948-44508948 on Build GRCh38
rs771633333 Chr.12: 44508993-44508993 on Build GRCh38

View Full Product Details