Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 80708748-80709280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYF6
Entrez Gene ID
4618
Gene Name
myogenic factor 6
Gene Aliases
CNM3, MRF4, bHLHc4, myf-6
UniGene
Hs.35937
Gene Chromosome Location
Chr.12: 80707629-80709478 on Build GRCh38
Transcripts
NM_002469.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5913429 c.700C>T p.P234S Chr.12: 80708931-80708931 on Build GRCh38
276199 c.629C>T p.S210L Chr.12: 80708860-80708860 on Build GRCh38
943858 c.638C>T p.S213L Chr.12: 80708869-80708869 on Build GRCh38
5376103 c.622A>T p.I208F Chr.12: 80708853-80708853 on Build GRCh38
1364476 c.667G>A p.V223M Chr.12: 80708898-80708898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566327783 Chr.12: 80709092-80709092 on Build GRCh38
rs550225751 Chr.12: 80709026-80709026 on Build GRCh38
rs556059811 Chr.12: 80709087-80709087 on Build GRCh38
rs886049857 Chr.12: 80709145-80709145 on Build GRCh38
rs201098963 Chr.12: 80708914-80708914 on Build GRCh38
rs762081940 Chr.12: 80708958-80708958 on Build GRCh38
rs746361902 Chr.12: 80708900-80708900 on Build GRCh38
rs914913149 Chr.12: 80709019-80709019 on Build GRCh38
rs537924366 Chr.12: 80709003-80709003 on Build GRCh38
rs539223659 Chr.12: 80709043-80709043 on Build GRCh38
rs1027725312 Chr.12: 80708981-80708981 on Build GRCh38
rs757639492 Chr.12: 80708876-80708876 on Build GRCh38
rs777044772 Chr.12: 80708953-80708953 on Build GRCh38
rs759406297 Chr.12: 80708989-80708989 on Build GRCh38
rs779208935 Chr.12: 80708880-80708880 on Build GRCh38
rs767550568 Chr.12: 80708991-80708991 on Build GRCh38
rs577457456 Chr.12: 80709257-80709257 on Build GRCh38
rs775844624 Chr.12: 80708915-80708915 on Build GRCh38
rs192420949 Chr.12: 80709067-80709067 on Build GRCh38
rs1014911545 Chr.12: 80709204-80709204 on Build GRCh38
rs1018032749 Chr.12: 80708969-80708969 on Build GRCh38
rs890591677 Chr.12: 80709165-80709165 on Build GRCh38
rs763544470 Chr.12: 80709008-80709008 on Build GRCh38
rs966247445 Chr.12: 80708885-80708885 on Build GRCh38
rs762867854 Chr.12: 80709184-80709184 on Build GRCh38
rs886049856 Chr.12: 80708865-80708865 on Build GRCh38
rs900528629 Chr.12: 80709193-80709193 on Build GRCh38
rs946386988 Chr.12: 80709090-80709090 on Build GRCh38
rs766941896 Chr.12: 80708854-80708854 on Build GRCh38
rs754909325 Chr.12: 80708848-80708848 on Build GRCh38
rs182525372 Chr.12: 80709163-80709163 on Build GRCh38
rs3120 Chr.12: 80709112-80709112 on Build GRCh38
rs934817396 Chr.12: 80708768-80708768 on Build GRCh38
rs752195466 Chr.12: 80708856-80708856 on Build GRCh38
rs867670483 Chr.12: 80708886-80708886 on Build GRCh38
rs1015397861 Chr.12: 80708865-80708865 on Build GRCh38
rs994838419 Chr.12: 80709223-80709223 on Build GRCh38
rs1039267743 Chr.12: 80709182-80709182 on Build GRCh38
rs79738065 Chr.12: 80708979-80708979 on Build GRCh38
rs549067340 Chr.12: 80708940-80708940 on Build GRCh38
rs367653508 Chr.12: 80708970-80708970 on Build GRCh38
rs751573649 Chr.12: 80708822-80708822 on Build GRCh38
rs756836102 Chr.12: 80709012-80709012 on Build GRCh38
rs923715427 Chr.12: 80709056-80709056 on Build GRCh38
rs976683088 Chr.12: 80708896-80708896 on Build GRCh38
rs762883381 Chr.12: 80708806-80708806 on Build GRCh38
rs201015986 Chr.12: 80708794-80708794 on Build GRCh38
rs754636641 Chr.12: 80709246-80709246 on Build GRCh38
rs766258230 Chr.12: 80708815-80708815 on Build GRCh38
rs868391302 Chr.12: 80708996-80708996 on Build GRCh38
rs141373187 Chr.12: 80709033-80709033 on Build GRCh38
rs369556625 Chr.12: 80709173-80709173 on Build GRCh38
rs753268041 Chr.12: 80708829-80708829 on Build GRCh38
rs555826001 Chr.12: 80709220-80709220 on Build GRCh38
rs939097257 Chr.12: 80709076-80709076 on Build GRCh38
rs755568133 Chr.12: 80708867-80708867 on Build GRCh38
rs3121 Chr.12: 80709142-80709142 on Build GRCh38
rs900475235 Chr.12: 80708971-80708971 on Build GRCh38
rs373402944 Chr.12: 80709010-80709010 on Build GRCh38
rs777471992 Chr.12: 80708871-80708871 on Build GRCh38
rs944968745 Chr.12: 80709180-80709180 on Build GRCh38
rs916251686 Chr.12: 80709061-80709061 on Build GRCh38
rs748561934 Chr.12: 80709029-80709029 on Build GRCh38
rs752575127 Chr.12: 80709002-80709002 on Build GRCh38
rs757260199 Chr.12: 80708829-80708830 on Build GRCh38
rs747177260 Chr.12: 80708926-80708926 on Build GRCh38
rs990722521 Chr.12: 80709038-80709038 on Build GRCh38
rs533796044 Chr.12: 80709107-80709107 on Build GRCh38
rs754416637 Chr.12: 80708873-80708873 on Build GRCh38

View Full Product Details