Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 81261839-81262338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPFIA2
Entrez Gene ID
8499
Gene Name
PTPRF interacting protein alpha 2
Gene Aliases
-
UniGene
Hs.506216
Gene Chromosome Location
Chr.12: 81257975-81759748 on Build GRCh38
Transcripts
NM_001220473.2 NM_001220474.2 NM_001220475.2 NM_001220476.2 NM_001220477.2 NM_001220478.2 NM_001220479.2 NM_001220480.2 NM_001282536.1 NM_003625.4 XM_006719670.3 XM_011538906.2 XM_011538907.2 XM_017020079.1 XM_017020080.1 XM_017020081.1 XM_017020082.1 XM_017020083.1 XM_017020084.1 XM_017020085.1 XM_017020086.1 XM_017020087.1 XM_017020088.1 XM_017020089.1 XM_017020090.1 XM_017020091.1 XM_017020092.1 XM_017020093.1 XM_017020094.1 XM_017020095.1 XM_017020096.1 XM_017020097.1 XM_017020098.1 XM_017020099.1 XM_017020100.1 XM_017020101.1 XM_017020102.1 XM_017020103.1 XM_017020104.1 XM_017020105.1 XM_017020106.1 XM_017020107.1 XM_017020108.1 XM_017020109.1 XM_017020110.1 XM_017020111.1 XM_017020112.1 XM_017020113.1 XM_017020114.1 XM_017020115.1 XM_017020116.1 XM_017020117.1 XM_017020118.1 XM_017020119.1 XM_017020120.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1993520 c.3757C>T p.R1253C Chr.12: 81261999-81261999 on Build GRCh38
299433 c.3716-9C>A p.? Chr.12: 81262049-81262049 on Build GRCh38
4778290 c.3765C>A p.Y1255* Chr.12: 81261991-81261991 on Build GRCh38
7332860 c.3761C>T p.T1254I Chr.12: 81261995-81261995 on Build GRCh38
7238998 c.3736A>T p.R1246W Chr.12: 81262020-81262020 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs975118893 Chr.12: 81261985-81261985 on Build GRCh38
rs370167407 Chr.12: 81262081-81262081 on Build GRCh38
rs780603198 Chr.12: 81262076-81262076 on Build GRCh38
rs1002649876 Chr.12: 81261888-81261888 on Build GRCh38
rs775327308 Chr.12: 81262032-81262032 on Build GRCh38
rs773379232 Chr.12: 81262015-81262015 on Build GRCh38
rs1006269432 Chr.12: 81262260-81262260 on Build GRCh38
rs549260926 Chr.12: 81262305-81262305 on Build GRCh38
rs572879102 Chr.12: 81261953-81261953 on Build GRCh38
rs964971602 Chr.12: 81262223-81262223 on Build GRCh38
rs566485653 Chr.12: 81262257-81262257 on Build GRCh38
rs1013451899 Chr.12: 81261904-81261905 on Build GRCh38
rs907974515 Chr.12: 81262039-81262039 on Build GRCh38
rs767353454 Chr.12: 81262026-81262026 on Build GRCh38
rs891551509 Chr.12: 81262196-81262196 on Build GRCh38
rs545929741 Chr.12: 81261901-81261901 on Build GRCh38
rs570392260 Chr.12: 81262189-81262191 on Build GRCh38
rs753303133 Chr.12: 81262051-81262051 on Build GRCh38
rs1023541006 Chr.12: 81261906-81261906 on Build GRCh38
rs529075425 Chr.12: 81262059-81262059 on Build GRCh38
rs117454231 Chr.12: 81262276-81262276 on Build GRCh38
rs747510642 Chr.12: 81261980-81261980 on Build GRCh38
rs371092727 Chr.12: 81262086-81262086 on Build GRCh38
rs748812574 Chr.12: 81261936-81261936 on Build GRCh38
rs964640473 Chr.12: 81261981-81261981 on Build GRCh38
rs10862276 Chr.12: 81262134-81262134 on Build GRCh38
rs2272501 Chr.12: 81262082-81262082 on Build GRCh38
rs1005907274 Chr.12: 81261914-81261914 on Build GRCh38
rs906934867 Chr.12: 81261872-81261872 on Build GRCh38
rs756761619 Chr.12: 81262053-81262053 on Build GRCh38
rs983590912 Chr.12: 81262029-81262029 on Build GRCh38
rs1045366431 Chr.12: 81262234-81262234 on Build GRCh38
rs989990124 Chr.12: 81261917-81261917 on Build GRCh38
rs933400379 Chr.12: 81262014-81262014 on Build GRCh38
rs978803720 Chr.12: 81262125-81262125 on Build GRCh38
rs1011389326 Chr.12: 81262220-81262220 on Build GRCh38
rs923323154 Chr.12: 81262002-81262002 on Build GRCh38
rs367984490 Chr.12: 81262085-81262085 on Build GRCh38
rs773939740 Chr.12: 81262120-81262120 on Build GRCh38
rs955890856 Chr.12: 81261913-81261913 on Build GRCh38
rs531308484 Chr.12: 81262194-81262194 on Build GRCh38
rs371448330 Chr.12: 81261942-81261942 on Build GRCh38
rs770005433 Chr.12: 81262013-81262013 on Build GRCh38
rs540492663 Chr.12: 81262004-81262004 on Build GRCh38
rs776630739 Chr.12: 81261991-81261991 on Build GRCh38
rs756921140 Chr.12: 81261938-81261938 on Build GRCh38
rs377033830 Chr.12: 81262080-81262080 on Build GRCh38
rs759345319 Chr.12: 81262022-81262022 on Build GRCh38
rs1031391117 Chr.12: 81262191-81262191 on Build GRCh38
rs1003026952 Chr.12: 81262183-81262183 on Build GRCh38
rs948359034 Chr.12: 81262105-81262105 on Build GRCh38
rs551483822 Chr.12: 81262240-81262240 on Build GRCh38
rs1057182623 Chr.12: 81262202-81262202 on Build GRCh38
rs190385979 Chr.12: 81261892-81261892 on Build GRCh38
rs370589815 Chr.12: 81261913-81261914 on Build GRCh38
rs533221724 Chr.12: 81262233-81262233 on Build GRCh38
rs1021478447 Chr.12: 81262221-81262221 on Build GRCh38
rs896314656 Chr.12: 81262210-81262210 on Build GRCh38
rs200340420 Chr.12: 81261982-81261982 on Build GRCh38
rs1052227382 Chr.12: 81261867-81261867 on Build GRCh38
rs780728977 Chr.12: 81261960-81261960 on Build GRCh38
rs778652217 Chr.12: 81261939-81261939 on Build GRCh38
rs750047916 Chr.12: 81262077-81262077 on Build GRCh38
rs764081959 Chr.12: 81262049-81262049 on Build GRCh38
rs182224954 Chr.12: 81261903-81261903 on Build GRCh38
rs187153200 Chr.12: 81262037-81262037 on Build GRCh38

View Full Product Details