Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 118380038-118380547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUDS3
Entrez Gene ID
64426
Gene Name
SDS3 homolog, SIN3A corepressor complex component
Gene Aliases
SAP45, SDS3
UniGene
Hs.416630
Gene Chromosome Location
Chr.12: 118373189-118418035 on Build GRCh38
Transcripts
NM_022491.2 XM_011538671.2 XM_017019818.1 XM_017019819.1 XM_017019820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4760592 c.205A>G p.K69E Chr.12: 118380224-118380224 on Build GRCh38
7292281 c.175A>G p.K59E Chr.12: 118380194-118380194 on Build GRCh38
5341908 c.143-3C>T p.? Chr.12: 118380159-118380159 on Build GRCh38
467870 c.210A>G p.E70E Chr.12: 118380229-118380229 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748835766 Chr.12: 118380187-118380187 on Build GRCh38
rs770017827 Chr.12: 118380262-118380262 on Build GRCh38
rs773211105 Chr.12: 118380264-118380264 on Build GRCh38
rs145380587 Chr.12: 118380070-118380070 on Build GRCh38
rs146694476 Chr.12: 118380475-118380475 on Build GRCh38
rs958298214 Chr.12: 118380342-118380342 on Build GRCh38
rs10850967 Chr.12: 118380072-118380072 on Build GRCh38
rs200129355 Chr.12: 118380214-118380214 on Build GRCh38
rs773338440 Chr.12: 118380173-118380173 on Build GRCh38
rs189537807 Chr.12: 118380502-118380502 on Build GRCh38
rs1025223801 Chr.12: 118380411-118380411 on Build GRCh38
rs766233481 Chr.12: 118380198-118380198 on Build GRCh38
rs747361603 Chr.12: 118380125-118380125 on Build GRCh38
rs746360515 Chr.12: 118380116-118380116 on Build GRCh38
rs918661889 Chr.12: 118380439-118380439 on Build GRCh38
rs903913067 Chr.12: 118380269-118380269 on Build GRCh38
rs758374839 Chr.12: 118380071-118380071 on Build GRCh38
rs892636931 Chr.12: 118380337-118380337 on Build GRCh38
rs566244952 Chr.12: 118380289-118380289 on Build GRCh38
rs139265823 Chr.12: 118380514-118380514 on Build GRCh38
rs774595917 Chr.12: 118380427-118380427 on Build GRCh38
rs970748364 Chr.12: 118380417-118380417 on Build GRCh38
rs762066191 Chr.12: 118380150-118380150 on Build GRCh38
rs538178067 Chr.12: 118380480-118380480 on Build GRCh38
rs775854397 Chr.12: 118380117-118380117 on Build GRCh38
rs756804964 Chr.12: 118380212-118380213 on Build GRCh38
rs747603752 Chr.12: 118380243-118380243 on Build GRCh38
rs1000971627 Chr.12: 118380287-118380287 on Build GRCh38
rs1048364989 Chr.12: 118380119-118380119 on Build GRCh38
rs758895527 Chr.12: 118380226-118380226 on Build GRCh38
rs550989702 Chr.12: 118380082-118380082 on Build GRCh38
rs762890494 Chr.12: 118380196-118380196 on Build GRCh38
rs767458289 Chr.12: 118380203-118380203 on Build GRCh38
rs990995043 Chr.12: 118380375-118380375 on Build GRCh38
rs769802793 Chr.12: 118380461-118380461 on Build GRCh38
rs143241090 Chr.12: 118380515-118380515 on Build GRCh38
rs748360560 Chr.12: 118380249-118380249 on Build GRCh38
rs972283139 Chr.12: 118380094-118380094 on Build GRCh38
rs751330932 Chr.12: 118380200-118380200 on Build GRCh38
rs777492004 Chr.12: 118380115-118380115 on Build GRCh38
rs576370208 Chr.12: 118380127-118380128 on Build GRCh38
rs376479816 Chr.12: 118380241-118380241 on Build GRCh38
rs531239979 Chr.12: 118380079-118380079 on Build GRCh38
rs79163346 Chr.12: 118380107-118380107 on Build GRCh38
rs572519735 Chr.12: 118380388-118380388 on Build GRCh38
rs1024606866 Chr.12: 118380067-118380067 on Build GRCh38
rs555540429 Chr.12: 118380381-118380381 on Build GRCh38
rs749534010 Chr.12: 118380341-118380341 on Build GRCh38
rs768877606 Chr.12: 118380133-118380133 on Build GRCh38
rs768830268 Chr.12: 118380202-118380202 on Build GRCh38
rs780445182 Chr.12: 118380228-118380228 on Build GRCh38
rs748749200 Chr.12: 118380221-118380221 on Build GRCh38
rs535733156 Chr.12: 118380294-118380294 on Build GRCh38
rs752392988 Chr.12: 118380209-118380209 on Build GRCh38
rs745577794 Chr.12: 118380273-118380275 on Build GRCh38
rs998376960 Chr.12: 118380244-118380244 on Build GRCh38
rs771095044 Chr.12: 118380419-118380419 on Build GRCh38
rs185515014 Chr.12: 118380314-118380314 on Build GRCh38
rs772475473 Chr.12: 118380281-118380282 on Build GRCh38
rs778635161 Chr.12: 118380272-118380272 on Build GRCh38
rs1015947645 Chr.12: 118380466-118380466 on Build GRCh38
rs577340815 Chr.12: 118380507-118380507 on Build GRCh38
rs889068689 Chr.12: 118380239-118380239 on Build GRCh38
rs776814042 Chr.12: 118380135-118380135 on Build GRCh38
rs181326809 Chr.12: 118380213-118380213 on Build GRCh38
rs117730538 Chr.12: 118380518-118380518 on Build GRCh38
rs768874158 Chr.12: 118380247-118380247 on Build GRCh38

View Full Product Details