Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120198476-120198974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPLP0
Entrez Gene ID
6175
Gene Name
ribosomal protein lateral stalk subunit P0
Gene Aliases
L10E, LP0, P0, PRLP0, RPP0
UniGene
Hs.546285
Gene Chromosome Location
Chr.12: 120196700-120201211 on Build GRCh38
Transcripts
NM_001002.3 NM_053275.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6769285 c.603T>C p.P201P Chr.12: 120198602-120198602 on Build GRCh38
6769284 c.519G>A p.T173T Chr.12: 120198686-120198686 on Build GRCh38
4039383 c.345C>A p.A115A Chr.12: 120198974-120198974 on Build GRCh38
4039382 c.617T>C p.I206T Chr.12: 120198588-120198588 on Build GRCh38
6769283 c.624G>A p.E208E Chr.12: 120198581-120198581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146041988 Chr.12: 120198626-120198626 on Build GRCh38
rs777061954 Chr.12: 120198945-120198945 on Build GRCh38
rs772038885 Chr.12: 120198647-120198647 on Build GRCh38
rs759435461 Chr.12: 120198528-120198528 on Build GRCh38
rs377500271 Chr.12: 120198763-120198763 on Build GRCh38
rs772044011 Chr.12: 120198529-120198529 on Build GRCh38
rs761399216 Chr.12: 120198524-120198524 on Build GRCh38
rs748865879 Chr.12: 120198928-120198928 on Build GRCh38
rs756378284 Chr.12: 120198620-120198620 on Build GRCh38
rs138718343 Chr.12: 120198695-120198695 on Build GRCh38
rs776555809 Chr.12: 120198760-120198760 on Build GRCh38
rs113306664 Chr.12: 120198735-120198735 on Build GRCh38
rs748520279 Chr.12: 120198513-120198513 on Build GRCh38
rs200612920 Chr.12: 120198567-120198567 on Build GRCh38
rs759148001 Chr.12: 120198932-120198932 on Build GRCh38
rs1049435751 Chr.12: 120198889-120198889 on Build GRCh38
rs371550795 Chr.12: 120198661-120198661 on Build GRCh38
rs372881911 Chr.12: 120198824-120198824 on Build GRCh38
rs867408911 Chr.12: 120198654-120198654 on Build GRCh38
rs369017873 Chr.12: 120198941-120198941 on Build GRCh38
rs750145678 Chr.12: 120198833-120198834 on Build GRCh38
rs377314568 Chr.12: 120198719-120198719 on Build GRCh38
rs781600167 Chr.12: 120198503-120198503 on Build GRCh38
rs151136387 Chr.12: 120198524-120198524 on Build GRCh38
rs771179332 Chr.12: 120198633-120198633 on Build GRCh38
rs182313909 Chr.12: 120198673-120198673 on Build GRCh38
rs775222896 Chr.12: 120198659-120198659 on Build GRCh38
rs750002971 Chr.12: 120198568-120198568 on Build GRCh38
rs769718181 Chr.12: 120198772-120198772 on Build GRCh38
rs777795514 Chr.12: 120198516-120198516 on Build GRCh38
rs761578930 Chr.12: 120198565-120198565 on Build GRCh38
rs553386311 Chr.12: 120198890-120198890 on Build GRCh38
rs781211506 Chr.12: 120198878-120198878 on Build GRCh38
rs374910964 Chr.12: 120198914-120198914 on Build GRCh38
rs369515748 Chr.12: 120198790-120198790 on Build GRCh38
rs115221652 Chr.12: 120198749-120198749 on Build GRCh38
rs754194367 Chr.12: 120198796-120198796 on Build GRCh38
rs749530115 Chr.12: 120198517-120198517 on Build GRCh38
rs1038364618 Chr.12: 120198794-120198794 on Build GRCh38
rs2074540 Chr.12: 120198746-120198746 on Build GRCh38
rs201156012 Chr.12: 120198523-120198523 on Build GRCh38
rs760450764 Chr.12: 120198542-120198542 on Build GRCh38
rs539541009 Chr.12: 120198823-120198823 on Build GRCh38
rs908063542 Chr.12: 120198768-120198768 on Build GRCh38
rs775741386 Chr.12: 120198578-120198578 on Build GRCh38
rs753098551 Chr.12: 120198704-120198704 on Build GRCh38
rs765930618 Chr.12: 120198701-120198701 on Build GRCh38
rs757547501 Chr.12: 120198806-120198806 on Build GRCh38
rs749950032 Chr.12: 120198690-120198690 on Build GRCh38
rs572962769 Chr.12: 120198929-120198929 on Build GRCh38
rs3180961 Chr.12: 120198650-120198650 on Build GRCh38
rs568378934 Chr.12: 120198757-120198757 on Build GRCh38
rs544200895 Chr.12: 120198598-120198598 on Build GRCh38
rs765771234 Chr.12: 120198779-120198779 on Build GRCh38
rs773174697 Chr.12: 120198923-120198923 on Build GRCh38
rs770051910 Chr.12: 120198514-120198514 on Build GRCh38
rs11548351 Chr.12: 120198635-120198635 on Build GRCh38
rs113815589 Chr.12: 120198562-120198562 on Build GRCh38
rs537068973 Chr.12: 120198777-120198777 on Build GRCh38
rs550538457 Chr.12: 120198803-120198803 on Build GRCh38
rs764875263 Chr.12: 120198783-120198785 on Build GRCh38
rs867197952 Chr.12: 120198604-120198604 on Build GRCh38
rs11548379 Chr.12: 120198605-120198605 on Build GRCh38
rs778226522 Chr.12: 120198621-120198621 on Build GRCh38
rs776598806 Chr.12: 120198669-120198669 on Build GRCh38
rs765041066 Chr.12: 120198680-120198680 on Build GRCh38
rs772943615 Chr.12: 120198774-120198774 on Build GRCh38
rs754247040 Chr.12: 120198734-120198734 on Build GRCh38
rs761061366 Chr.12: 120198793-120198793 on Build GRCh38
rs754867572 Chr.12: 120198834-120198834 on Build GRCh38
rs753374361 Chr.12: 120198564-120198564 on Build GRCh38
rs990874508 Chr.12: 120198738-120198738 on Build GRCh38
rs746003325 Chr.12: 120198636-120198636 on Build GRCh38
rs764639794 Chr.12: 120198795-120198795 on Build GRCh38
rs966508594 Chr.12: 120198563-120198563 on Build GRCh38
rs369822884 Chr.12: 120198577-120198577 on Build GRCh38
rs753254215 Chr.12: 120198606-120198606 on Build GRCh38
rs370074305 Chr.12: 120198745-120198745 on Build GRCh38
rs755528442 Chr.12: 120198588-120198588 on Build GRCh38
rs139992927 Chr.12: 120198812-120198812 on Build GRCh38
rs746881067 Chr.12: 120198753-120198753 on Build GRCh38
rs756680488 Chr.12: 120198710-120198710 on Build GRCh38
rs775337487 Chr.12: 120198540-120198540 on Build GRCh38
rs937755896 Chr.12: 120198559-120198559 on Build GRCh38
rs763953360 Chr.12: 120198544-120198544 on Build GRCh38
rs187195275 Chr.12: 120198736-120198736 on Build GRCh38

View Full Product Details