Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120302185-120302683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRT4
Entrez Gene ID
23409
Gene Name
sirtuin 4
Gene Aliases
SIR2L4
UniGene
Hs.50861
Gene Chromosome Location
Chr.12: 120292199-120313242 on Build GRCh38
Transcripts
NM_012240.2 XM_005253865.4 XM_006719308.2 XM_006719309.3

SNP Detail

SNP ID Chromosome Location
rs1041828039 Chr.12: 120302443-120302443 on Build GRCh38
rs367733916 Chr.12: 120302287-120302287 on Build GRCh38
rs765200215 Chr.12: 120302369-120302369 on Build GRCh38
rs983253791 Chr.12: 120302267-120302267 on Build GRCh38
rs567703467 Chr.12: 120302532-120302532 on Build GRCh38
rs547905248 Chr.12: 120302461-120302461 on Build GRCh38
rs941893398 Chr.12: 120302296-120302296 on Build GRCh38
rs375051312 Chr.12: 120302396-120302396 on Build GRCh38
rs953214860 Chr.12: 120302581-120302581 on Build GRCh38
rs200367607 Chr.12: 120302345-120302345 on Build GRCh38
rs945960816 Chr.12: 120302600-120302600 on Build GRCh38
rs114623397 Chr.12: 120302566-120302566 on Build GRCh38
rs202240486 Chr.12: 120302337-120302337 on Build GRCh38
rs750367828 Chr.12: 120302390-120302390 on Build GRCh38
rs1043347386 Chr.12: 120302474-120302474 on Build GRCh38
rs911787090 Chr.12: 120302598-120302598 on Build GRCh38
rs965875514 Chr.12: 120302217-120302217 on Build GRCh38
rs150226600 Chr.12: 120302546-120302546 on Build GRCh38
rs909092841 Chr.12: 120302349-120302349 on Build GRCh38
rs561594961 Chr.12: 120302659-120302659 on Build GRCh38
rs894607930 Chr.12: 120302519-120302519 on Build GRCh38
rs11613753 Chr.12: 120302367-120302367 on Build GRCh38
rs751782448 Chr.12: 120302257-120302257 on Build GRCh38
rs1024645000 Chr.12: 120302233-120302233 on Build GRCh38
rs946032350 Chr.12: 120302436-120302436 on Build GRCh38
rs149355355 Chr.12: 120302607-120302607 on Build GRCh38
rs987441773 Chr.12: 120302590-120302590 on Build GRCh38
rs569898335 Chr.12: 120302554-120302554 on Build GRCh38
rs138792334 Chr.12: 120302576-120302576 on Build GRCh38
rs903407608 Chr.12: 120302498-120302498 on Build GRCh38
rs938471466 Chr.12: 120302437-120302437 on Build GRCh38
rs970796176 Chr.12: 120302238-120302238 on Build GRCh38
rs201746486 Chr.12: 120302378-120302378 on Build GRCh38
rs959869671 Chr.12: 120302627-120302627 on Build GRCh38
rs112396223 Chr.12: 120302403-120302403 on Build GRCh38
rs991247739 Chr.12: 120302442-120302442 on Build GRCh38
rs904622639 Chr.12: 120302451-120302451 on Build GRCh38
rs536629574 Chr.12: 120302543-120302543 on Build GRCh38
rs1000250371 Chr.12: 120302494-120302494 on Build GRCh38
rs757978937 Chr.12: 120302535-120302535 on Build GRCh38
rs925908066 Chr.12: 120302630-120302630 on Build GRCh38
rs1028320022 Chr.12: 120302237-120302237 on Build GRCh38
rs1055979425 Chr.12: 120302496-120302496 on Build GRCh38
rs994937423 Chr.12: 120302513-120302513 on Build GRCh38
rs934915996 Chr.12: 120302505-120302505 on Build GRCh38
rs534247577 Chr.12: 120302438-120302438 on Build GRCh38
rs371516938 Chr.12: 120302435-120302435 on Build GRCh38
rs991241536 Chr.12: 120302633-120302633 on Build GRCh38
rs34369955 Chr.12: 120302618-120302619 on Build GRCh38
rs751125740 Chr.12: 120302569-120302569 on Build GRCh38
rs778371608 Chr.12: 120302528-120302529 on Build GRCh38
rs761537584 Chr.12: 120302275-120302275 on Build GRCh38
rs367870250 Chr.12: 120302636-120302636 on Build GRCh38
rs1051594320 Chr.12: 120302370-120302370 on Build GRCh38
rs61665022 Chr.12: 120302559-120302559 on Build GRCh38
rs571876040 Chr.12: 120302395-120302395 on Build GRCh38
rs973228713 Chr.12: 120302316-120302316 on Build GRCh38
rs866145245 Chr.12: 120302509-120302509 on Build GRCh38
rs753631629 Chr.12: 120302248-120302248 on Build GRCh38
rs993045185 Chr.12: 120302230-120302230 on Build GRCh38
rs531925766 Chr.12: 120302353-120302353 on Build GRCh38
rs1014553739 Chr.12: 120302533-120302533 on Build GRCh38
rs764307000 Chr.12: 120302214-120302214 on Build GRCh38
rs181958355 Chr.12: 120302645-120302645 on Build GRCh38
rs200766343 Chr.12: 120302355-120302355 on Build GRCh38
rs915640793 Chr.12: 120302642-120302642 on Build GRCh38
rs201127747 Chr.12: 120302327-120302327 on Build GRCh38

View Full Product Details