Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121154480-121154983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P2RX7
Entrez Gene ID
5027
Gene Name
purinergic receptor P2X 7
Gene Aliases
P2X7
UniGene
Hs.729169
Gene Chromosome Location
Chr.12: 121132819-121189478 on Build GRCh38
Transcripts
NM_002562.5 NR_033948.1 NR_033949.1 NR_033950.1 NR_033951.1 NR_033952.1 NR_033953.1 NR_033954.1 NR_033955.1 NR_033956.1 XM_011538419.2 XM_011538420.2 XM_017019364.1 XM_017019365.1 XM_017019366.1 XM_017019367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5071698 c.158G>A p.R53Q Chr.12: 121154817-121154817 on Build GRCh38
6041592 c.205G>T p.A69S Chr.12: 121154864-121154864 on Build GRCh38
4484845 c.285C>T p.F95F Chr.12: 121154944-121154944 on Build GRCh38
6861661 c.171C>T p.V57V Chr.12: 121154830-121154830 on Build GRCh38
4531223 c.176G>A p.S59N Chr.12: 121154835-121154835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910391503 Chr.12: 121154780-121154780 on Build GRCh38
rs548882788 Chr.12: 121154961-121154961 on Build GRCh38
rs200957980 Chr.12: 121154756-121154756 on Build GRCh38
rs746705484 Chr.12: 121154887-121154887 on Build GRCh38
rs1018702931 Chr.12: 121154821-121154821 on Build GRCh38
rs28360443 Chr.12: 121154722-121154722 on Build GRCh38
rs202115621 Chr.12: 121154878-121154878 on Build GRCh38
rs753908144 Chr.12: 121154931-121154931 on Build GRCh38
rs775831502 Chr.12: 121154927-121154927 on Build GRCh38
rs200496294 Chr.12: 121154896-121154896 on Build GRCh38
rs200300690 Chr.12: 121154846-121154846 on Build GRCh38
rs768107697 Chr.12: 121154804-121154804 on Build GRCh38
rs202077960 Chr.12: 121154915-121154915 on Build GRCh38
rs754634787 Chr.12: 121154956-121154956 on Build GRCh38
rs940532812 Chr.12: 121154733-121154733 on Build GRCh38
rs954666339 Chr.12: 121154771-121154771 on Build GRCh38
rs898543517 Chr.12: 121154550-121154550 on Build GRCh38
rs548756208 Chr.12: 121154711-121154711 on Build GRCh38
rs201601422 Chr.12: 121154739-121154739 on Build GRCh38
rs370478786 Chr.12: 121154833-121154833 on Build GRCh38
rs199707157 Chr.12: 121154933-121154933 on Build GRCh38
rs201754454 Chr.12: 121154912-121154912 on Build GRCh38
rs957819576 Chr.12: 121154585-121154585 on Build GRCh38
rs201451288 Chr.12: 121154930-121154930 on Build GRCh38
rs756627952 Chr.12: 121154851-121154851 on Build GRCh38
rs915872551 Chr.12: 121154622-121154622 on Build GRCh38
rs760016181 Chr.12: 121154916-121154916 on Build GRCh38
rs753893226 Chr.12: 121154861-121154861 on Build GRCh38
rs778459697 Chr.12: 121154519-121154519 on Build GRCh38
rs772486470 Chr.12: 121154926-121154926 on Build GRCh38
rs751355725 Chr.12: 121154950-121154950 on Build GRCh38
rs776199891 Chr.12: 121154806-121154806 on Build GRCh38
rs942810068 Chr.12: 121154503-121154503 on Build GRCh38
rs771879719 Chr.12: 121154777-121154777 on Build GRCh38
rs200026479 Chr.12: 121154755-121154755 on Build GRCh38
rs1014231044 Chr.12: 121154666-121154666 on Build GRCh38
rs762147937 Chr.12: 121154651-121154651 on Build GRCh38
rs763374310 Chr.12: 121154809-121154809 on Build GRCh38
rs575422540 Chr.12: 121154723-121154723 on Build GRCh38
rs148167930 Chr.12: 121154541-121154541 on Build GRCh38
rs766851766 Chr.12: 121154810-121154810 on Build GRCh38
rs186488525 Chr.12: 121154860-121154860 on Build GRCh38
rs778950777 Chr.12: 121154879-121154879 on Build GRCh38
rs771374027 Chr.12: 121154908-121154908 on Build GRCh38
rs750352416 Chr.12: 121154882-121154882 on Build GRCh38
rs778581720 Chr.12: 121154770-121154770 on Build GRCh38
rs201644110 Chr.12: 121154944-121154944 on Build GRCh38
rs767838566 Chr.12: 121154830-121154830 on Build GRCh38
rs200006807 Chr.12: 121154791-121154791 on Build GRCh38
rs940080307 Chr.12: 121154935-121154935 on Build GRCh38
rs143171240 Chr.12: 121154817-121154817 on Build GRCh38
rs200965514 Chr.12: 121154869-121154869 on Build GRCh38
rs991516796 Chr.12: 121154600-121154600 on Build GRCh38
rs1000623422 Chr.12: 121154731-121154731 on Build GRCh38
rs761986892 Chr.12: 121154932-121154932 on Build GRCh38
rs145127272 Chr.12: 121154884-121154884 on Build GRCh38
rs112275589 Chr.12: 121154893-121154893 on Build GRCh38
rs28360444 Chr.12: 121154774-121154774 on Build GRCh38
rs200942168 Chr.12: 121154937-121154937 on Build GRCh38
rs746636395 Chr.12: 121154803-121154803 on Build GRCh38
rs531620812 Chr.12: 121154868-121154868 on Build GRCh38
rs376579155 Chr.12: 121154740-121154740 on Build GRCh38
rs139154098 Chr.12: 121154816-121154816 on Build GRCh38
rs17525809 Chr.12: 121154886-121154886 on Build GRCh38
rs567612383 Chr.12: 121154524-121154524 on Build GRCh38
rs138943772 Chr.12: 121154885-121154885 on Build GRCh38
rs747646614 Chr.12: 121154905-121154905 on Build GRCh38
rs200456779 Chr.12: 121154929-121154929 on Build GRCh38
rs139064140 Chr.12: 121154940-121154940 on Build GRCh38
rs112717437 Chr.12: 121154665-121154665 on Build GRCh38
rs764013575 Chr.12: 121154938-121154939 on Build GRCh38
rs757160496 Chr.12: 121154870-121154870 on Build GRCh38

View Full Product Details