Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121770806-121771300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM120B
Entrez Gene ID
144404
Gene Name
transmembrane protein 120B
Gene Aliases
-
UniGene
Hs.644504
Gene Chromosome Location
Chr.12: 121712752-121782070 on Build GRCh38
Transcripts
NM_001080825.2 XM_005253843.4 XM_017018853.1 XM_017018854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1359497 c.610C>T p.L204L Chr.12: 121770965-121770965 on Build GRCh38
4518292 c.594_595CC>TT p.(=) Chr.12: 121770949-121770950 on Build GRCh38
5573442 c.617+1G>T p.? Chr.12: 121770973-121770973 on Build GRCh38
4204067 c.601G>A p.G201R Chr.12: 121770956-121770956 on Build GRCh38
4518293 c.594_595CC>TT p.(=) Chr.12: 121770949-121770950 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766484696 Chr.12: 121770985-121770985 on Build GRCh38
rs946696716 Chr.12: 121771163-121771163 on Build GRCh38
rs575641179 Chr.12: 121770986-121770986 on Build GRCh38
rs1018520105 Chr.12: 121771279-121771279 on Build GRCh38
rs1040859885 Chr.12: 121770891-121770891 on Build GRCh38
rs534755619 Chr.12: 121770898-121770898 on Build GRCh38
rs952954003 Chr.12: 121771117-121771117 on Build GRCh38
rs773442629 Chr.12: 121770965-121770965 on Build GRCh38
rs942464758 Chr.12: 121771055-121771055 on Build GRCh38
rs998477656 Chr.12: 121771195-121771195 on Build GRCh38
rs536549611 Chr.12: 121770987-121770987 on Build GRCh38
rs765582075 Chr.12: 121770961-121770961 on Build GRCh38
rs761227394 Chr.12: 121770871-121770871 on Build GRCh38
rs11608693 Chr.12: 121771062-121771062 on Build GRCh38
rs910925643 Chr.12: 121771121-121771121 on Build GRCh38
rs892043093 Chr.12: 121771173-121771173 on Build GRCh38
rs1036996971 Chr.12: 121771100-121771100 on Build GRCh38
rs754877510 Chr.12: 121770918-121770918 on Build GRCh38
rs569065859 Chr.12: 121771278-121771278 on Build GRCh38
rs968150376 Chr.12: 121771013-121771013 on Build GRCh38
rs905894704 Chr.12: 121771187-121771187 on Build GRCh38
rs1035717132 Chr.12: 121771104-121771104 on Build GRCh38
rs528075153 Chr.12: 121771125-121771125 on Build GRCh38
rs562721388 Chr.12: 121771064-121771064 on Build GRCh38
rs930975714 Chr.12: 121770848-121770848 on Build GRCh38
rs748991352 Chr.12: 121771023-121771023 on Build GRCh38
rs956162169 Chr.12: 121770851-121770851 on Build GRCh38
rs367663330 Chr.12: 121771063-121771063 on Build GRCh38
rs550843617 Chr.12: 121771255-121771255 on Build GRCh38
rs200861818 Chr.12: 121770937-121770937 on Build GRCh38
rs1035853080 Chr.12: 121770967-121770967 on Build GRCh38
rs1052800150 Chr.12: 121771061-121771061 on Build GRCh38
rs79559664 Chr.12: 121771127-121771127 on Build GRCh38
rs34599985 Chr.12: 121771019-121771019 on Build GRCh38
rs566919388 Chr.12: 121770888-121770888 on Build GRCh38
rs952596362 Chr.12: 121771150-121771150 on Build GRCh38
rs960183311 Chr.12: 121770970-121770970 on Build GRCh38
rs777120828 Chr.12: 121770956-121770956 on Build GRCh38
rs769305240 Chr.12: 121770859-121770859 on Build GRCh38
rs773970595 Chr.12: 121770952-121770952 on Build GRCh38
rs28685863 Chr.12: 121771060-121771060 on Build GRCh38
rs751528567 Chr.12: 121770910-121770910 on Build GRCh38
rs540576216 Chr.12: 121771058-121771058 on Build GRCh38
rs34623055 Chr.12: 121771083-121771083 on Build GRCh38
rs760897409 Chr.12: 121770895-121770895 on Build GRCh38
rs1045371008 Chr.12: 121771170-121771170 on Build GRCh38
rs930984456 Chr.12: 121771020-121771020 on Build GRCh38
rs752610499 Chr.12: 121770998-121770998 on Build GRCh38
rs372468519 Chr.12: 121771019-121771019 on Build GRCh38
rs776165930 Chr.12: 121770864-121770864 on Build GRCh38
rs777587616 Chr.12: 121771022-121771022 on Build GRCh38
rs558806929 Chr.12: 121771059-121771059 on Build GRCh38
rs1030906423 Chr.12: 121770923-121770923 on Build GRCh38
rs1029818143 Chr.12: 121771200-121771200 on Build GRCh38
rs772833234 Chr.12: 121770860-121770860 on Build GRCh38
rs188387103 Chr.12: 121771118-121771118 on Build GRCh38
rs139299980 Chr.12: 121770904-121770904 on Build GRCh38
rs1005830607 Chr.12: 121771263-121771263 on Build GRCh38
rs960214127 Chr.12: 121771113-121771113 on Build GRCh38
rs556688948 Chr.12: 121771114-121771114 on Build GRCh38
rs1048637046 Chr.12: 121770853-121770853 on Build GRCh38
rs962835284 Chr.12: 121771224-121771224 on Build GRCh38
rs985665375 Chr.12: 121771211-121771211 on Build GRCh38
rs983768847 Chr.12: 121771028-121771028 on Build GRCh38
rs994746777 Chr.12: 121771249-121771249 on Build GRCh38
rs114979626 Chr.12: 121771142-121771142 on Build GRCh38
rs895871488 Chr.12: 121771225-121771225 on Build GRCh38
rs991802282 Chr.12: 121770931-121770941 on Build GRCh38
rs368861780 Chr.12: 121770938-121770938 on Build GRCh38
rs79944789 Chr.12: 121770861-121770861 on Build GRCh38
rs562585896 Chr.12: 121771110-121771119 on Build GRCh38
rs747572526 Chr.12: 121770955-121770955 on Build GRCh38
rs542106552 Chr.12: 121771115-121771115 on Build GRCh38
rs370439191 Chr.12: 121770957-121770957 on Build GRCh38
rs754659235 Chr.12: 121771213-121771213 on Build GRCh38
rs901006049 Chr.12: 121770892-121770892 on Build GRCh38
rs573285737 Chr.12: 121771029-121771029 on Build GRCh38
rs1043840685 Chr.12: 121771137-121771137 on Build GRCh38
rs1057156568 Chr.12: 121771217-121771217 on Build GRCh38
rs1037237033 Chr.12: 121770845-121770845 on Build GRCh38
rs909912970 Chr.12: 121770879-121770879 on Build GRCh38
rs1001601619 Chr.12: 121771093-121771093 on Build GRCh38
rs184767447 Chr.12: 121770903-121770903 on Build GRCh38
rs947818708 Chr.12: 121771131-121771131 on Build GRCh38
rs1052243050 Chr.12: 121771143-121771143 on Build GRCh38
rs763119520 Chr.12: 121770984-121770984 on Build GRCh38
rs560372001 Chr.12: 121771124-121771124 on Build GRCh38
rs561349532 Chr.12: 121771250-121771250 on Build GRCh38
rs375249240 Chr.12: 121770931-121770931 on Build GRCh38
rs770600129 Chr.12: 121770946-121770946 on Build GRCh38
rs1001464534 Chr.12: 121771192-121771192 on Build GRCh38
rs542469949 Chr.12: 121771220-121771220 on Build GRCh38
rs747874966 Chr.12: 121770856-121770856 on Build GRCh38
rs528852180 Chr.12: 121771065-121771065 on Build GRCh38
rs376403880 Chr.12: 121770942-121770942 on Build GRCh38
rs764563331 Chr.12: 121770887-121770887 on Build GRCh38
rs748518189 Chr.12: 121770846-121770846 on Build GRCh38
rs954384533 Chr.12: 121771209-121771209 on Build GRCh38
rs549547120 Chr.12: 121771140-121771140 on Build GRCh38

View Full Product Details