Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122184438-122184935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC43
Entrez Gene ID
254050
Gene Name
leucine rich repeat containing 43
Gene Aliases
-
UniGene
Hs.374856
Gene Chromosome Location
Chr.12: 122167719-122203471 on Build GRCh38
Transcripts
NM_001098519.1 NM_152759.4 XM_017019124.1 XM_017019125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2176487 c.326G>A p.R109K Chr.12: 122184694-122184694 on Build GRCh38
2176484 c.253C>T p.L85L Chr.12: 122184621-122184621 on Build GRCh38
1191678 c.274C>T p.H92Y Chr.12: 122184642-122184642 on Build GRCh38
2176489 c.386G>A p.R129Q Chr.12: 122184754-122184754 on Build GRCh38
1127527 c.169C>T p.P57S Chr.12: 122184537-122184537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753706800 Chr.12: 122184684-122184684 on Build GRCh38
rs200132936 Chr.12: 122184608-122184608 on Build GRCh38
rs201504564 Chr.12: 122184557-122184557 on Build GRCh38
rs544082638 Chr.12: 122184850-122184850 on Build GRCh38
rs200547603 Chr.12: 122184822-122184822 on Build GRCh38
rs371279456 Chr.12: 122184887-122184887 on Build GRCh38
rs866461267 Chr.12: 122184598-122184598 on Build GRCh38
rs374939130 Chr.12: 122184795-122184795 on Build GRCh38
rs1040044921 Chr.12: 122184764-122184764 on Build GRCh38
rs752770340 Chr.12: 122184696-122184696 on Build GRCh38
rs761218989 Chr.12: 122184731-122184731 on Build GRCh38
rs780102378 Chr.12: 122184480-122184480 on Build GRCh38
rs926958702 Chr.12: 122184697-122184697 on Build GRCh38
rs762531779 Chr.12: 122184567-122184567 on Build GRCh38
rs942541007 Chr.12: 122184907-122184907 on Build GRCh38
rs887228102 Chr.12: 122184810-122184811 on Build GRCh38
rs774506201 Chr.12: 122184528-122184528 on Build GRCh38
rs760336260 Chr.12: 122184675-122184675 on Build GRCh38
rs1050982943 Chr.12: 122184583-122184583 on Build GRCh38
rs574275977 Chr.12: 122184894-122184894 on Build GRCh38
rs773376494 Chr.12: 122184846-122184846 on Build GRCh38
rs763722269 Chr.12: 122184576-122184576 on Build GRCh38
rs760408826 Chr.12: 122184750-122184750 on Build GRCh38
rs772802777 Chr.12: 122184804-122184804 on Build GRCh38
rs970934978 Chr.12: 122184580-122184580 on Build GRCh38
rs990260418 Chr.12: 122184872-122184872 on Build GRCh38
rs113382732 Chr.12: 122184581-122184581 on Build GRCh38
rs768770730 Chr.12: 122184527-122184527 on Build GRCh38
rs749526407 Chr.12: 122184585-122184587 on Build GRCh38
rs771378312 Chr.12: 122184640-122184640 on Build GRCh38
rs771628026 Chr.12: 122184734-122184734 on Build GRCh38
rs945974799 Chr.12: 122184864-122184864 on Build GRCh38
rs182755986 Chr.12: 122184820-122184820 on Build GRCh38
rs71453589 Chr.12: 122184857-122184858 on Build GRCh38
rs879181284 Chr.12: 122184625-122184625 on Build GRCh38
rs1033034010 Chr.12: 122184863-122184863 on Build GRCh38
rs763082790 Chr.12: 122184876-122184876 on Build GRCh38
rs775258822 Chr.12: 122184734-122184736 on Build GRCh38
rs777230754 Chr.12: 122184646-122184646 on Build GRCh38
rs771445471 Chr.12: 122184802-122184802 on Build GRCh38
rs3741449 Chr.12: 122184865-122184865 on Build GRCh38
rs765154228 Chr.12: 122184761-122184761 on Build GRCh38
rs774015043 Chr.12: 122184667-122184667 on Build GRCh38
rs904314364 Chr.12: 122184707-122184707 on Build GRCh38
rs549848383 Chr.12: 122184765-122184765 on Build GRCh38
rs761380375 Chr.12: 122184587-122184587 on Build GRCh38
rs779830177 Chr.12: 122184602-122184602 on Build GRCh38
rs771203566 Chr.12: 122184596-122184597 on Build GRCh38
rs761159183 Chr.12: 122184556-122184556 on Build GRCh38
rs776333065 Chr.12: 122184819-122184819 on Build GRCh38
rs963544074 Chr.12: 122184788-122184788 on Build GRCh38
rs746845862 Chr.12: 122184736-122184737 on Build GRCh38
rs755987347 Chr.12: 122184479-122184479 on Build GRCh38
rs750136239 Chr.12: 122184674-122184674 on Build GRCh38
rs570255416 Chr.12: 122184521-122184521 on Build GRCh38
rs371653161 Chr.12: 122184768-122184768 on Build GRCh38
rs138682950 Chr.12: 122184709-122184709 on Build GRCh38
rs752737247 Chr.12: 122184762-122184762 on Build GRCh38
rs770398014 Chr.12: 122184722-122184722 on Build GRCh38
rs781609921 Chr.12: 122184771-122184771 on Build GRCh38
rs192088829 Chr.12: 122184786-122184786 on Build GRCh38
rs749332423 Chr.12: 122184526-122184526 on Build GRCh38
rs895229367 Chr.12: 122184747-122184747 on Build GRCh38
rs921719408 Chr.12: 122184817-122184817 on Build GRCh38
rs201851503 Chr.12: 122184694-122184694 on Build GRCh38
rs199909089 Chr.12: 122184634-122184634 on Build GRCh38
rs373684289 Chr.12: 122184699-122184699 on Build GRCh38
rs372181783 Chr.12: 122184754-122184754 on Build GRCh38
rs754921318 Chr.12: 122184689-122184689 on Build GRCh38
rs200849229 Chr.12: 122184769-122184769 on Build GRCh38
rs567947496 Chr.12: 122184794-122184794 on Build GRCh38
rs778967458 Chr.12: 122184621-122184621 on Build GRCh38
rs746235604 Chr.12: 122184778-122184778 on Build GRCh38
rs768218856 Chr.12: 122184666-122184666 on Build GRCh38
rs766433910 Chr.12: 122184908-122184908 on Build GRCh38
rs748399342 Chr.12: 122184529-122184529 on Build GRCh38
rs746550306 Chr.12: 122184815-122184815 on Build GRCh38
rs777985499 Chr.12: 122184520-122184521 on Build GRCh38
rs1002433951 Chr.12: 122184824-122184824 on Build GRCh38
rs570495541 Chr.12: 122184716-122184716 on Build GRCh38
rs200229491 Chr.12: 122184549-122184549 on Build GRCh38
rs983696964 Chr.12: 122184773-122184773 on Build GRCh38
rs746323946 Chr.12: 122184721-122184721 on Build GRCh38
rs189080619 Chr.12: 122184547-122184547 on Build GRCh38
rs367754491 Chr.12: 122184662-122184662 on Build GRCh38
rs370251743 Chr.12: 122184677-122184677 on Build GRCh38
rs753792682 Chr.12: 122184607-122184607 on Build GRCh38
rs777320283 Chr.12: 122184563-122184563 on Build GRCh38
rs750177453 Chr.12: 122184597-122184597 on Build GRCh38
rs986550080 Chr.12: 122184898-122184898 on Build GRCh38
rs768938779 Chr.12: 122184742-122184742 on Build GRCh38
rs200583842 Chr.12: 122184639-122184639 on Build GRCh38
rs368439340 Chr.12: 122184818-122184818 on Build GRCh38
rs912375333 Chr.12: 122184904-122184904 on Build GRCh38
rs372375067 Chr.12: 122184708-122184708 on Build GRCh38
rs61732476 Chr.12: 122184633-122184633 on Build GRCh38
rs368951248 Chr.12: 122184748-122184748 on Build GRCh38
rs767289151 Chr.12: 122184596-122184596 on Build GRCh38
rs931886904 Chr.12: 122184622-122184622 on Build GRCh38
rs767042268 Chr.12: 122184670-122184670 on Build GRCh38
rs959330706 Chr.12: 122184534-122184534 on Build GRCh38
rs1055534263 Chr.12: 122184896-122184896 on Build GRCh38
rs992326617 Chr.12: 122184648-122184648 on Build GRCh38
rs902627097 Chr.12: 122184504-122184504 on Build GRCh38
rs576077334 Chr.12: 122184828-122184828 on Build GRCh38
rs199554806 Chr.12: 122184763-122184763 on Build GRCh38
rs951531195 Chr.12: 122184647-122184647 on Build GRCh38
rs538114744 Chr.12: 122184753-122184753 on Build GRCh38

View Full Product Details