Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123620931-123621432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX55
Entrez Gene ID
57696
Gene Name
DEAD-box helicase 55
Gene Aliases
-
UniGene
Hs.286173
Gene Chromosome Location
Chr.12: 123602077-123620943 on Build GRCh38
Transcripts
NM_020936.2 NR_135104.1 NR_135105.1 XM_017019710.1 XM_017019711.1 XM_017019712.1 XM_017019713.1 XM_017019714.1 XM_017019715.1 XM_017019716.1 XM_017019717.1 XM_017019718.1 XM_017019719.1 XM_017019720.1 NM_001414.3

Gene Symbol
EIF2B1
Entrez Gene ID
1967
Gene Name
eukaryotic translation initiation factor 2B subunit alpha
Gene Aliases
EIF2B, EIF2BA
UniGene
Hs.741273
Gene Chromosome Location
Chr.12: 123621023-123633776 on Build GRCh38
Transcripts
NM_020936.2 NR_135104.1 NR_135105.1 XM_017019710.1 XM_017019711.1 XM_017019712.1 XM_017019713.1 XM_017019714.1 XM_017019715.1 XM_017019716.1 XM_017019717.1 XM_017019718.1 XM_017019719.1 XM_017019720.1 NM_001414.3

SNP Detail

SNP ID Chromosome Location
rs1035582102 Chr.12: 123621385-123621385 on Build GRCh38
rs1039731499 Chr.12: 123621116-123621116 on Build GRCh38
rs959918171 Chr.12: 123621324-123621324 on Build GRCh38
rs890070779 Chr.12: 123621118-123621118 on Build GRCh38
rs948627119 Chr.12: 123621028-123621028 on Build GRCh38
rs886049050 Chr.12: 123621331-123621331 on Build GRCh38
rs1052810183 Chr.12: 123621076-123621076 on Build GRCh38
rs934505838 Chr.12: 123621307-123621307 on Build GRCh38
rs10846532 Chr.12: 123621025-123621025 on Build GRCh38
rs187113416 Chr.12: 123621103-123621103 on Build GRCh38
rs916556316 Chr.12: 123621273-123621273 on Build GRCh38
rs138494784 Chr.12: 123621235-123621235 on Build GRCh38
rs1050448 Chr.12: 123621378-123621378 on Build GRCh38
rs571156978 Chr.12: 123621097-123621097 on Build GRCh38
rs542350295 Chr.12: 123621182-123621182 on Build GRCh38
rs1025390999 Chr.12: 123621332-123621332 on Build GRCh38
rs1050449 Chr.12: 123621361-123621361 on Build GRCh38
rs763860706 Chr.12: 123621243-123621244 on Build GRCh38
rs141505281 Chr.12: 123621402-123621402 on Build GRCh38
rs543958349 Chr.12: 123621308-123621308 on Build GRCh38
rs992605750 Chr.12: 123620990-123620990 on Build GRCh38
rs572343813 Chr.12: 123621250-123621250 on Build GRCh38
rs1050443 Chr.12: 123621401-123621401 on Build GRCh38
rs1035126065 Chr.12: 123621222-123621222 on Build GRCh38
rs960748477 Chr.12: 123620967-123620967 on Build GRCh38
rs1052583400 Chr.12: 123621183-123621183 on Build GRCh38
rs9919 Chr.12: 123621292-123621292 on Build GRCh38
rs574912798 Chr.12: 123621114-123621114 on Build GRCh38
rs150711091 Chr.12: 123621404-123621404 on Build GRCh38
rs2707 Chr.12: 123621091-123621091 on Build GRCh38
rs1048959916 Chr.12: 123621057-123621057 on Build GRCh38
rs933722140 Chr.12: 123621331-123621331 on Build GRCh38
rs959969390 Chr.12: 123620988-123620988 on Build GRCh38
rs1034046440 Chr.12: 123621305-123621305 on Build GRCh38
rs1005797340 Chr.12: 123621125-123621125 on Build GRCh38
rs1042899844 Chr.12: 123621030-123621030 on Build GRCh38
rs886049049 Chr.12: 123621116-123621116 on Build GRCh38
rs771167598 Chr.12: 123621196-123621196 on Build GRCh38
rs547879408 Chr.12: 123621341-123621341 on Build GRCh38
rs7137332 Chr.12: 123621301-123621301 on Build GRCh38
rs961054040 Chr.12: 123621248-123621248 on Build GRCh38
rs546204360 Chr.12: 123621270-123621270 on Build GRCh38
rs529880752 Chr.12: 123621334-123621334 on Build GRCh38
rs1009958383 Chr.12: 123621219-123621219 on Build GRCh38
rs994305144 Chr.12: 123621205-123621205 on Build GRCh38
rs1052858492 Chr.12: 123621319-123621319 on Build GRCh38
rs547744395 Chr.12: 123620960-123620960 on Build GRCh38
rs951225305 Chr.12: 123621333-123621333 on Build GRCh38
rs113041500 Chr.12: 123621330-123621330 on Build GRCh38
rs918432019 Chr.12: 123621012-123621012 on Build GRCh38
rs372071677 Chr.12: 123621348-123621348 on Build GRCh38
rs1017151704 Chr.12: 123621139-123621139 on Build GRCh38
rs901413445 Chr.12: 123621154-123621154 on Build GRCh38
rs566337000 Chr.12: 123621024-123621024 on Build GRCh38
rs998451866 Chr.12: 123621158-123621158 on Build GRCh38
rs927197065 Chr.12: 123620980-123620980 on Build GRCh38
rs191665709 Chr.12: 123621147-123621147 on Build GRCh38
rs934454881 Chr.12: 123621075-123621075 on Build GRCh38
rs775222743 Chr.12: 123621068-123621068 on Build GRCh38

View Full Product Details