Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 112647093-112647588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11AUN
Entrez Gene ID
400165
Gene Name
ATP11A upstream neighbor
Gene Aliases
C13orf35, SMABLO1
UniGene
Hs.591213
Gene Chromosome Location
Chr.13: 112647044-112684497 on Build GRCh38
Transcripts
NM_207440.2

SNP Detail

SNP ID Chromosome Location
rs538476917 Chr.13: 112647392-112647392 on Build GRCh38
rs1019673843 Chr.13: 112647461-112647461 on Build GRCh38
rs937374152 Chr.13: 112647325-112647325 on Build GRCh38
rs933589067 Chr.13: 112647268-112647268 on Build GRCh38
rs557650060 Chr.13: 112647447-112647447 on Build GRCh38
rs928812952 Chr.13: 112647348-112647348 on Build GRCh38
rs180917315 Chr.13: 112647479-112647479 on Build GRCh38
rs540196651 Chr.13: 112647121-112647121 on Build GRCh38
rs968415601 Chr.13: 112647158-112647158 on Build GRCh38
rs908052740 Chr.13: 112647288-112647288 on Build GRCh38
rs1046361246 Chr.13: 112647144-112647144 on Build GRCh38
rs1036054335 Chr.13: 112647120-112647120 on Build GRCh38
rs1019203566 Chr.13: 112647251-112647251 on Build GRCh38
rs1008743358 Chr.13: 112647417-112647417 on Build GRCh38
rs556663043 Chr.13: 112647404-112647404 on Build GRCh38
rs898780788 Chr.13: 112647376-112647376 on Build GRCh38
rs4907733 Chr.13: 112647475-112647475 on Build GRCh38
rs981495298 Chr.13: 112647543-112647543 on Build GRCh38
rs548171693 Chr.13: 112647295-112647295 on Build GRCh38
rs9603923 Chr.13: 112647274-112647274 on Build GRCh38
rs531446120 Chr.13: 112647205-112647205 on Build GRCh38
rs543886483 Chr.13: 112647254-112647254 on Build GRCh38
rs78311413 Chr.13: 112647190-112647190 on Build GRCh38
rs185323007 Chr.13: 112647517-112647517 on Build GRCh38
rs1028317980 Chr.13: 112647244-112647244 on Build GRCh38
rs757766566 Chr.13: 112647407-112647407 on Build GRCh38
rs1057140774 Chr.13: 112647339-112647339 on Build GRCh38
rs954308252 Chr.13: 112647245-112647245 on Build GRCh38
rs562259194 Chr.13: 112647262-112647262 on Build GRCh38
rs548138381 Chr.13: 112647229-112647229 on Build GRCh38
rs915577778 Chr.13: 112647337-112647337 on Build GRCh38
rs540113438 Chr.13: 112647504-112647504 on Build GRCh38
rs527978427 Chr.13: 112647347-112647347 on Build GRCh38
rs995534244 Chr.13: 112647238-112647238 on Build GRCh38
rs535439078 Chr.13: 112647472-112647472 on Build GRCh38
rs907331167 Chr.13: 112647170-112647170 on Build GRCh38
rs35800964 Chr.13: 112647504-112647505 on Build GRCh38
rs781757339 Chr.13: 112647469-112647469 on Build GRCh38
rs924556812 Chr.13: 112647289-112647289 on Build GRCh38
rs779647658 Chr.13: 112647505-112647505 on Build GRCh38
rs975013278 Chr.13: 112647249-112647249 on Build GRCh38
rs368316167 Chr.13: 112647143-112647143 on Build GRCh38
rs1010452087 Chr.13: 112647122-112647122 on Build GRCh38
rs553134151 Chr.13: 112647210-112647210 on Build GRCh38
rs576594338 Chr.13: 112647551-112647551 on Build GRCh38
rs116468782 Chr.13: 112647353-112647353 on Build GRCh38
rs187953640 Chr.13: 112647145-112647145 on Build GRCh38
rs995754184 Chr.13: 112647382-112647382 on Build GRCh38
rs112433197 Chr.13: 112647315-112647315 on Build GRCh38
rs948478502 Chr.13: 112647343-112647343 on Build GRCh38
rs115623242 Chr.13: 112647415-112647415 on Build GRCh38
rs140007433 Chr.13: 112647518-112647518 on Build GRCh38
rs986863303 Chr.13: 112647250-112647250 on Build GRCh38
rs750157043 Chr.13: 112647316-112647316 on Build GRCh38
rs957323545 Chr.13: 112647500-112647500 on Build GRCh38
rs755637842 Chr.13: 112647483-112647483 on Build GRCh38
rs573779 Chr.13: 112647352-112647352 on Build GRCh38
rs535801717 Chr.13: 112647403-112647403 on Build GRCh38

View Full Product Details