Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20560095-20560596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNASE9
Entrez Gene ID
390443
Gene Name
ribonuclease A family member 9 (inactive)
Gene Aliases
HEL128, RAK1, h461
UniGene
Hs.533972
Gene Chromosome Location
Chr.14: 20556093-20560931 on Build GRCh38
Transcripts
NM_001001673.3 NM_001110356.1 NM_001110357.1 NM_001110358.1 NM_001110359.1 NM_001110360.1 NM_001110361.1 NM_001289110.1

SNP Detail

SNP ID Chromosome Location
rs925656244 Chr.14: 20560137-20560137 on Build GRCh38
rs944367987 Chr.14: 20560527-20560527 on Build GRCh38
rs1037136930 Chr.14: 20560390-20560390 on Build GRCh38
rs891023688 Chr.14: 20560148-20560153 on Build GRCh38
rs957363530 Chr.14: 20560523-20560523 on Build GRCh38
rs144240632 Chr.14: 20560407-20560407 on Build GRCh38
rs921311446 Chr.14: 20560356-20560357 on Build GRCh38
rs1013862925 Chr.14: 20560367-20560367 on Build GRCh38
rs944667977 Chr.14: 20560244-20560244 on Build GRCh38
rs999110387 Chr.14: 20560514-20560514 on Build GRCh38
rs1031669873 Chr.14: 20560324-20560324 on Build GRCh38
rs1054819493 Chr.14: 20560439-20560439 on Build GRCh38
rs575383268 Chr.14: 20560428-20560455 on Build GRCh38
rs1243643 Chr.14: 20560344-20560344 on Build GRCh38
rs988546010 Chr.14: 20560466-20560466 on Build GRCh38
rs1049142716 Chr.14: 20560303-20560303 on Build GRCh38
rs932493096 Chr.14: 20560363-20560363 on Build GRCh38
rs543291932 Chr.14: 20560274-20560274 on Build GRCh38
rs907488328 Chr.14: 20560504-20560504 on Build GRCh38
rs995207702 Chr.14: 20560256-20560256 on Build GRCh38
rs930940617 Chr.14: 20560429-20560429 on Build GRCh38
rs747496885 Chr.14: 20560501-20560501 on Build GRCh38
rs960979941 Chr.14: 20560348-20560348 on Build GRCh38
rs561447156 Chr.14: 20560284-20560284 on Build GRCh38
rs1052746680 Chr.14: 20560203-20560203 on Build GRCh38
rs1033355649 Chr.14: 20560271-20560271 on Build GRCh38
rs1032308062 Chr.14: 20560522-20560523 on Build GRCh38
rs913163873 Chr.14: 20560221-20560221 on Build GRCh38
rs935653464 Chr.14: 20560169-20560169 on Build GRCh38
rs565137436 Chr.14: 20560443-20560443 on Build GRCh38
rs912924870 Chr.14: 20560469-20560469 on Build GRCh38
rs971660680 Chr.14: 20560300-20560300 on Build GRCh38
rs925709081 Chr.14: 20560428-20560428 on Build GRCh38
rs900372223 Chr.14: 20560276-20560276 on Build GRCh38
rs1014850588 Chr.14: 20560347-20560347 on Build GRCh38
rs1024306749 Chr.14: 20560388-20560388 on Build GRCh38
rs1024524692 Chr.14: 20560293-20560293 on Build GRCh38
rs912491835 Chr.14: 20560387-20560387 on Build GRCh38
rs549180739 Chr.14: 20560329-20560329 on Build GRCh38
rs932442185 Chr.14: 20560129-20560129 on Build GRCh38
rs1036680193 Chr.14: 20560237-20560237 on Build GRCh38
rs772857501 Chr.14: 20560376-20560379 on Build GRCh38
rs969589004 Chr.14: 20560406-20560406 on Build GRCh38
rs1051013358 Chr.14: 20560140-20560140 on Build GRCh38
rs887824456 Chr.14: 20560325-20560325 on Build GRCh38

View Full Product Details