Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15440330-15440729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTRC
Entrez Gene ID
11330
Gene Name
chymotrypsin C
Gene Aliases
CLCR, ELA4
UniGene
Hs.631869
Gene Chromosome Location
Chr.1: 15438442-15449247 on Build GRCh38
Transcripts
NM_007272.2 XM_011540550.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
424186 c.189G>C p.L63F Chr.1: 15440549-15440549 on Build GRCh38
3475812 c.135C>T p.I45I Chr.1: 15440495-15440495 on Build GRCh38
4142866 c.180C>T p.G60G Chr.1: 15440540-15440540 on Build GRCh38
897573 c.204C>T p.F68F Chr.1: 15440564-15440564 on Build GRCh38
1335135 c.82G>A p.A28T Chr.1: 15440341-15440341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770423335 Chr.1: 15440483-15440483 on Build GRCh38
rs761118020 Chr.1: 15440517-15440517 on Build GRCh38
rs1009858072 Chr.1: 15440670-15440670 on Build GRCh38
rs545837377 Chr.1: 15440635-15440635 on Build GRCh38
rs1057381063 Chr.1: 15440630-15440630 on Build GRCh38
rs773776887 Chr.1: 15440634-15440634 on Build GRCh38
rs559326793 Chr.1: 15440521-15440521 on Build GRCh38
rs750844200 Chr.1: 15440422-15440422 on Build GRCh38
rs767242562 Chr.1: 15440547-15440550 on Build GRCh38
rs552041210 Chr.1: 15440662-15440662 on Build GRCh38
rs763886872 Chr.1: 15440613-15440613 on Build GRCh38
rs772186675 Chr.1: 15440473-15440473 on Build GRCh38
rs755134647 Chr.1: 15440437-15440437 on Build GRCh38
rs769324644 Chr.1: 15440499-15440499 on Build GRCh38
rs754126755 Chr.1: 15440597-15440597 on Build GRCh38
rs887666879 Chr.1: 15440626-15440626 on Build GRCh38
rs515726209 Chr.1: 15440577-15440577 on Build GRCh38
rs267598094 Chr.1: 15440513-15440513 on Build GRCh38
rs370612719 Chr.1: 15440406-15440406 on Build GRCh38
rs497078 Chr.1: 15440540-15440540 on Build GRCh38
rs111996339 Chr.1: 15440445-15440445 on Build GRCh38
rs755756826 Chr.1: 15440629-15440629 on Build GRCh38
rs377475504 Chr.1: 15440591-15440591 on Build GRCh38
rs368277420 Chr.1: 15440639-15440639 on Build GRCh38
rs769482036 Chr.1: 15440541-15440541 on Build GRCh38
rs757098417 Chr.1: 15440536-15440536 on Build GRCh38
rs910219514 Chr.1: 15440586-15440586 on Build GRCh38
rs764525500 Chr.1: 15440399-15440399 on Build GRCh38
rs979580964 Chr.1: 15440677-15440677 on Build GRCh38
rs772513025 Chr.1: 15440562-15440562 on Build GRCh38
rs993807060 Chr.1: 15440416-15440416 on Build GRCh38
rs746890215 Chr.1: 15440480-15440480 on Build GRCh38
rs79256239 Chr.1: 15440672-15440672 on Build GRCh38
rs1047580748 Chr.1: 15440431-15440431 on Build GRCh38
rs560916216 Chr.1: 15440572-15440572 on Build GRCh38
rs1005668610 Chr.1: 15440438-15440438 on Build GRCh38
rs371127925 Chr.1: 15440625-15440625 on Build GRCh38
rs201904257 Chr.1: 15440434-15440434 on Build GRCh38
rs778956887 Chr.1: 15440439-15440439 on Build GRCh38
rs748069207 Chr.1: 15440461-15440461 on Build GRCh38
rs896477812 Chr.1: 15440663-15440663 on Build GRCh38
rs757180999 Chr.1: 15440624-15440624 on Build GRCh38
rs112086354 Chr.1: 15440661-15440661 on Build GRCh38
rs746721892 Chr.1: 15440534-15440534 on Build GRCh38
rs759090519 Chr.1: 15440397-15440397 on Build GRCh38
rs78247007 Chr.1: 15440505-15440505 on Build GRCh38
rs760911831 Chr.1: 15440565-15440565 on Build GRCh38
rs773793601 Chr.1: 15440564-15440564 on Build GRCh38
rs748861167 Chr.1: 15440557-15440557 on Build GRCh38
rs901779500 Chr.1: 15440391-15440391 on Build GRCh38
rs115742618 Chr.1: 15440628-15440628 on Build GRCh38
rs756273786 Chr.1: 15440428-15440428 on Build GRCh38
rs77373944 Chr.1: 15440516-15440516 on Build GRCh38
rs1042720496 Chr.1: 15440604-15440604 on Build GRCh38
rs767834265 Chr.1: 15440421-15440421 on Build GRCh38
rs536812916 Chr.1: 15440503-15440503 on Build GRCh38
rs1012984310 Chr.1: 15440430-15440430 on Build GRCh38
rs1017712066 Chr.1: 15440684-15440684 on Build GRCh38
rs774966069 Chr.1: 15440545-15440545 on Build GRCh38
rs1002224359 Chr.1: 15440511-15440511 on Build GRCh38
rs968303542 Chr.1: 15440673-15440673 on Build GRCh38
rs200910331 Chr.1: 15440533-15440533 on Build GRCh38
rs777418401 Chr.1: 15440474-15440474 on Build GRCh38
rs576621137 Chr.1: 15440507-15440507 on Build GRCh38
rs765600406 Chr.1: 15440660-15440660 on Build GRCh38
rs140059353 Chr.1: 15440576-15440576 on Build GRCh38
rs758920912 Chr.1: 15440494-15440494 on Build GRCh38
rs758564235 Chr.1: 15440611-15440611 on Build GRCh38
rs184977421 Chr.1: 15440362-15440362 on Build GRCh38
rs200859966 Chr.1: 15440632-15440632 on Build GRCh38
rs756090858 Chr.1: 15440522-15440522 on Build GRCh38
rs121909294 Chr.1: 15440524-15440524 on Build GRCh38
rs766739002 Chr.1: 15440570-15440570 on Build GRCh38
rs768248232 Chr.1: 15440633-15440633 on Build GRCh38
rs751545869 Chr.1: 15440622-15440622 on Build GRCh38
rs373205674 Chr.1: 15440435-15440435 on Build GRCh38
rs770675516 Chr.1: 15440368-15440368 on Build GRCh38
rs762121956 Chr.1: 15440407-15440407 on Build GRCh38
rs758406950 Chr.1: 15440528-15440528 on Build GRCh38
rs201395459 Chr.1: 15440462-15440462 on Build GRCh38
rs866213803 Chr.1: 15440530-15440530 on Build GRCh38
rs777132640 Chr.1: 15440640-15440640 on Build GRCh38
rs145868278 Chr.1: 15440369-15440369 on Build GRCh38
rs553816883 Chr.1: 15440349-15440349 on Build GRCh38

View Full Product Details