Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15462641-15463144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELA2A
Entrez Gene ID
63036
Gene Name
chymotrypsin like elastase family member 2A
Gene Aliases
ELA2A, PE-1
UniGene
Hs.631866
Gene Chromosome Location
Chr.1: 15456728-15472091 on Build GRCh38
Transcripts
NM_033440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4671398 c.279G>A p.A93A Chr.1: 15462784-15462784 on Build GRCh38
3788869 c.243C>T p.Y81Y Chr.1: 15462748-15462748 on Build GRCh38
378583 c.232T>A p.S78T Chr.1: 15462737-15462737 on Build GRCh38
1985573 c.273C>T p.Y91Y Chr.1: 15462778-15462778 on Build GRCh38
5968892 c.356+1G>A p.? Chr.1: 15462862-15462862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771741733 Chr.1: 15462803-15462803 on Build GRCh38
rs967957338 Chr.1: 15462989-15462989 on Build GRCh38
rs889492833 Chr.1: 15463098-15463098 on Build GRCh38
rs1006789466 Chr.1: 15462798-15462798 on Build GRCh38
rs529396199 Chr.1: 15462745-15462745 on Build GRCh38
rs770452292 Chr.1: 15462894-15462894 on Build GRCh38
rs538432757 Chr.1: 15463072-15463072 on Build GRCh38
rs767995731 Chr.1: 15462784-15462784 on Build GRCh38
rs757901311 Chr.1: 15462862-15462862 on Build GRCh38
rs367677257 Chr.1: 15462812-15462812 on Build GRCh38
rs759151072 Chr.1: 15462836-15462836 on Build GRCh38
rs545240114 Chr.1: 15462890-15462890 on Build GRCh38
rs3815791 Chr.1: 15463050-15463050 on Build GRCh38
rs960809069 Chr.1: 15463012-15463012 on Build GRCh38
rs148630095 Chr.1: 15462752-15462752 on Build GRCh38
rs527616408 Chr.1: 15462925-15462925 on Build GRCh38
rs138061779 Chr.1: 15462705-15462705 on Build GRCh38
rs375040985 Chr.1: 15462790-15462790 on Build GRCh38
rs902008363 Chr.1: 15462950-15462950 on Build GRCh38
rs558493952 Chr.1: 15462758-15462758 on Build GRCh38
rs752094557 Chr.1: 15462861-15462861 on Build GRCh38
rs571952876 Chr.1: 15462975-15462975 on Build GRCh38
rs750869865 Chr.1: 15462799-15462800 on Build GRCh38
rs576153482 Chr.1: 15463071-15463071 on Build GRCh38
rs751420468 Chr.1: 15462693-15462693 on Build GRCh38
rs1049360969 Chr.1: 15462931-15462931 on Build GRCh38
rs558358974 Chr.1: 15463094-15463094 on Build GRCh38
rs764702337 Chr.1: 15462705-15462705 on Build GRCh38
rs540212600 Chr.1: 15462932-15462932 on Build GRCh38
rs1029321283 Chr.1: 15462783-15462783 on Build GRCh38
rs765802257 Chr.1: 15462769-15462769 on Build GRCh38
rs779012449 Chr.1: 15462883-15462883 on Build GRCh38
rs764020357 Chr.1: 15462755-15462755 on Build GRCh38
rs767169289 Chr.1: 15462698-15462698 on Build GRCh38
rs111335133 Chr.1: 15462667-15462667 on Build GRCh38
rs755854436 Chr.1: 15462766-15462766 on Build GRCh38
rs573956736 Chr.1: 15463120-15463120 on Build GRCh38
rs756230930 Chr.1: 15462865-15462865 on Build GRCh38
rs756477005 Chr.1: 15462875-15462876 on Build GRCh38
rs746890964 Chr.1: 15462942-15462942 on Build GRCh38
rs190173301 Chr.1: 15462751-15462751 on Build GRCh38
rs753239633 Chr.1: 15462827-15462827 on Build GRCh38
rs766070348 Chr.1: 15462820-15462820 on Build GRCh38
rs146661868 Chr.1: 15462765-15462765 on Build GRCh38
rs763858920 Chr.1: 15462682-15462682 on Build GRCh38
rs564853506 Chr.1: 15462917-15462917 on Build GRCh38
rs747392824 Chr.1: 15462728-15462728 on Build GRCh38
rs960855749 Chr.1: 15462856-15462856 on Build GRCh38
rs776751440 Chr.1: 15462740-15462740 on Build GRCh38
rs3815788 Chr.1: 15462951-15462951 on Build GRCh38
rs1036568860 Chr.1: 15462948-15462949 on Build GRCh38
rs144479223 Chr.1: 15463114-15463114 on Build GRCh38
rs781660282 Chr.1: 15462784-15462784 on Build GRCh38
rs146502483 Chr.1: 15462795-15462795 on Build GRCh38
rs780360200 Chr.1: 15462911-15462912 on Build GRCh38
rs748074322 Chr.1: 15462886-15462886 on Build GRCh38
rs141105892 Chr.1: 15462779-15462779 on Build GRCh38
rs547412942 Chr.1: 15463008-15463008 on Build GRCh38
rs191777690 Chr.1: 15463010-15463010 on Build GRCh38
rs771232071 Chr.1: 15462735-15462735 on Build GRCh38
rs923490582 Chr.1: 15463065-15463065 on Build GRCh38
rs747775883 Chr.1: 15462996-15462996 on Build GRCh38
rs540971068 Chr.1: 15463113-15463113 on Build GRCh38
rs531834108 Chr.1: 15462969-15462969 on Build GRCh38
rs752216098 Chr.1: 15462765-15462765 on Build GRCh38
rs552066572 Chr.1: 15462970-15462970 on Build GRCh38
rs1037193659 Chr.1: 15462764-15462764 on Build GRCh38
rs3815789 Chr.1: 15462955-15462955 on Build GRCh38
rs201222086 Chr.1: 15462703-15462703 on Build GRCh38
rs3815787 Chr.1: 15462802-15462802 on Build GRCh38
rs534522393 Chr.1: 15463006-15463006 on Build GRCh38
rs948008389 Chr.1: 15462896-15462896 on Build GRCh38
rs560160322 Chr.1: 15462941-15462941 on Build GRCh38
rs772051136 Chr.1: 15462887-15462887 on Build GRCh38
rs992257106 Chr.1: 15463013-15463013 on Build GRCh38
rs867241766 Chr.1: 15462750-15462750 on Build GRCh38
rs376499556 Chr.1: 15463090-15463090 on Build GRCh38
rs750800980 Chr.1: 15462864-15462864 on Build GRCh38
rs768691083 Chr.1: 15462872-15462872 on Build GRCh38
rs3815790 Chr.1: 15463045-15463045 on Build GRCh38
rs774073473 Chr.1: 15462756-15462756 on Build GRCh38
rs531628579 Chr.1: 15462662-15462662 on Build GRCh38
rs757767208 Chr.1: 15462782-15462783 on Build GRCh38
rs753395373 Chr.1: 15462776-15462776 on Build GRCh38
rs754163336 Chr.1: 15462723-15462723 on Build GRCh38
rs573754320 Chr.1: 15462791-15462791 on Build GRCh38
rs41307796 Chr.1: 15462718-15462718 on Build GRCh38
rs909206370 Chr.1: 15463048-15463048 on Build GRCh38
rs371892099 Chr.1: 15462866-15462866 on Build GRCh38
rs201476668 Chr.1: 15462778-15462778 on Build GRCh38
rs764883532 Chr.1: 15462851-15462851 on Build GRCh38
rs768529968 Chr.1: 15462800-15462800 on Build GRCh38
rs749897239 Chr.1: 15462699-15462699 on Build GRCh38
rs1022170260 Chr.1: 15462976-15462976 on Build GRCh38
rs916645358 Chr.1: 15463016-15463016 on Build GRCh38
rs772826357 Chr.1: 15462888-15462888 on Build GRCh38
rs972705326 Chr.1: 15463062-15463062 on Build GRCh38
rs756833751 Chr.1: 15462696-15462696 on Build GRCh38
rs978593808 Chr.1: 15462676-15462676 on Build GRCh38
rs572006598 Chr.1: 15463095-15463095 on Build GRCh38
rs948307921 Chr.1: 15462721-15462721 on Build GRCh38
rs3815786 Chr.1: 15462681-15462681 on Build GRCh38
rs749380925 Chr.1: 15462796-15462796 on Build GRCh38
rs150179544 Chr.1: 15462809-15462809 on Build GRCh38
rs186428650 Chr.1: 15462952-15462952 on Build GRCh38
rs184977621 Chr.1: 15462749-15462749 on Build GRCh38

View Full Product Details