Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16044380-16044886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCNKB
Entrez Gene ID
1188
Gene Name
chloride voltage-gated channel Kb
Gene Aliases
CLCKB, ClC-K2, ClC-Kb
UniGene
Hs.352243
Gene Chromosome Location
Chr.1: 16043736-16057326 on Build GRCh38
Transcripts
NM_000085.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3802675 c.7G>C p.E3Q Chr.1: 16044499-16044499 on Build GRCh38
6629932 c.79C>T p.R27C Chr.1: 16044571-16044571 on Build GRCh38
2153859 c.85C>T p.R29C Chr.1: 16044577-16044577 on Build GRCh38
3750546 c.80G>T p.R27L Chr.1: 16044572-16044572 on Build GRCh38
5491029 c.98G>A p.R33Q Chr.1: 16044590-16044590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530322888 Chr.1: 16044713-16044713 on Build GRCh38
rs768338456 Chr.1: 16044471-16044471 on Build GRCh38
rs547016093 Chr.1: 16044826-16044826 on Build GRCh38
rs760357463 Chr.1: 16044524-16044524 on Build GRCh38
rs766622963 Chr.1: 16044564-16044565 on Build GRCh38
rs916648566 Chr.1: 16044743-16044743 on Build GRCh38
rs781680908 Chr.1: 16044470-16044470 on Build GRCh38
rs1037234777 Chr.1: 16044652-16044652 on Build GRCh38
rs370860323 Chr.1: 16044489-16044489 on Build GRCh38
rs562549002 Chr.1: 16044719-16044719 on Build GRCh38
rs34476179 Chr.1: 16044459-16044459 on Build GRCh38
rs985164800 Chr.1: 16044514-16044514 on Build GRCh38
rs889563690 Chr.1: 16044670-16044670 on Build GRCh38
rs1022537062 Chr.1: 16044521-16044521 on Build GRCh38
rs767867132 Chr.1: 16044458-16044458 on Build GRCh38
rs368792715 Chr.1: 16044637-16044637 on Build GRCh38
rs941249096 Chr.1: 16044809-16044809 on Build GRCh38
rs992758067 Chr.1: 16044737-16044737 on Build GRCh38
rs768090520 Chr.1: 16044620-16044620 on Build GRCh38
rs1006548698 Chr.1: 16044674-16044674 on Build GRCh38
rs34851419 Chr.1: 16044504-16044504 on Build GRCh38
rs2015352 Chr.1: 16044572-16044572 on Build GRCh38
rs771963940 Chr.1: 16044584-16044584 on Build GRCh38
rs115953063 Chr.1: 16044646-16044646 on Build GRCh38
rs6604907 Chr.1: 16044669-16044669 on Build GRCh38
rs969455305 Chr.1: 16044756-16044756 on Build GRCh38
rs759045664 Chr.1: 16044544-16044544 on Build GRCh38
rs13306234 Chr.1: 16044787-16044787 on Build GRCh38
rs918539289 Chr.1: 16044837-16044837 on Build GRCh38
rs1021988481 Chr.1: 16044681-16044681 on Build GRCh38
rs200835418 Chr.1: 16044580-16044580 on Build GRCh38
rs997950438 Chr.1: 16044490-16044490 on Build GRCh38
rs945394 Chr.1: 16044404-16044404 on Build GRCh38
rs374439723 Chr.1: 16044589-16044589 on Build GRCh38
rs924135805 Chr.1: 16044574-16044574 on Build GRCh38
rs757638010 Chr.1: 16044464-16044464 on Build GRCh38
rs13306233 Chr.1: 16044728-16044728 on Build GRCh38
rs747945822 Chr.1: 16044509-16044509 on Build GRCh38
rs762461903 Chr.1: 16044617-16044617 on Build GRCh38
rs531495195 Chr.1: 16044436-16044436 on Build GRCh38
rs376899737 Chr.1: 16044570-16044570 on Build GRCh38
rs953686324 Chr.1: 16044506-16044507 on Build GRCh38
rs551608685 Chr.1: 16044439-16044439 on Build GRCh38
rs768510442 Chr.1: 16044501-16044501 on Build GRCh38
rs776026475 Chr.1: 16044537-16044537 on Build GRCh38
rs772984104 Chr.1: 16044519-16044519 on Build GRCh38
rs188028488 Chr.1: 16044475-16044475 on Build GRCh38
rs992246057 Chr.1: 16044635-16044635 on Build GRCh38
rs780928130 Chr.1: 16044429-16044429 on Build GRCh38
rs767165327 Chr.1: 16044442-16044442 on Build GRCh38
rs866959922 Chr.1: 16044542-16044542 on Build GRCh38
rs536104811 Chr.1: 16044498-16044498 on Build GRCh38
rs766031319 Chr.1: 16044525-16044525 on Build GRCh38
rs765869565 Chr.1: 16044457-16044457 on Build GRCh38
rs749342666 Chr.1: 16044494-16044494 on Build GRCh38
rs774622318 Chr.1: 16044758-16044758 on Build GRCh38
rs746767761 Chr.1: 16044590-16044590 on Build GRCh38
rs770156509 Chr.1: 16044460-16044461 on Build GRCh38
rs1050863925 Chr.1: 16044659-16044659 on Build GRCh38
rs756330716 Chr.1: 16044488-16044488 on Build GRCh38
rs998462008 Chr.1: 16044654-16044654 on Build GRCh38
rs387907412 Chr.1: 16044563-16044563 on Build GRCh38
rs770519624 Chr.1: 16044593-16044593 on Build GRCh38
rs140923370 Chr.1: 16044577-16044577 on Build GRCh38
rs760303999 Chr.1: 16044450-16044450 on Build GRCh38
rs769642303 Chr.1: 16044460-16044460 on Build GRCh38
rs776279574 Chr.1: 16044595-16044595 on Build GRCh38
rs387907411 Chr.1: 16044515-16044515 on Build GRCh38
rs758980868 Chr.1: 16044603-16044603 on Build GRCh38
rs896996647 Chr.1: 16044653-16044653 on Build GRCh38
rs542452776 Chr.1: 16044718-16044718 on Build GRCh38
rs941302853 Chr.1: 16044651-16044651 on Build GRCh38
rs753962055 Chr.1: 16044594-16044595 on Build GRCh38
rs540090938 Chr.1: 16044685-16044685 on Build GRCh38
rs933822646 Chr.1: 16044853-16044853 on Build GRCh38
rs138927935 Chr.1: 16044624-16044624 on Build GRCh38
rs761974859 Chr.1: 16044764-16044764 on Build GRCh38
rs761036682 Chr.1: 16044630-16044630 on Build GRCh38
rs764512896 Chr.1: 16044552-16044552 on Build GRCh38
rs746902905 Chr.1: 16044433-16044433 on Build GRCh38
rs757836001 Chr.1: 16044564-16044564 on Build GRCh38
rs551404925 Chr.1: 16044736-16044736 on Build GRCh38
rs528950572 Chr.1: 16044657-16044657 on Build GRCh38
rs529086393 Chr.1: 16044416-16044416 on Build GRCh38
rs766528068 Chr.1: 16044639-16044639 on Build GRCh38
rs577234299 Chr.1: 16044616-16044616 on Build GRCh38
rs569178192 Chr.1: 16044426-16044426 on Build GRCh38
rs571505155 Chr.1: 16044786-16044786 on Build GRCh38
rs750016311 Chr.1: 16044449-16044449 on Build GRCh38
rs370236747 Chr.1: 16044571-16044571 on Build GRCh38
rs1049076799 Chr.1: 16044640-16044640 on Build GRCh38
rs764908276 Chr.1: 16044605-16044605 on Build GRCh38
rs775410059 Chr.1: 16044528-16044528 on Build GRCh38
rs374883047 Chr.1: 16044424-16044424 on Build GRCh38
rs544582273 Chr.1: 16044578-16044578 on Build GRCh38

View Full Product Details