Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16980380-16980868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFAP2
Entrez Gene ID
4237
Gene Name
microfibrillar associated protein 2
Gene Aliases
MAGP, MAGP-1, MAGP1
UniGene
Hs.389137
Gene Chromosome Location
Chr.1: 16974502-16981649 on Build GRCh38
Transcripts
NM_001135247.1 NM_001135248.1 NM_002403.3 NM_017459.2 XM_011541476.2

SNP Detail

SNP ID Chromosome Location
rs747294630 Chr.1: 16980650-16980650 on Build GRCh38
rs778671828 Chr.1: 16980588-16980588 on Build GRCh38
rs567378816 Chr.1: 16980646-16980646 on Build GRCh38
rs745744836 Chr.1: 16980771-16980771 on Build GRCh38
rs570879895 Chr.1: 16980603-16980603 on Build GRCh38
rs905780150 Chr.1: 16980564-16980564 on Build GRCh38
rs56265117 Chr.1: 16980428-16980428 on Build GRCh38
rs974017072 Chr.1: 16980532-16980532 on Build GRCh38
rs536445271 Chr.1: 16980720-16980720 on Build GRCh38
rs60284130 Chr.1: 16980770-16980770 on Build GRCh38
rs1001165953 Chr.1: 16980849-16980849 on Build GRCh38
rs1039904075 Chr.1: 16980803-16980803 on Build GRCh38
rs970580547 Chr.1: 16980493-16980493 on Build GRCh38
rs1045641748 Chr.1: 16980539-16980539 on Build GRCh38
rs1018920012 Chr.1: 16980647-16980647 on Build GRCh38
rs891260612 Chr.1: 16980614-16980614 on Build GRCh38
rs1025250132 Chr.1: 16980457-16980457 on Build GRCh38
rs537502186 Chr.1: 16980451-16980451 on Build GRCh38
rs988626281 Chr.1: 16980479-16980479 on Build GRCh38
rs575705250 Chr.1: 16980727-16980727 on Build GRCh38
rs988591888 Chr.1: 16980695-16980695 on Build GRCh38
rs974802859 Chr.1: 16980662-16980662 on Build GRCh38
rs533848910 Chr.1: 16980617-16980617 on Build GRCh38
rs1009769034 Chr.1: 16980670-16980670 on Build GRCh38
rs1038757974 Chr.1: 16980772-16980772 on Build GRCh38
rs564716063 Chr.1: 16980779-16980780 on Build GRCh38
rs949938204 Chr.1: 16980509-16980509 on Build GRCh38
rs1039982189 Chr.1: 16980632-16980632 on Build GRCh38
rs138941532 Chr.1: 16980735-16980738 on Build GRCh38
rs944152798 Chr.1: 16980593-16980593 on Build GRCh38
rs900066726 Chr.1: 16980835-16980835 on Build GRCh38
rs913111229 Chr.1: 16980491-16980491 on Build GRCh38
rs983278624 Chr.1: 16980775-16980775 on Build GRCh38
rs937313578 Chr.1: 16980570-16980570 on Build GRCh38
rs964339631 Chr.1: 16980657-16980657 on Build GRCh38
rs1027135159 Chr.1: 16980508-16980508 on Build GRCh38
rs898540428 Chr.1: 16980773-16980773 on Build GRCh38
rs370557362 Chr.1: 16980530-16980530 on Build GRCh38
rs771129890 Chr.1: 16980658-16980658 on Build GRCh38
rs558205225 Chr.1: 16980778-16980778 on Build GRCh38
rs1008353826 Chr.1: 16980629-16980629 on Build GRCh38
rs919925072 Chr.1: 16980579-16980579 on Build GRCh38
rs760618983 Chr.1: 16980769-16980770 on Build GRCh38
rs957109068 Chr.1: 16980689-16980689 on Build GRCh38
rs191772276 Chr.1: 16980726-16980726 on Build GRCh38
rs958719421 Chr.1: 16980514-16980515 on Build GRCh38
rs891229644 Chr.1: 16980791-16980791 on Build GRCh38
rs578046664 Chr.1: 16980793-16980793 on Build GRCh38
rs898562260 Chr.1: 16980586-16980586 on Build GRCh38
rs55779591 Chr.1: 16980453-16980453 on Build GRCh38
rs975548743 Chr.1: 16980777-16980777 on Build GRCh38
rs929987125 Chr.1: 16980774-16980774 on Build GRCh38
rs553632753 Chr.1: 16980618-16980618 on Build GRCh38
rs879506743 Chr.1: 16980590-16980590 on Build GRCh38
rs1024694093 Chr.1: 16980684-16980684 on Build GRCh38
rs529617685 Chr.1: 16980770-16980771 on Build GRCh38
rs1038384950 Chr.1: 16980571-16980571 on Build GRCh38
rs1032705712 Chr.1: 16980663-16980663 on Build GRCh38
rs1006947761 Chr.1: 16980585-16980585 on Build GRCh38
rs906379809 Chr.1: 16980705-16980705 on Build GRCh38
rs1033520843 Chr.1: 16980514-16980514 on Build GRCh38
rs1032108942 Chr.1: 16980427-16980427 on Build GRCh38
rs937301734 Chr.1: 16980771-16980771 on Build GRCh38
rs900061154 Chr.1: 16980644-16980644 on Build GRCh38
rs530088830 Chr.1: 16980728-16980728 on Build GRCh38
rs757276787 Chr.1: 16980769-16980769 on Build GRCh38

View Full Product Details