Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17270875-17271365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PADI3
Entrez Gene ID
51702
Gene Name
peptidyl arginine deiminase 3
Gene Aliases
PAD3, PDI3
UniGene
Hs.149195
Gene Chromosome Location
Chr.1: 17249079-17284233 on Build GRCh38
Transcripts
NM_016233.2 XM_011541571.2 XM_011541572.2 XM_017001463.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6373045 c.841G>A p.A281T Chr.1: 17270888-17270888 on Build GRCh38
7195701 c.995G>A p.C332Y Chr.1: 17271126-17271126 on Build GRCh38
390376 c.980C>A p.A327D Chr.1: 17271111-17271111 on Build GRCh38
6271185 c.1028G>T p.R343L Chr.1: 17271159-17271159 on Build GRCh38
2093690 c.1023G>T p.E341D Chr.1: 17271154-17271154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373280246 Chr.1: 17271041-17271041 on Build GRCh38
rs552971232 Chr.1: 17271092-17271092 on Build GRCh38
rs764860340 Chr.1: 17271159-17271159 on Build GRCh38
rs143396189 Chr.1: 17271151-17271151 on Build GRCh38
rs568276808 Chr.1: 17270922-17270922 on Build GRCh38
rs144080386 Chr.1: 17270928-17270928 on Build GRCh38
rs1031518225 Chr.1: 17271233-17271233 on Build GRCh38
rs779715839 Chr.1: 17271164-17271164 on Build GRCh38
rs200041897 Chr.1: 17271214-17271214 on Build GRCh38
rs745786433 Chr.1: 17270944-17270944 on Build GRCh38
rs768543947 Chr.1: 17271107-17271107 on Build GRCh38
rs752815659 Chr.1: 17271061-17271061 on Build GRCh38
rs147183978 Chr.1: 17271163-17271163 on Build GRCh38
rs151130447 Chr.1: 17270966-17270966 on Build GRCh38
rs1039220581 Chr.1: 17270977-17270977 on Build GRCh38
rs779877401 Chr.1: 17271103-17271103 on Build GRCh38
rs375585925 Chr.1: 17270993-17270993 on Build GRCh38
rs761986987 Chr.1: 17271066-17271066 on Build GRCh38
rs750716007 Chr.1: 17270990-17270990 on Build GRCh38
rs764523694 Chr.1: 17270982-17270982 on Build GRCh38
rs970615797 Chr.1: 17271093-17271093 on Build GRCh38
rs541935186 Chr.1: 17271142-17271142 on Build GRCh38
rs35340855 Chr.1: 17271084-17271084 on Build GRCh38
rs752484702 Chr.1: 17271251-17271251 on Build GRCh38
rs372305068 Chr.1: 17271213-17271213 on Build GRCh38
rs4595409 Chr.1: 17271226-17271226 on Build GRCh38
rs771856526 Chr.1: 17270947-17270947 on Build GRCh38
rs747010919 Chr.1: 17271056-17271056 on Build GRCh38
rs890699798 Chr.1: 17270967-17270967 on Build GRCh38
rs562060102 Chr.1: 17271275-17271275 on Build GRCh38
rs774896280 Chr.1: 17271138-17271138 on Build GRCh38
rs969031681 Chr.1: 17270957-17270957 on Build GRCh38
rs780639796 Chr.1: 17270920-17270920 on Build GRCh38
rs760682098 Chr.1: 17270962-17270962 on Build GRCh38
rs772634367 Chr.1: 17271221-17271221 on Build GRCh38
rs778365044 Chr.1: 17271096-17271096 on Build GRCh38
rs557261332 Chr.1: 17271062-17271062 on Build GRCh38
rs748115863 Chr.1: 17271116-17271116 on Build GRCh38
rs766932761 Chr.1: 17270996-17270996 on Build GRCh38
rs1009632930 Chr.1: 17271034-17271034 on Build GRCh38
rs141411323 Chr.1: 17271158-17271158 on Build GRCh38
rs753254314 Chr.1: 17271157-17271157 on Build GRCh38
rs368643899 Chr.1: 17271190-17271190 on Build GRCh38
rs551221702 Chr.1: 17270943-17270943 on Build GRCh38
rs775422448 Chr.1: 17270950-17270950 on Build GRCh38
rs140596018 Chr.1: 17270939-17270939 on Build GRCh38
rs973904055 Chr.1: 17270929-17270929 on Build GRCh38
rs978614612 Chr.1: 17270971-17270971 on Build GRCh38
rs773494983 Chr.1: 17271071-17271071 on Build GRCh38
rs748925952 Chr.1: 17271023-17271023 on Build GRCh38
rs758451448 Chr.1: 17271073-17271074 on Build GRCh38
rs781133131 Chr.1: 17271114-17271114 on Build GRCh38
rs906215717 Chr.1: 17271303-17271303 on Build GRCh38
rs761993755 Chr.1: 17270981-17270981 on Build GRCh38
rs1037921293 Chr.1: 17271121-17271121 on Build GRCh38
rs775402350 Chr.1: 17271055-17271055 on Build GRCh38
rs776734446 Chr.1: 17271063-17271063 on Build GRCh38
rs750156839 Chr.1: 17271097-17271097 on Build GRCh38
rs773298347 Chr.1: 17271122-17271122 on Build GRCh38
rs777498129 Chr.1: 17270924-17270924 on Build GRCh38
rs140711335 Chr.1: 17271298-17271298 on Build GRCh38
rs747557126 Chr.1: 17271287-17271287 on Build GRCh38
rs368973782 Chr.1: 17271027-17271027 on Build GRCh38
rs572833699 Chr.1: 17271126-17271126 on Build GRCh38
rs775848586 Chr.1: 17271222-17271222 on Build GRCh38
rs748179947 Chr.1: 17271181-17271181 on Build GRCh38
rs765324453 Chr.1: 17271069-17271069 on Build GRCh38
rs963352210 Chr.1: 17271326-17271326 on Build GRCh38
rs777567027 Chr.1: 17271299-17271299 on Build GRCh38
rs140529865 Chr.1: 17271167-17271167 on Build GRCh38
rs758872744 Chr.1: 17270909-17270909 on Build GRCh38
rs376973180 Chr.1: 17271026-17271026 on Build GRCh38
rs2293916 Chr.1: 17271160-17271160 on Build GRCh38
rs1002037027 Chr.1: 17271336-17271336 on Build GRCh38
rs550839371 Chr.1: 17271319-17271319 on Build GRCh38
rs763156968 Chr.1: 17271075-17271075 on Build GRCh38
rs528557680 Chr.1: 17271252-17271252 on Build GRCh38
rs769745732 Chr.1: 17271185-17271185 on Build GRCh38
rs763664949 Chr.1: 17271086-17271086 on Build GRCh38
rs771399492 Chr.1: 17271132-17271132 on Build GRCh38
rs371836063 Chr.1: 17270969-17270969 on Build GRCh38
rs752098569 Chr.1: 17271083-17271083 on Build GRCh38
rs756806020 Chr.1: 17271094-17271094 on Build GRCh38
rs374292518 Chr.1: 17271005-17271005 on Build GRCh38
rs149889085 Chr.1: 17270974-17270974 on Build GRCh38
rs3900818 Chr.1: 17271250-17271250 on Build GRCh38
rs765582813 Chr.1: 17270897-17270897 on Build GRCh38
rs765334620 Chr.1: 17271016-17271022 on Build GRCh38
rs369400143 Chr.1: 17271012-17271012 on Build GRCh38
rs372942922 Chr.1: 17271015-17271015 on Build GRCh38
rs1014022390 Chr.1: 17271247-17271247 on Build GRCh38
rs980688561 Chr.1: 17271099-17271099 on Build GRCh38
rs755678365 Chr.1: 17271011-17271011 on Build GRCh38
rs747611201 Chr.1: 17270921-17270921 on Build GRCh38
rs189676862 Chr.1: 17271246-17271246 on Build GRCh38
rs769919248 Chr.1: 17271117-17271117 on Build GRCh38
rs377689956 Chr.1: 17271104-17271104 on Build GRCh38
rs147256594 Chr.1: 17271081-17271081 on Build GRCh38
rs548637617 Chr.1: 17271258-17271258 on Build GRCh38
rs780921729 Chr.1: 17271178-17271178 on Build GRCh38
rs139426141 Chr.1: 17270903-17270903 on Build GRCh38

View Full Product Details