Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17381746-17382242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PADI6
Entrez Gene ID
353238
Gene Name
peptidyl arginine deiminase 6
Gene Aliases
hPADVI
UniGene
Hs.531598
Gene Chromosome Location
Chr.1: 17372196-17401699 on Build GRCh38
Transcripts
NM_207421.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6997592 c.659C>T p.A220V Chr.1: 17382072-17382072 on Build GRCh38
4226465 c.639C>T p.S213S Chr.1: 17382052-17382052 on Build GRCh38
676836 c.593G>T p.G198V Chr.1: 17382006-17382006 on Build GRCh38
3746675 c.620G>A p.R207Q Chr.1: 17382033-17382033 on Build GRCh38
3400022 c.651G>A p.S217S Chr.1: 17382064-17382064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748698648 Chr.1: 17381935-17381935 on Build GRCh38
rs781608134 Chr.1: 17382083-17382083 on Build GRCh38
rs766424489 Chr.1: 17382135-17382135 on Build GRCh38
rs377202883 Chr.1: 17381904-17381904 on Build GRCh38
rs761739312 Chr.1: 17381974-17381974 on Build GRCh38
rs769663751 Chr.1: 17382029-17382030 on Build GRCh38
rs188669403 Chr.1: 17381884-17381884 on Build GRCh38
rs552700086 Chr.1: 17382151-17382151 on Build GRCh38
rs373085990 Chr.1: 17382129-17382129 on Build GRCh38
rs773094687 Chr.1: 17382134-17382134 on Build GRCh38
rs561583700 Chr.1: 17381885-17381885 on Build GRCh38
rs776811887 Chr.1: 17382133-17382133 on Build GRCh38
rs996930220 Chr.1: 17382212-17382212 on Build GRCh38
rs570384831 Chr.1: 17382143-17382143 on Build GRCh38
rs79493782 Chr.1: 17381851-17381851 on Build GRCh38
rs1037829412 Chr.1: 17382146-17382146 on Build GRCh38
rs745905938 Chr.1: 17382006-17382007 on Build GRCh38
rs114337556 Chr.1: 17381858-17381858 on Build GRCh38
rs770500103 Chr.1: 17381947-17381947 on Build GRCh38
rs779431094 Chr.1: 17382111-17382111 on Build GRCh38
rs965964200 Chr.1: 17382158-17382160 on Build GRCh38
rs746610372 Chr.1: 17382118-17382118 on Build GRCh38
rs766402946 Chr.1: 17382072-17382072 on Build GRCh38
rs376937636 Chr.1: 17382064-17382064 on Build GRCh38
rs199828844 Chr.1: 17381973-17381973 on Build GRCh38
rs955138124 Chr.1: 17381963-17381963 on Build GRCh38
rs774012598 Chr.1: 17381950-17381950 on Build GRCh38
rs1052338417 Chr.1: 17381838-17381838 on Build GRCh38
rs55754476 Chr.1: 17382132-17382132 on Build GRCh38
rs373091309 Chr.1: 17382060-17382060 on Build GRCh38
rs979044675 Chr.1: 17381781-17381781 on Build GRCh38
rs750559850 Chr.1: 17381984-17381984 on Build GRCh38
rs775156958 Chr.1: 17382046-17382046 on Build GRCh38
rs150265019 Chr.1: 17382193-17382193 on Build GRCh38
rs755169325 Chr.1: 17382077-17382077 on Build GRCh38
rs763141031 Chr.1: 17382069-17382069 on Build GRCh38
rs753042161 Chr.1: 17381800-17381800 on Build GRCh38
rs758613912 Chr.1: 17381986-17381986 on Build GRCh38
rs747066917 Chr.1: 17382073-17382073 on Build GRCh38
rs779806717 Chr.1: 17381770-17381770 on Build GRCh38
rs372730011 Chr.1: 17382032-17382032 on Build GRCh38
rs777149297 Chr.1: 17381933-17381933 on Build GRCh38
rs543766470 Chr.1: 17381900-17381900 on Build GRCh38
rs756661415 Chr.1: 17382102-17382102 on Build GRCh38
rs1037691676 Chr.1: 17381887-17381887 on Build GRCh38
rs776420072 Chr.1: 17382061-17382061 on Build GRCh38
rs745413780 Chr.1: 17382039-17382039 on Build GRCh38
rs781444420 Chr.1: 17382000-17382001 on Build GRCh38
rs775120426 Chr.1: 17381964-17381964 on Build GRCh38
rs750469901 Chr.1: 17382067-17382067 on Build GRCh38
rs951684530 Chr.1: 17381910-17381911 on Build GRCh38
rs956332450 Chr.1: 17382048-17382048 on Build GRCh38
rs369994332 Chr.1: 17382035-17382035 on Build GRCh38
rs552744225 Chr.1: 17381913-17381913 on Build GRCh38
rs572609515 Chr.1: 17382171-17382171 on Build GRCh38
rs765239729 Chr.1: 17381977-17381977 on Build GRCh38
rs563442299 Chr.1: 17381829-17381829 on Build GRCh38
rs1030544558 Chr.1: 17381919-17381919 on Build GRCh38
rs777715785 Chr.1: 17382059-17382059 on Build GRCh38
rs112057062 Chr.1: 17381851-17381852 on Build GRCh38
rs1020178936 Chr.1: 17381939-17381939 on Build GRCh38
rs753183881 Chr.1: 17382095-17382095 on Build GRCh38
rs747472866 Chr.1: 17381927-17381927 on Build GRCh38
rs201174737 Chr.1: 17381965-17381965 on Build GRCh38
rs945235936 Chr.1: 17381856-17381856 on Build GRCh38
rs778566039 Chr.1: 17382038-17382038 on Build GRCh38
rs1027243824 Chr.1: 17381906-17381906 on Build GRCh38
rs747976426 Chr.1: 17382131-17382131 on Build GRCh38
rs201403576 Chr.1: 17381852-17381853 on Build GRCh38
rs781415187 Chr.1: 17382015-17382015 on Build GRCh38
rs370127465 Chr.1: 17382218-17382218 on Build GRCh38
rs759253886 Chr.1: 17381953-17381953 on Build GRCh38
rs768246834 Chr.1: 17382128-17382128 on Build GRCh38
rs61766772 Chr.1: 17382033-17382033 on Build GRCh38
rs532616882 Chr.1: 17381905-17381905 on Build GRCh38
rs761724874 Chr.1: 17382063-17382063 on Build GRCh38
rs113858406 Chr.1: 17381895-17381895 on Build GRCh38
rs80026107 Chr.1: 17382145-17382145 on Build GRCh38
rs369167478 Chr.1: 17382011-17382011 on Build GRCh38
rs965556589 Chr.1: 17381944-17381944 on Build GRCh38
rs905251552 Chr.1: 17381866-17381866 on Build GRCh38
rs191095230 Chr.1: 17381929-17381929 on Build GRCh38
rs760273114 Chr.1: 17382055-17382055 on Build GRCh38
rs780361897 Chr.1: 17381992-17381992 on Build GRCh38
rs924975928 Chr.1: 17381785-17381785 on Build GRCh38
rs565879071 Chr.1: 17381914-17381914 on Build GRCh38
rs920952860 Chr.1: 17381841-17381841 on Build GRCh38
rs917827550 Chr.1: 17381845-17381845 on Build GRCh38
rs749837509 Chr.1: 17382106-17382106 on Build GRCh38
rs751753335 Chr.1: 17382007-17382007 on Build GRCh38
rs771604783 Chr.1: 17382040-17382040 on Build GRCh38
rs112817962 Chr.1: 17381769-17381769 on Build GRCh38
rs769011107 Chr.1: 17381932-17381932 on Build GRCh38
rs7556072 Chr.1: 17381854-17381854 on Build GRCh38
rs771828240 Chr.1: 17381955-17381955 on Build GRCh38

View Full Product Details