Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17392071-17392564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PADI6
Entrez Gene ID
353238
Gene Name
peptidyl arginine deiminase 6
Gene Aliases
hPADVI
UniGene
Hs.531598
Gene Chromosome Location
Chr.1: 17372196-17401699 on Build GRCh38
Transcripts
NM_207421.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1158176 c.1044C>T p.D348D Chr.1: 17392198-17392198 on Build GRCh38
3771529 c.1026+1G>C p.? Chr.1: 17392178-17392178 on Build GRCh38
5516132 c.998C>T p.T333M Chr.1: 17392149-17392149 on Build GRCh38
5713025 c.1026delG p.A343fs*32 Chr.1: 17392177-17392177 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200649188 Chr.1: 17392178-17392178 on Build GRCh38
rs1009575047 Chr.1: 17392522-17392522 on Build GRCh38
rs184514891 Chr.1: 17392430-17392430 on Build GRCh38
rs188550053 Chr.1: 17392305-17392305 on Build GRCh38
rs764055065 Chr.1: 17392248-17392248 on Build GRCh38
rs189613221 Chr.1: 17392444-17392444 on Build GRCh38
rs907403006 Chr.1: 17392484-17392484 on Build GRCh38
rs369052562 Chr.1: 17392207-17392207 on Build GRCh38
rs74058825 Chr.1: 17392260-17392260 on Build GRCh38
rs578077400 Chr.1: 17392400-17392400 on Build GRCh38
rs754291664 Chr.1: 17392277-17392277 on Build GRCh38
rs531826273 Chr.1: 17392476-17392476 on Build GRCh38
rs113887714 Chr.1: 17392527-17392527 on Build GRCh38
rs777388185 Chr.1: 17392185-17392185 on Build GRCh38
rs375267418 Chr.1: 17392149-17392149 on Build GRCh38
rs574017970 Chr.1: 17392427-17392427 on Build GRCh38
rs929014466 Chr.1: 17392436-17392436 on Build GRCh38
rs562617192 Chr.1: 17392208-17392208 on Build GRCh38
rs976219306 Chr.1: 17392418-17392418 on Build GRCh38
rs540831324 Chr.1: 17392402-17392402 on Build GRCh38
rs755386206 Chr.1: 17392299-17392299 on Build GRCh38
rs760604887 Chr.1: 17392245-17392245 on Build GRCh38
rs74058826 Chr.1: 17392358-17392358 on Build GRCh38
rs184870201 Chr.1: 17392139-17392139 on Build GRCh38
rs536225740 Chr.1: 17392323-17392323 on Build GRCh38
rs369227850 Chr.1: 17392150-17392150 on Build GRCh38
rs758747780 Chr.1: 17392174-17392174 on Build GRCh38
rs1011843731 Chr.1: 17392345-17392345 on Build GRCh38
rs1034724702 Chr.1: 17392328-17392328 on Build GRCh38
rs745342727 Chr.1: 17392104-17392104 on Build GRCh38
rs536529522 Chr.1: 17392495-17392496 on Build GRCh38
rs764568882 Chr.1: 17392174-17392174 on Build GRCh38
rs74503949 Chr.1: 17392375-17392375 on Build GRCh38
rs750629079 Chr.1: 17392172-17392172 on Build GRCh38
rs868721033 Chr.1: 17392265-17392265 on Build GRCh38
rs2800691 Chr.1: 17392509-17392509 on Build GRCh38
rs112535805 Chr.1: 17392546-17392546 on Build GRCh38
rs1017424322 Chr.1: 17392128-17392128 on Build GRCh38
rs774017113 Chr.1: 17392220-17392220 on Build GRCh38
rs1027236710 Chr.1: 17392531-17392531 on Build GRCh38
rs12737852 Chr.1: 17392254-17392254 on Build GRCh38
rs1057517683 Chr.1: 17392121-17392121 on Build GRCh38
rs907958783 Chr.1: 17392200-17392200 on Build GRCh38
rs556223000 Chr.1: 17392327-17392327 on Build GRCh38
rs533668017 Chr.1: 17392104-17392104 on Build GRCh38
rs543626039 Chr.1: 17392354-17392354 on Build GRCh38
rs375305818 Chr.1: 17392201-17392201 on Build GRCh38
rs776666748 Chr.1: 17392255-17392255 on Build GRCh38
rs1042359960 Chr.1: 17392470-17392470 on Build GRCh38
rs757584809 Chr.1: 17392255-17392255 on Build GRCh38
rs533992573 Chr.1: 17392270-17392270 on Build GRCh38
rs12031545 Chr.1: 17392515-17392515 on Build GRCh38
rs951929892 Chr.1: 17392365-17392365 on Build GRCh38
rs527459136 Chr.1: 17392488-17392488 on Build GRCh38
rs931990758 Chr.1: 17392421-17392421 on Build GRCh38
rs964486097 Chr.1: 17392130-17392130 on Build GRCh38
rs946282704 Chr.1: 17392452-17392452 on Build GRCh38
rs1041856168 Chr.1: 17392303-17392303 on Build GRCh38
rs769342993 Chr.1: 17392183-17392183 on Build GRCh38
rs911523021 Chr.1: 17392472-17392472 on Build GRCh38
rs906540091 Chr.1: 17392528-17392529 on Build GRCh38
rs200954407 Chr.1: 17392179-17392179 on Build GRCh38
rs914837170 Chr.1: 17392426-17392426 on Build GRCh38
rs765329691 Chr.1: 17392261-17392261 on Build GRCh38
rs372214311 Chr.1: 17392168-17392168 on Build GRCh38
rs980949660 Chr.1: 17392404-17392404 on Build GRCh38
rs181943218 Chr.1: 17392521-17392521 on Build GRCh38
rs752616539 Chr.1: 17392144-17392144 on Build GRCh38
rs941777599 Chr.1: 17392206-17392206 on Build GRCh38
rs74419307 Chr.1: 17392487-17392487 on Build GRCh38
rs970931516 Chr.1: 17392182-17392182 on Build GRCh38
rs192264745 Chr.1: 17392423-17392423 on Build GRCh38
rs748682379 Chr.1: 17392420-17392420 on Build GRCh38
rs398089152 Chr.1: 17392178-17392178 on Build GRCh38
rs551535333 Chr.1: 17392481-17392481 on Build GRCh38
rs918951598 Chr.1: 17392232-17392232 on Build GRCh38
rs759328284 Chr.1: 17392129-17392129 on Build GRCh38
rs773082407 Chr.1: 17392134-17392134 on Build GRCh38
rs1002444343 Chr.1: 17392534-17392534 on Build GRCh38
rs774266573 Chr.1: 17392117-17392117 on Build GRCh38
rs753981238 Chr.1: 17392163-17392163 on Build GRCh38
rs747491479 Chr.1: 17392177-17392177 on Build GRCh38
rs572104363 Chr.1: 17392190-17392190 on Build GRCh38
rs371891091 Chr.1: 17392142-17392142 on Build GRCh38
rs775210631 Chr.1: 17392236-17392236 on Build GRCh38
rs531744275 Chr.1: 17392222-17392222 on Build GRCh38
rs576071991 Chr.1: 17392344-17392344 on Build GRCh38
rs111789712 Chr.1: 17392383-17392383 on Build GRCh38
rs779152121 Chr.1: 17392171-17392171 on Build GRCh38
rs567581297 Chr.1: 17392306-17392306 on Build GRCh38
rs1038945366 Chr.1: 17392477-17392477 on Build GRCh38

View Full Product Details