Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19814659-19815168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF186
Entrez Gene ID
54546
Gene Name
ring finger protein 186
Gene Aliases
-
UniGene
Hs.124835
Gene Chromosome Location
Chr.1: 19814029-19815278 on Build GRCh38
Transcripts
NM_019062.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
678177 c.261G>C p.K87N Chr.1: 19814841-19814841 on Build GRCh38
3864101 c.76C>T p.P26S Chr.1: 19815026-19815026 on Build GRCh38
678176 c.103G>T p.E35* Chr.1: 19814999-19814999 on Build GRCh38
3864100 c.309G>A p.E103E Chr.1: 19814793-19814793 on Build GRCh38
3976844 c.404G>T p.S135I Chr.1: 19814698-19814698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779912959 Chr.1: 19814832-19814832 on Build GRCh38
rs41264113 Chr.1: 19814912-19814912 on Build GRCh38
rs200902005 Chr.1: 19814972-19814972 on Build GRCh38
rs767057932 Chr.1: 19815110-19815110 on Build GRCh38
rs780765955 Chr.1: 19815023-19815023 on Build GRCh38
rs573777317 Chr.1: 19814799-19814799 on Build GRCh38
rs774433130 Chr.1: 19814949-19814949 on Build GRCh38
rs1054204777 Chr.1: 19814839-19814839 on Build GRCh38
rs116045850 Chr.1: 19814804-19814804 on Build GRCh38
rs142776324 Chr.1: 19814732-19814732 on Build GRCh38
rs368552116 Chr.1: 19814803-19814803 on Build GRCh38
rs964224247 Chr.1: 19815009-19815009 on Build GRCh38
rs775762632 Chr.1: 19814954-19814954 on Build GRCh38
rs199500273 Chr.1: 19814831-19814831 on Build GRCh38
rs878995541 Chr.1: 19815013-19815013 on Build GRCh38
rs767724635 Chr.1: 19814872-19814872 on Build GRCh38
rs758050679 Chr.1: 19814895-19814895 on Build GRCh38
rs370363967 Chr.1: 19814768-19814768 on Build GRCh38
rs201325675 Chr.1: 19814962-19814962 on Build GRCh38
rs751568791 Chr.1: 19814856-19814856 on Build GRCh38
rs951574937 Chr.1: 19814815-19814815 on Build GRCh38
rs748876956 Chr.1: 19814698-19814698 on Build GRCh38
rs150996095 Chr.1: 19815031-19815031 on Build GRCh38
rs146325708 Chr.1: 19814971-19814971 on Build GRCh38
rs374960430 Chr.1: 19814740-19814740 on Build GRCh38
rs769659519 Chr.1: 19814845-19814845 on Build GRCh38
rs193217692 Chr.1: 19814884-19814884 on Build GRCh38
rs149641709 Chr.1: 19814925-19814925 on Build GRCh38
rs773110875 Chr.1: 19814846-19814846 on Build GRCh38
rs770882872 Chr.1: 19815043-19815043 on Build GRCh38
rs200709475 Chr.1: 19814913-19814913 on Build GRCh38
rs769778620 Chr.1: 19814951-19814951 on Build GRCh38
rs750533210 Chr.1: 19814801-19814801 on Build GRCh38
rs774317437 Chr.1: 19815048-19815048 on Build GRCh38
rs771841790 Chr.1: 19815132-19815132 on Build GRCh38
rs201411238 Chr.1: 19814835-19814835 on Build GRCh38
rs751960462 Chr.1: 19814758-19814758 on Build GRCh38
rs1054659114 Chr.1: 19814742-19814742 on Build GRCh38
rs778777865 Chr.1: 19815126-19815126 on Build GRCh38
rs545485178 Chr.1: 19814763-19814763 on Build GRCh38
rs770794921 Chr.1: 19814935-19814935 on Build GRCh38
rs368683659 Chr.1: 19814892-19814892 on Build GRCh38
rs761620972 Chr.1: 19814841-19814841 on Build GRCh38
rs779935601 Chr.1: 19814920-19814920 on Build GRCh38
rs371529547 Chr.1: 19814960-19814960 on Build GRCh38
rs543596067 Chr.1: 19814784-19814784 on Build GRCh38
rs1034315093 Chr.1: 19814919-19814919 on Build GRCh38
rs185666442 Chr.1: 19815115-19815115 on Build GRCh38
rs755311819 Chr.1: 19814760-19814760 on Build GRCh38
rs745415535 Chr.1: 19814827-19814827 on Build GRCh38
rs751063588 Chr.1: 19814976-19814976 on Build GRCh38
rs367913080 Chr.1: 19814764-19814764 on Build GRCh38
rs891580247 Chr.1: 19814894-19814894 on Build GRCh38
rs376655245 Chr.1: 19815044-19815044 on Build GRCh38
rs780209634 Chr.1: 19814754-19814754 on Build GRCh38
rs776699070 Chr.1: 19814791-19814791 on Build GRCh38
rs912800988 Chr.1: 19814822-19814822 on Build GRCh38
rs939536327 Chr.1: 19814875-19814875 on Build GRCh38
rs150494893 Chr.1: 19815117-19815117 on Build GRCh38
rs377351426 Chr.1: 19814736-19814736 on Build GRCh38
rs774117323 Chr.1: 19814708-19814708 on Build GRCh38
rs201407976 Chr.1: 19815112-19815112 on Build GRCh38
rs112876882 Chr.1: 19814687-19814687 on Build GRCh38
rs34693363 Chr.1: 19814766-19814766 on Build GRCh38
rs375096308 Chr.1: 19814928-19814928 on Build GRCh38
rs147586351 Chr.1: 19815063-19815063 on Build GRCh38
rs555165614 Chr.1: 19815042-19815042 on Build GRCh38
rs201262383 Chr.1: 19814802-19814802 on Build GRCh38
rs777554238 Chr.1: 19815036-19815036 on Build GRCh38
rs765894281 Chr.1: 19815083-19815083 on Build GRCh38
rs748212664 Chr.1: 19814859-19814860 on Build GRCh38
rs1045568639 Chr.1: 19815090-19815090 on Build GRCh38
rs764552514 Chr.1: 19814963-19814963 on Build GRCh38
rs750437587 Chr.1: 19814751-19814751 on Build GRCh38
rs746469604 Chr.1: 19814908-19814908 on Build GRCh38
rs760509691 Chr.1: 19814726-19814726 on Build GRCh38
rs763077781 Chr.1: 19814853-19814853 on Build GRCh38
rs75270523 Chr.1: 19815118-19815118 on Build GRCh38
rs755783405 Chr.1: 19815113-19815113 on Build GRCh38
rs139260117 Chr.1: 19814744-19814744 on Build GRCh38
rs759262417 Chr.1: 19814718-19814718 on Build GRCh38
rs780196592 Chr.1: 19815133-19815133 on Build GRCh38
rs759033786 Chr.1: 19815108-19815108 on Build GRCh38
rs775644469 Chr.1: 19815062-19815062 on Build GRCh38
rs771689670 Chr.1: 19814829-19814829 on Build GRCh38
rs928065236 Chr.1: 19815068-19815068 on Build GRCh38
rs762229219 Chr.1: 19815072-19815072 on Build GRCh38
rs773198909 Chr.1: 19814931-19814931 on Build GRCh38
rs773419653 Chr.1: 19815037-19815038 on Build GRCh38
rs777158876 Chr.1: 19815067-19815067 on Build GRCh38
rs781750387 Chr.1: 19814761-19814761 on Build GRCh38
rs749224399 Chr.1: 19814932-19814932 on Build GRCh38
rs1541184 Chr.1: 19814790-19814790 on Build GRCh38
rs767676001 Chr.1: 19814951-19814951 on Build GRCh38
rs770331438 Chr.1: 19814703-19814703 on Build GRCh38
rs760597725 Chr.1: 19814788-19814788 on Build GRCh38
rs750928036 Chr.1: 19815085-19815085 on Build GRCh38
rs201766425 Chr.1: 19814765-19814765 on Build GRCh38
rs141513805 Chr.1: 19814886-19814886 on Build GRCh38
rs148594830 Chr.1: 19814752-19814752 on Build GRCh38
rs768529602 Chr.1: 19814692-19814692 on Build GRCh38
rs146963497 Chr.1: 19814871-19814871 on Build GRCh38
rs745815435 Chr.1: 19815131-19815131 on Build GRCh38
rs535476759 Chr.1: 19814722-19814722 on Build GRCh38
rs754488757 Chr.1: 19815004-19815004 on Build GRCh38
rs767216940 Chr.1: 19814720-19814720 on Build GRCh38
rs142010502 Chr.1: 19814969-19814969 on Build GRCh38
rs765163524 Chr.1: 19814800-19814800 on Build GRCh38
rs755435727 Chr.1: 19814819-19814819 on Build GRCh38
rs372014586 Chr.1: 19814897-19814897 on Build GRCh38
rs368042107 Chr.1: 19814950-19814950 on Build GRCh38
rs138663359 Chr.1: 19814826-19814826 on Build GRCh38
rs753044028 Chr.1: 19814823-19814823 on Build GRCh38
rs1541185 Chr.1: 19815035-19815035 on Build GRCh38
rs768289183 Chr.1: 19814834-19814834 on Build GRCh38
rs568188617 Chr.1: 19814885-19814885 on Build GRCh38
rs181898691 Chr.1: 19815060-19815060 on Build GRCh38
rs141102528 Chr.1: 19815059-19815059 on Build GRCh38
rs145074386 Chr.1: 19814825-19814825 on Build GRCh38

View Full Product Details