Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20827535-20828024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4G3
Entrez Gene ID
8672
Gene Name
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases
eIF-4G 3, eIF4G 3, eIF4GII
UniGene
Hs.467084
Gene Chromosome Location
Chr.1: 20806292-21177320 on Build GRCh38
Transcripts
NM_001198801.1 NM_001198802.1 NM_003760.4 XM_011542356.2 XM_011542361.2 XM_011542370.1 XM_011542371.1 XM_011542374.1 XM_011542375.2 XM_011542381.2 XM_017002672.1 XM_017002673.1 XM_017002674.1 XM_017002675.1 XM_017002676.1 XM_017002677.1 XM_017002678.1 XM_017002679.1 XM_017002680.1 XM_017002681.1 XM_017002682.1 XM_017002683.1 XM_017002684.1 XM_017002685.1 XM_017002686.1 XM_017002687.1 XM_017002688.1 XM_017002689.1 XM_017002690.1 XM_017002691.1 XM_017002692.1 XM_017002693.1 XM_017002694.1 XM_017002695.1 XM_017002696.1 XM_017002697.1 XM_017002698.1 XM_017002699.1 XM_017002700.1 XM_017002701.1 XM_017002702.1 XM_017002703.1 XM_017002704.1 XM_017002705.1 XM_017002706.1 XM_017002707.1 XM_017002708.1 XM_017002709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7101915 c.4108T>C p.C1370R Chr.1: 20827628-20827628 on Build GRCh38
903473 c.4042T>C p.F1348L Chr.1: 20827694-20827694 on Build GRCh38
3789449 c.4070G>C p.R1357T Chr.1: 20827666-20827666 on Build GRCh38
3943437 c.4077A>C p.I1359I Chr.1: 20827641-20827641 on Build GRCh38
7101916 c.4090T>C p.C1364R Chr.1: 20827628-20827628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898397197 Chr.1: 20827972-20827972 on Build GRCh38
rs760958641 Chr.1: 20827652-20827652 on Build GRCh38
rs780034907 Chr.1: 20827900-20827900 on Build GRCh38
rs535336814 Chr.1: 20827877-20827877 on Build GRCh38
rs554558342 Chr.1: 20827879-20827880 on Build GRCh38
rs760073491 Chr.1: 20827610-20827610 on Build GRCh38
rs777205424 Chr.1: 20827606-20827607 on Build GRCh38
rs367795928 Chr.1: 20827761-20827761 on Build GRCh38
rs767974703 Chr.1: 20827622-20827622 on Build GRCh38
rs968900083 Chr.1: 20827624-20827624 on Build GRCh38
rs773581982 Chr.1: 20827567-20827567 on Build GRCh38
rs1039983934 Chr.1: 20827724-20827724 on Build GRCh38
rs746703453 Chr.1: 20827670-20827670 on Build GRCh38
rs78515631 Chr.1: 20827861-20827861 on Build GRCh38
rs892576095 Chr.1: 20827808-20827808 on Build GRCh38
rs747919877 Chr.1: 20827736-20827736 on Build GRCh38
rs368311897 Chr.1: 20827786-20827786 on Build GRCh38
rs753035646 Chr.1: 20827627-20827627 on Build GRCh38
rs1046436254 Chr.1: 20827874-20827874 on Build GRCh38
rs397967710 Chr.1: 20827892-20827893 on Build GRCh38
rs34303992 Chr.1: 20827887-20827888 on Build GRCh38
rs907052351 Chr.1: 20827909-20827909 on Build GRCh38
rs779521367 Chr.1: 20827707-20827707 on Build GRCh38
rs191837120 Chr.1: 20827912-20827912 on Build GRCh38
rs754941442 Chr.1: 20827712-20827712 on Build GRCh38
rs376746220 Chr.1: 20827709-20827709 on Build GRCh38
rs1034308302 Chr.1: 20827918-20827918 on Build GRCh38
rs888375576 Chr.1: 20827852-20827852 on Build GRCh38
rs560272931 Chr.1: 20827944-20827944 on Build GRCh38
rs57117079 Chr.1: 20827888-20827889 on Build GRCh38
rs1048288939 Chr.1: 20827847-20827847 on Build GRCh38
rs372159887 Chr.1: 20827571-20827571 on Build GRCh38
rs764817596 Chr.1: 20827664-20827664 on Build GRCh38
rs150435979 Chr.1: 20827562-20827562 on Build GRCh38
rs544417261 Chr.1: 20827694-20827694 on Build GRCh38
rs758090814 Chr.1: 20827702-20827702 on Build GRCh38
rs1052853000 Chr.1: 20827805-20827805 on Build GRCh38
rs925524571 Chr.1: 20827759-20827759 on Build GRCh38
rs563407165 Chr.1: 20827878-20827878 on Build GRCh38
rs774622215 Chr.1: 20827606-20827606 on Build GRCh38
rs948746795 Chr.1: 20827828-20827828 on Build GRCh38

View Full Product Details