Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 21218959-21219461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECE1
Entrez Gene ID
1889
Gene Name
endothelin converting enzyme 1
Gene Aliases
ECE
UniGene
Hs.195080
Gene Chromosome Location
Chr.1: 21217247-21345541 on Build GRCh38
Transcripts
NM_001113347.1 NM_001113348.1 NM_001113349.1 NM_001397.2 XM_006710398.2 XM_011540872.2 XM_011540873.2 XM_017000511.1

SNP Detail

SNP ID Chromosome Location
rs554478099 Chr.1: 21219398-21219398 on Build GRCh38
rs956495641 Chr.1: 21219309-21219309 on Build GRCh38
rs946047777 Chr.1: 21218981-21218981 on Build GRCh38
rs1056581018 Chr.1: 21219102-21219102 on Build GRCh38
rs370563227 Chr.1: 21219007-21219007 on Build GRCh38
rs946663951 Chr.1: 21219062-21219062 on Build GRCh38
rs762017322 Chr.1: 21219422-21219422 on Build GRCh38
rs994596210 Chr.1: 21219073-21219073 on Build GRCh38
rs3026911 Chr.1: 21219089-21219089 on Build GRCh38
rs6701890 Chr.1: 21219285-21219285 on Build GRCh38
rs897968732 Chr.1: 21219125-21219125 on Build GRCh38
rs1045090599 Chr.1: 21219071-21219071 on Build GRCh38
rs527788834 Chr.1: 21219175-21219175 on Build GRCh38
rs888032295 Chr.1: 21219202-21219202 on Build GRCh38
rs550880787 Chr.1: 21219313-21219313 on Build GRCh38
rs939389960 Chr.1: 21219374-21219374 on Build GRCh38
rs572630539 Chr.1: 21219396-21219396 on Build GRCh38
rs763268181 Chr.1: 21219188-21219188 on Build GRCh38
rs1052327081 Chr.1: 21219000-21219000 on Build GRCh38
rs980746248 Chr.1: 21219330-21219330 on Build GRCh38
rs1019057509 Chr.1: 21219153-21219153 on Build GRCh38
rs975432078 Chr.1: 21219214-21219214 on Build GRCh38
rs963778193 Chr.1: 21219277-21219277 on Build GRCh38
rs976891922 Chr.1: 21219281-21219281 on Build GRCh38
rs532844615 Chr.1: 21219423-21219423 on Build GRCh38
rs375249865 Chr.1: 21219074-21219074 on Build GRCh38
rs1045733413 Chr.1: 21219024-21219024 on Build GRCh38
rs907187567 Chr.1: 21219028-21219028 on Build GRCh38
rs1018352996 Chr.1: 21219271-21219272 on Build GRCh38
rs532432401 Chr.1: 21219293-21219293 on Build GRCh38
rs1055460478 Chr.1: 21219417-21219417 on Build GRCh38
rs775955731 Chr.1: 21219122-21219122 on Build GRCh38
rs915120259 Chr.1: 21219315-21219315 on Build GRCh38
rs895007072 Chr.1: 21219060-21219060 on Build GRCh38
rs935177599 Chr.1: 21218987-21218988 on Build GRCh38
rs937350867 Chr.1: 21219410-21219410 on Build GRCh38
rs566185770 Chr.1: 21219419-21219419 on Build GRCh38
rs926602831 Chr.1: 21219331-21219331 on Build GRCh38
rs1029210673 Chr.1: 21219229-21219229 on Build GRCh38
rs990292551 Chr.1: 21219263-21219263 on Build GRCh38
rs1055048607 Chr.1: 21219052-21219052 on Build GRCh38
rs1004520459 Chr.1: 21219135-21219135 on Build GRCh38
rs568954735 Chr.1: 21219329-21219329 on Build GRCh38
rs946611693 Chr.1: 21219318-21219318 on Build GRCh38
rs28368054 Chr.1: 21219225-21219226 on Build GRCh38
rs3026910 Chr.1: 21219240-21219240 on Build GRCh38
rs746337307 Chr.1: 21219010-21219010 on Build GRCh38
rs1003894904 Chr.1: 21219050-21219050 on Build GRCh38
rs913279876 Chr.1: 21218980-21218980 on Build GRCh38
rs993640573 Chr.1: 21219142-21219142 on Build GRCh38
rs1027829707 Chr.1: 21219199-21219199 on Build GRCh38

View Full Product Details