Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40471205-40471714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST14
Entrez Gene ID
113189
Gene Name
carbohydrate sulfotransferase 14
Gene Aliases
ATCS, D4ST1, EDSMC1, HNK1ST
UniGene
Hs.442449
Gene Chromosome Location
Chr.15: 40470961-40473158 on Build GRCh38
Transcripts
NM_130468.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6628902 c.416G>A p.R139H Chr.15: 40471629-40471629 on Build GRCh38
5778290 c.455G>A p.C152Y Chr.15: 40471668-40471668 on Build GRCh38
6628903 c.441C>T p.Y147Y Chr.15: 40471654-40471654 on Build GRCh38
1188959 c.181G>T p.E61* Chr.15: 40471394-40471394 on Build GRCh38
6411449 c.229G>A p.G77S Chr.15: 40471442-40471442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770839880 Chr.15: 40471506-40471506 on Build GRCh38
rs910863196 Chr.15: 40471323-40471323 on Build GRCh38
rs562582335 Chr.15: 40471317-40471317 on Build GRCh38
rs866817984 Chr.15: 40471611-40471611 on Build GRCh38
rs960537765 Chr.15: 40471318-40471318 on Build GRCh38
rs986123630 Chr.15: 40471302-40471302 on Build GRCh38
rs754720949 Chr.15: 40471488-40471488 on Build GRCh38
rs1035955777 Chr.15: 40471291-40471291 on Build GRCh38
rs1055352198 Chr.15: 40471399-40471399 on Build GRCh38
rs774196289 Chr.15: 40471511-40471511 on Build GRCh38
rs577809616 Chr.15: 40471271-40471271 on Build GRCh38
rs753381343 Chr.15: 40471618-40471618 on Build GRCh38
rs1017593366 Chr.15: 40471680-40471680 on Build GRCh38
rs1007569346 Chr.15: 40471649-40471649 on Build GRCh38
rs896003662 Chr.15: 40471371-40471371 on Build GRCh38
rs754252026 Chr.15: 40471311-40471311 on Build GRCh38
rs774512581 Chr.15: 40471236-40471236 on Build GRCh38
rs770562379 Chr.15: 40471401-40471401 on Build GRCh38
rs770191510 Chr.15: 40471544-40471545 on Build GRCh38
rs760649086 Chr.15: 40471441-40471441 on Build GRCh38
rs910816020 Chr.15: 40471308-40471308 on Build GRCh38
rs760814663 Chr.15: 40471526-40471526 on Build GRCh38
rs561964206 Chr.15: 40471463-40471463 on Build GRCh38
rs1046833753 Chr.15: 40471361-40471361 on Build GRCh38
rs780919131 Chr.15: 40471356-40471356 on Build GRCh38
rs749118579 Chr.15: 40471504-40471504 on Build GRCh38
rs112899398 Chr.15: 40471579-40471579 on Build GRCh38
rs946414569 Chr.15: 40471328-40471328 on Build GRCh38
rs1043447552 Chr.15: 40471343-40471343 on Build GRCh38
rs191997027 Chr.15: 40471423-40471423 on Build GRCh38
rs775525832 Chr.15: 40471523-40471523 on Build GRCh38
rs200761477 Chr.15: 40471684-40471684 on Build GRCh38
rs999093521 Chr.15: 40471565-40471565 on Build GRCh38
rs139908298 Chr.15: 40471376-40471376 on Build GRCh38
rs955261065 Chr.15: 40471588-40471588 on Build GRCh38
rs747719839 Chr.15: 40471494-40471494 on Build GRCh38
rs749135787 Chr.15: 40471386-40471386 on Build GRCh38
rs35318007 Chr.15: 40471548-40471548 on Build GRCh38
rs1030815676 Chr.15: 40471245-40471245 on Build GRCh38
rs146457413 Chr.15: 40471650-40471650 on Build GRCh38
rs750814626 Chr.15: 40471481-40471481 on Build GRCh38
rs766643057 Chr.15: 40471233-40471233 on Build GRCh38
rs750869813 Chr.15: 40471563-40471563 on Build GRCh38
rs886042864 Chr.15: 40471287-40471287 on Build GRCh38
rs1030451531 Chr.15: 40471570-40471570 on Build GRCh38
rs545629118 Chr.15: 40471312-40471312 on Build GRCh38
rs267606730 Chr.15: 40471418-40471418 on Build GRCh38
rs1014902072 Chr.15: 40471411-40471411 on Build GRCh38
rs533124457 Chr.15: 40471639-40471639 on Build GRCh38
rs564319358 Chr.15: 40471583-40471583 on Build GRCh38
rs985415192 Chr.15: 40471301-40471301 on Build GRCh38
rs199749498 Chr.15: 40471364-40471364 on Build GRCh38
rs752221539 Chr.15: 40471591-40471591 on Build GRCh38
rs772328190 Chr.15: 40471514-40471514 on Build GRCh38
rs780337527 Chr.15: 40471642-40471642 on Build GRCh38
rs780973904 Chr.15: 40471493-40471493 on Build GRCh38
rs765811260 Chr.15: 40471549-40471549 on Build GRCh38
rs757224402 Chr.15: 40471630-40471630 on Build GRCh38
rs759613375 Chr.15: 40471261-40471261 on Build GRCh38
rs922470463 Chr.15: 40471238-40471238 on Build GRCh38
rs754666043 Chr.15: 40471325-40471325 on Build GRCh38
rs748369785 Chr.15: 40471419-40471420 on Build GRCh38
rs778171410 Chr.15: 40471616-40471630 on Build GRCh38
rs765756208 Chr.15: 40471480-40471480 on Build GRCh38
rs112549048 Chr.15: 40471544-40471544 on Build GRCh38
rs745630272 Chr.15: 40471512-40471512 on Build GRCh38
rs774567579 Chr.15: 40471416-40471416 on Build GRCh38
rs781489212 Chr.15: 40471412-40471413 on Build GRCh38
rs371189136 Chr.15: 40471452-40471452 on Build GRCh38
rs371684007 Chr.15: 40471501-40471501 on Build GRCh38
rs977564675 Chr.15: 40471227-40471227 on Build GRCh38
rs762296206 Chr.15: 40471548-40471548 on Build GRCh38
rs775350610 Chr.15: 40471652-40471652 on Build GRCh38
rs368203130 Chr.15: 40471613-40471613 on Build GRCh38
rs746603977 Chr.15: 40471324-40471324 on Build GRCh38
rs768920700 Chr.15: 40471651-40471651 on Build GRCh38
rs1018182086 Chr.15: 40471612-40471612 on Build GRCh38
rs764476073 Chr.15: 40471281-40471281 on Build GRCh38
rs550038340 Chr.15: 40471640-40471640 on Build GRCh38
rs772970656 Chr.15: 40471375-40471375 on Build GRCh38
rs201632751 Chr.15: 40471538-40471538 on Build GRCh38
rs397518432 Chr.15: 40471358-40471358 on Build GRCh38
rs903042632 Chr.15: 40471528-40471528 on Build GRCh38
rs267606727 Chr.15: 40471616-40471616 on Build GRCh38
rs761965484 Chr.15: 40471660-40471660 on Build GRCh38
rs267606728 Chr.15: 40471623-40471623 on Build GRCh38
rs757043277 Chr.15: 40471412-40471412 on Build GRCh38
rs755535418 Chr.15: 40471496-40471496 on Build GRCh38
rs755659406 Chr.15: 40471602-40471602 on Build GRCh38
rs777203532 Chr.15: 40471607-40471607 on Build GRCh38
rs747584345 Chr.15: 40471358-40471358 on Build GRCh38
rs752758290 Chr.15: 40471262-40471262 on Build GRCh38
rs759809750 Chr.15: 40471421-40471421 on Build GRCh38
rs938393246 Chr.15: 40471393-40471393 on Build GRCh38

View Full Product Details