Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40844617-40845127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPINT1
Entrez Gene ID
6692
Gene Name
serine peptidase inhibitor, Kunitz type 1
Gene Aliases
HAI, HAI1, MANSC2
UniGene
Hs.233950
Gene Chromosome Location
Chr.15: 40844018-40857655 on Build GRCh38
Transcripts
NM_001032367.1 NM_003710.3 NM_181642.2 XM_006720657.1 XM_011521957.1 XM_011521958.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6237764 c.254G>T p.W85L Chr.15: 40844808-40844808 on Build GRCh38
6789017 c.285G>T p.Q95H Chr.15: 40844839-40844839 on Build GRCh38
5740284 c.175C>T p.P59S Chr.15: 40844729-40844729 on Build GRCh38
7332253 c.207C>T p.V69V Chr.15: 40844761-40844761 on Build GRCh38
315564 c.307G>T p.E103* Chr.15: 40844861-40844861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779964722 Chr.15: 40844658-40844658 on Build GRCh38
rs756638082 Chr.15: 40844838-40844838 on Build GRCh38
rs111695130 Chr.15: 40844894-40844894 on Build GRCh38
rs565266707 Chr.15: 40844883-40844883 on Build GRCh38
rs776710634 Chr.15: 40844719-40844719 on Build GRCh38
rs372286432 Chr.15: 40844780-40844780 on Build GRCh38
rs368309266 Chr.15: 40844734-40844734 on Build GRCh38
rs770138846 Chr.15: 40844675-40844675 on Build GRCh38
rs758550828 Chr.15: 40844646-40844646 on Build GRCh38
rs373492914 Chr.15: 40844969-40844969 on Build GRCh38
rs190902803 Chr.15: 40844977-40844977 on Build GRCh38
rs545760845 Chr.15: 40844848-40844848 on Build GRCh38
rs144779474 Chr.15: 40844976-40844976 on Build GRCh38
rs768721508 Chr.15: 40844710-40844710 on Build GRCh38
rs551303454 Chr.15: 40845082-40845082 on Build GRCh38
rs202049185 Chr.15: 40845034-40845034 on Build GRCh38
rs779633506 Chr.15: 40844855-40844855 on Build GRCh38
rs145999931 Chr.15: 40844909-40844909 on Build GRCh38
rs770574048 Chr.15: 40844876-40844876 on Build GRCh38
rs747611192 Chr.15: 40844882-40844882 on Build GRCh38
rs750828679 Chr.15: 40845055-40845055 on Build GRCh38
rs754984432 Chr.15: 40844761-40844761 on Build GRCh38
rs758002274 Chr.15: 40844773-40844773 on Build GRCh38
rs780877965 Chr.15: 40844863-40844863 on Build GRCh38
rs11549915 Chr.15: 40844922-40844922 on Build GRCh38
rs754360257 Chr.15: 40844959-40844959 on Build GRCh38
rs764578411 Chr.15: 40844950-40844950 on Build GRCh38
rs372434962 Chr.15: 40845000-40845000 on Build GRCh38
rs745605901 Chr.15: 40845006-40845006 on Build GRCh38
rs148755059 Chr.15: 40844915-40844915 on Build GRCh38
rs763340218 Chr.15: 40845070-40845070 on Build GRCh38
rs763112135 Chr.15: 40844742-40844742 on Build GRCh38
rs971251491 Chr.15: 40844673-40844673 on Build GRCh38
rs759670416 Chr.15: 40844828-40844828 on Build GRCh38
rs749617310 Chr.15: 40844771-40844771 on Build GRCh38
rs201697101 Chr.15: 40844678-40844678 on Build GRCh38
rs767343780 Chr.15: 40845075-40845075 on Build GRCh38
rs755497485 Chr.15: 40844667-40844667 on Build GRCh38
rs760684271 Chr.15: 40844945-40844945 on Build GRCh38
rs749103967 Chr.15: 40844913-40844913 on Build GRCh38
rs199965511 Chr.15: 40844861-40844861 on Build GRCh38
rs543267493 Chr.15: 40844750-40844750 on Build GRCh38
rs762409531 Chr.15: 40845041-40845041 on Build GRCh38
rs201791058 Chr.15: 40844692-40844692 on Build GRCh38
rs759786834 Chr.15: 40844921-40844921 on Build GRCh38
rs762551018 Chr.15: 40844936-40844936 on Build GRCh38
rs746642409 Chr.15: 40844859-40844859 on Build GRCh38
rs141793805 Chr.15: 40845053-40845053 on Build GRCh38
rs754696591 Chr.15: 40844970-40844970 on Build GRCh38
rs200164851 Chr.15: 40844991-40844991 on Build GRCh38
rs12323939 Chr.15: 40844979-40844979 on Build GRCh38
rs750002104 Chr.15: 40844769-40844769 on Build GRCh38
rs746570646 Chr.15: 40844789-40844789 on Build GRCh38
rs777048289 Chr.15: 40845039-40845039 on Build GRCh38
rs1034696456 Chr.15: 40845053-40845053 on Build GRCh38
rs762138463 Chr.15: 40844960-40844960 on Build GRCh38
rs913937036 Chr.15: 40845003-40845003 on Build GRCh38
rs376333579 Chr.15: 40844720-40844720 on Build GRCh38
rs751601912 Chr.15: 40844758-40844758 on Build GRCh38
rs769812130 Chr.15: 40844810-40844810 on Build GRCh38
rs200486135 Chr.15: 40844802-40844802 on Build GRCh38
rs145578767 Chr.15: 40844820-40844820 on Build GRCh38
rs768272269 Chr.15: 40844794-40844794 on Build GRCh38
rs376462674 Chr.15: 40845007-40845007 on Build GRCh38
rs750594103 Chr.15: 40844644-40844644 on Build GRCh38
rs776753954 Chr.15: 40844795-40844795 on Build GRCh38
rs765098289 Chr.15: 40844722-40844722 on Build GRCh38
rs553670634 Chr.15: 40844762-40844762 on Build GRCh38
rs765625282 Chr.15: 40844638-40844638 on Build GRCh38
rs781620292 Chr.15: 40844668-40844668 on Build GRCh38
rs905116590 Chr.15: 40845029-40845029 on Build GRCh38
rs755644673 Chr.15: 40844988-40844988 on Build GRCh38
rs777628182 Chr.15: 40844989-40844989 on Build GRCh38
rs142567105 Chr.15: 40844835-40844835 on Build GRCh38
rs544670776 Chr.15: 40844912-40844912 on Build GRCh38
rs1046305723 Chr.15: 40845016-40845016 on Build GRCh38
rs375042808 Chr.15: 40844914-40844914 on Build GRCh38
rs796510478 Chr.15: 40845032-40845032 on Build GRCh38
rs371381706 Chr.15: 40845077-40845077 on Build GRCh38
rs751615505 Chr.15: 40844664-40844664 on Build GRCh38
rs765609743 Chr.15: 40845051-40845051 on Build GRCh38
rs373463208 Chr.15: 40844799-40844799 on Build GRCh38
rs138089109 Chr.15: 40844822-40844822 on Build GRCh38
rs748516131 Chr.15: 40844669-40844669 on Build GRCh38
rs760177902 Chr.15: 40844703-40844703 on Build GRCh38
rs914334103 Chr.15: 40844774-40844774 on Build GRCh38
rs757639019 Chr.15: 40844849-40844849 on Build GRCh38
rs756511656 Chr.15: 40844765-40844765 on Build GRCh38
rs544733342 Chr.15: 40844836-40844836 on Build GRCh38
rs200006775 Chr.15: 40845022-40845022 on Build GRCh38
rs775688195 Chr.15: 40845010-40845010 on Build GRCh38
rs764606165 Chr.15: 40844842-40844842 on Build GRCh38
rs376170333 Chr.15: 40844783-40844783 on Build GRCh38
rs897821279 Chr.15: 40844890-40844890 on Build GRCh38
rs11549914 Chr.15: 40844687-40844687 on Build GRCh38
rs772902109 Chr.15: 40844819-40844819 on Build GRCh38
rs745480373 Chr.15: 40844679-40844679 on Build GRCh38
rs993297271 Chr.15: 40845081-40845081 on Build GRCh38
rs368898985 Chr.15: 40844966-40844966 on Build GRCh38

View Full Product Details