Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41309747-41310257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OIP5
Entrez Gene ID
11339
Gene Name
Opa interacting protein 5
Gene Aliases
5730547N13Rik, CT86, LINT-25, MIS18B, MIS18beta, hMIS18beta
UniGene
Hs.661645
Gene Chromosome Location
Chr.15: 41309268-41332621 on Build GRCh38
Transcripts
NM_001317860.1 NM_007280.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6463867 c.606G>A p.K202K Chr.15: 41309838-41309838 on Build GRCh38
961386 c.617C>T p.T206M Chr.15: 41309827-41309827 on Build GRCh38
1158363 c.645G>T p.K215N Chr.15: 41309799-41309799 on Build GRCh38
5913695 c.642G>A p.M214I Chr.15: 41309802-41309802 on Build GRCh38
5760059 c.649C>T p.L217L Chr.15: 41309795-41309795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200589092 Chr.15: 41309821-41309821 on Build GRCh38
rs767482883 Chr.15: 41309923-41309923 on Build GRCh38
rs77683997 Chr.15: 41310158-41310158 on Build GRCh38
rs762272534 Chr.15: 41309808-41309808 on Build GRCh38
rs747096194 Chr.15: 41309850-41309850 on Build GRCh38
rs150485331 Chr.15: 41309819-41309819 on Build GRCh38
rs777320440 Chr.15: 41309857-41309857 on Build GRCh38
rs1020499509 Chr.15: 41310035-41310036 on Build GRCh38
rs143255947 Chr.15: 41309791-41309791 on Build GRCh38
rs780241051 Chr.15: 41309900-41309907 on Build GRCh38
rs777218157 Chr.15: 41309794-41309794 on Build GRCh38
rs758688316 Chr.15: 41309885-41309890 on Build GRCh38
rs1048334897 Chr.15: 41310089-41310089 on Build GRCh38
rs1039054011 Chr.15: 41309967-41309967 on Build GRCh38
rs777709218 Chr.15: 41309827-41309827 on Build GRCh38
rs752210328 Chr.15: 41309822-41309822 on Build GRCh38
rs775694150 Chr.15: 41309772-41309772 on Build GRCh38
rs1000453604 Chr.15: 41310080-41310080 on Build GRCh38
rs554128442 Chr.15: 41309811-41309811 on Build GRCh38
rs778549081 Chr.15: 41309835-41309835 on Build GRCh38
rs1012640993 Chr.15: 41309805-41309805 on Build GRCh38
rs556963181 Chr.15: 41310104-41310104 on Build GRCh38
rs753733170 Chr.15: 41309831-41309831 on Build GRCh38
rs370443628 Chr.15: 41310020-41310020 on Build GRCh38
rs572778978 Chr.15: 41310097-41310099 on Build GRCh38
rs377350927 Chr.15: 41309840-41309840 on Build GRCh38
rs959082436 Chr.15: 41310097-41310097 on Build GRCh38
rs537065800 Chr.15: 41310045-41310045 on Build GRCh38
rs970479988 Chr.15: 41310163-41310163 on Build GRCh38
rs1032047252 Chr.15: 41309782-41309782 on Build GRCh38
rs780354656 Chr.15: 41309848-41309848 on Build GRCh38
rs757115076 Chr.15: 41309834-41309834 on Build GRCh38
rs1012675913 Chr.15: 41310181-41310181 on Build GRCh38
rs750652978 Chr.15: 41309883-41309883 on Build GRCh38
rs777137142 Chr.15: 41310200-41310200 on Build GRCh38
rs775076610 Chr.15: 41309889-41309889 on Build GRCh38
rs930631924 Chr.15: 41310066-41310066 on Build GRCh38
rs890570888 Chr.15: 41310009-41310009 on Build GRCh38
rs931939397 Chr.15: 41309962-41309962 on Build GRCh38
rs370924216 Chr.15: 41309856-41309856 on Build GRCh38
rs900501596 Chr.15: 41309920-41309920 on Build GRCh38
rs765472877 Chr.15: 41309882-41309882 on Build GRCh38
rs374289568 Chr.15: 41309815-41309815 on Build GRCh38
rs770254563 Chr.15: 41309865-41309865 on Build GRCh38
rs763310510 Chr.15: 41309867-41309867 on Build GRCh38
rs1010314936 Chr.15: 41310023-41310023 on Build GRCh38
rs895615296 Chr.15: 41310162-41310162 on Build GRCh38
rs139348470 Chr.15: 41309816-41309816 on Build GRCh38
rs775595534 Chr.15: 41309913-41309913 on Build GRCh38
rs755564722 Chr.15: 41309826-41309826 on Build GRCh38
rs943470605 Chr.15: 41309869-41309869 on Build GRCh38
rs979078627 Chr.15: 41309818-41309818 on Build GRCh38
rs767171708 Chr.15: 41309875-41309875 on Build GRCh38
rs761977741 Chr.15: 41309809-41309809 on Build GRCh38
rs760765825 Chr.15: 41310065-41310065 on Build GRCh38
rs573662575 Chr.15: 41310115-41310115 on Build GRCh38
rs547051997 Chr.15: 41309786-41309786 on Build GRCh38
rs959794488 Chr.15: 41309790-41309790 on Build GRCh38
rs748585712 Chr.15: 41309860-41309860 on Build GRCh38

View Full Product Details