Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41350801-41351200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUSAP1
Entrez Gene ID
51203
Gene Name
nucleolar and spindle associated protein 1
Gene Aliases
ANKT, BM037, LNP, NUSAP, PRO0310p1, Q0310, SAPL
UniGene
Hs.615092
Gene Chromosome Location
Chr.15: 41332694-41381050 on Build GRCh38
Transcripts
NM_001243142.1 NM_001243143.1 NM_001243144.1 NM_001301136.1 NM_016359.4 NM_018454.7 XM_005254428.2 XM_005254430.4 XM_005254431.2 XM_006720559.2 XM_006720560.3 XM_006720561.2 XM_006720562.2 XM_006720563.2 XM_017022294.1 XM_017022295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6456087 c.430G>T p.E144* Chr.15: 41351111-41351111 on Build GRCh38
3886931 c.355C>T p.H119Y Chr.15: 41351036-41351036 on Build GRCh38
961392 c.330A>G p.I110M Chr.15: 41351011-41351011 on Build GRCh38
961393 c.412G>C p.D138H Chr.15: 41351093-41351093 on Build GRCh38
4835136 c.381C>T p.L127L Chr.15: 41351062-41351062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022692579 Chr.15: 41350842-41350842 on Build GRCh38
rs368317188 Chr.15: 41350957-41350957 on Build GRCh38
rs747363394 Chr.15: 41350976-41350976 on Build GRCh38
rs1031307891 Chr.15: 41350900-41350900 on Build GRCh38
rs1048360282 Chr.15: 41350895-41350895 on Build GRCh38
rs757467371 Chr.15: 41350956-41350956 on Build GRCh38
rs779216392 Chr.15: 41351061-41351061 on Build GRCh38
rs780560157 Chr.15: 41350967-41350967 on Build GRCh38
rs373496789 Chr.15: 41351082-41351082 on Build GRCh38
rs964890325 Chr.15: 41350965-41350965 on Build GRCh38
rs755901337 Chr.15: 41350942-41350942 on Build GRCh38
rs772529323 Chr.15: 41350984-41350984 on Build GRCh38
rs760653952 Chr.15: 41351163-41351163 on Build GRCh38
rs748456499 Chr.15: 41351113-41351113 on Build GRCh38
rs968410197 Chr.15: 41350851-41350851 on Build GRCh38
rs192933106 Chr.15: 41350865-41350865 on Build GRCh38
rs200386583 Chr.15: 41351025-41351025 on Build GRCh38
rs763940798 Chr.15: 41351046-41351046 on Build GRCh38
rs777902249 Chr.15: 41350943-41350943 on Build GRCh38
rs547089226 Chr.15: 41350962-41350962 on Build GRCh38
rs368868122 Chr.15: 41351071-41351071 on Build GRCh38
rs189325557 Chr.15: 41351093-41351093 on Build GRCh38
rs200519598 Chr.15: 41351022-41351022 on Build GRCh38
rs920672223 Chr.15: 41350982-41350982 on Build GRCh38
rs778622787 Chr.15: 41351083-41351083 on Build GRCh38
rs376382140 Chr.15: 41351121-41351121 on Build GRCh38
rs772787580 Chr.15: 41350989-41350989 on Build GRCh38
rs199580182 Chr.15: 41351089-41351089 on Build GRCh38
rs565898344 Chr.15: 41350968-41350968 on Build GRCh38
rs999907013 Chr.15: 41351135-41351135 on Build GRCh38
rs754072126 Chr.15: 41350953-41350953 on Build GRCh38
rs772738465 Chr.15: 41351128-41351128 on Build GRCh38
rs745563327 Chr.15: 41351085-41351087 on Build GRCh38
rs928449171 Chr.15: 41350827-41350827 on Build GRCh38
rs184240094 Chr.15: 41351013-41351013 on Build GRCh38
rs763866474 Chr.15: 41351165-41351165 on Build GRCh38
rs773955590 Chr.15: 41351127-41351127 on Build GRCh38
rs746766695 Chr.15: 41351148-41351148 on Build GRCh38
rs375265995 Chr.15: 41350985-41350985 on Build GRCh38
rs750550611 Chr.15: 41351075-41351075 on Build GRCh38
rs779705461 Chr.15: 41350951-41350951 on Build GRCh38
rs1006943870 Chr.15: 41350904-41350904 on Build GRCh38
rs897191951 Chr.15: 41350870-41350870 on Build GRCh38
rs531453630 Chr.15: 41350848-41350852 on Build GRCh38
rs369330327 Chr.15: 41350994-41350994 on Build GRCh38
rs771513120 Chr.15: 41350933-41350934 on Build GRCh38
rs375966047 Chr.15: 41351032-41351032 on Build GRCh38
rs956276952 Chr.15: 41350930-41350930 on Build GRCh38
rs762590744 Chr.15: 41350993-41350993 on Build GRCh38
rs747528263 Chr.15: 41351095-41351095 on Build GRCh38
rs771593156 Chr.15: 41351141-41351141 on Build GRCh38
rs776578468 Chr.15: 41351175-41351175 on Build GRCh38
rs773867437 Chr.15: 41351004-41351004 on Build GRCh38
rs372347241 Chr.15: 41350961-41350961 on Build GRCh38
rs556917845 Chr.15: 41351096-41351096 on Build GRCh38
rs757553840 Chr.15: 41351053-41351053 on Build GRCh38
rs769126935 Chr.15: 41351098-41351098 on Build GRCh38
rs931296779 Chr.15: 41350876-41350876 on Build GRCh38
rs754072283 Chr.15: 41351052-41351052 on Build GRCh38
rs550993263 Chr.15: 41350832-41350832 on Build GRCh38

View Full Product Details