Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42079397-42079906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G4D
Entrez Gene ID
283748
Gene Name
phospholipase A2 group IVD
Gene Aliases
cPLA2delta
UniGene
Hs.380225
Gene Chromosome Location
Chr.15: 42067562-42094559 on Build GRCh38
Transcripts
NM_178034.3 XM_011521467.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5362876 c.1210C>T p.R404C Chr.15: 42079644-42079644 on Build GRCh38
3969105 c.1281G>C p.L427L Chr.15: 42079573-42079573 on Build GRCh38
3927506 c.1095-10G>A p.? Chr.15: 42079769-42079769 on Build GRCh38
3501215 c.1100T>C p.M367T Chr.15: 42079754-42079754 on Build GRCh38
2186873 c.1225C>T p.R409C Chr.15: 42079629-42079629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776830539 Chr.15: 42079653-42079653 on Build GRCh38
rs747181263 Chr.15: 42079739-42079739 on Build GRCh38
rs762723864 Chr.15: 42079591-42079591 on Build GRCh38
rs564129708 Chr.15: 42079794-42079794 on Build GRCh38
rs770742077 Chr.15: 42079770-42079770 on Build GRCh38
rs763787378 Chr.15: 42079841-42079842 on Build GRCh38
rs755724653 Chr.15: 42079629-42079629 on Build GRCh38
rs745806594 Chr.15: 42079773-42079773 on Build GRCh38
rs921334601 Chr.15: 42079542-42079542 on Build GRCh38
rs768924485 Chr.15: 42079650-42079650 on Build GRCh38
rs541133267 Chr.15: 42079793-42079793 on Build GRCh38
rs990961668 Chr.15: 42079608-42079608 on Build GRCh38
rs549615926 Chr.15: 42079881-42079881 on Build GRCh38
rs779546424 Chr.15: 42079567-42079567 on Build GRCh38
rs984187840 Chr.15: 42079838-42079838 on Build GRCh38
rs757688840 Chr.15: 42079636-42079636 on Build GRCh38
rs549341305 Chr.15: 42079459-42079459 on Build GRCh38
rs563253408 Chr.15: 42079875-42079875 on Build GRCh38
rs1027464041 Chr.15: 42079833-42079833 on Build GRCh38
rs761317866 Chr.15: 42079433-42079433 on Build GRCh38
rs369952855 Chr.15: 42079487-42079487 on Build GRCh38
rs750858244 Chr.15: 42079801-42079801 on Build GRCh38
rs150477834 Chr.15: 42079600-42079600 on Build GRCh38
rs751376810 Chr.15: 42079698-42079698 on Build GRCh38
rs775282416 Chr.15: 42079785-42079785 on Build GRCh38
rs375189436 Chr.15: 42079688-42079688 on Build GRCh38
rs891041131 Chr.15: 42079831-42079831 on Build GRCh38
rs201310162 Chr.15: 42079533-42079533 on Build GRCh38
rs112756552 Chr.15: 42079517-42079517 on Build GRCh38
rs781443451 Chr.15: 42079755-42079755 on Build GRCh38
rs780118874 Chr.15: 42079644-42079644 on Build GRCh38
rs145134059 Chr.15: 42079693-42079693 on Build GRCh38
rs750384290 Chr.15: 42079507-42079507 on Build GRCh38
rs752371395 Chr.15: 42079552-42079552 on Build GRCh38
rs543490926 Chr.15: 42079849-42079849 on Build GRCh38
rs774177182 Chr.15: 42079662-42079662 on Build GRCh38
rs150236163 Chr.15: 42079842-42079844 on Build GRCh38
rs959508458 Chr.15: 42079565-42079565 on Build GRCh38
rs747210167 Chr.15: 42079648-42079648 on Build GRCh38
rs751084346 Chr.15: 42079585-42079585 on Build GRCh38
rs956153365 Chr.15: 42079490-42079490 on Build GRCh38
rs750901921 Chr.15: 42079713-42079713 on Build GRCh38
rs542084472 Chr.15: 42079634-42079634 on Build GRCh38
rs372877660 Chr.15: 42079799-42079799 on Build GRCh38
rs763223901 Chr.15: 42079806-42079806 on Build GRCh38
rs1012653149 Chr.15: 42079448-42079448 on Build GRCh38
rs754509238 Chr.15: 42079548-42079548 on Build GRCh38
rs566247594 Chr.15: 42079468-42079468 on Build GRCh38
rs1031762178 Chr.15: 42079716-42079716 on Build GRCh38
rs758442248 Chr.15: 42079641-42079641 on Build GRCh38
rs771503723 Chr.15: 42079566-42079566 on Build GRCh38
rs751469539 Chr.15: 42079603-42079603 on Build GRCh38
rs746000051 Chr.15: 42079568-42079568 on Build GRCh38
rs748319464 Chr.15: 42079769-42079769 on Build GRCh38
rs1008317118 Chr.15: 42079523-42079523 on Build GRCh38
rs775599625 Chr.15: 42079588-42079588 on Build GRCh38
rs772171805 Chr.15: 42079585-42079585 on Build GRCh38
rs556925463 Chr.15: 42079589-42079589 on Build GRCh38
rs868324508 Chr.15: 42079737-42079737 on Build GRCh38
rs760087005 Chr.15: 42079679-42079679 on Build GRCh38
rs766940241 Chr.15: 42079673-42079673 on Build GRCh38
rs541337149 Chr.15: 42079493-42079493 on Build GRCh38
rs775774497 Chr.15: 42079509-42079509 on Build GRCh38
rs151093842 Chr.15: 42079557-42079557 on Build GRCh38
rs968443098 Chr.15: 42079461-42079461 on Build GRCh38
rs942141667 Chr.15: 42079723-42079723 on Build GRCh38
rs772100342 Chr.15: 42079777-42079777 on Build GRCh38
rs758886570 Chr.15: 42079719-42079719 on Build GRCh38
rs917946541 Chr.15: 42079424-42079424 on Build GRCh38
rs766951636 Chr.15: 42079811-42079811 on Build GRCh38
rs765922845 Chr.15: 42079488-42079489 on Build GRCh38
rs202036676 Chr.15: 42079637-42079637 on Build GRCh38
rs759477315 Chr.15: 42079663-42079663 on Build GRCh38
rs577617981 Chr.15: 42079524-42079524 on Build GRCh38
rs755021428 Chr.15: 42079745-42079745 on Build GRCh38
rs753508253 Chr.15: 42079689-42079689 on Build GRCh38
rs4924618 Chr.15: 42079554-42079554 on Build GRCh38
rs999909538 Chr.15: 42079692-42079692 on Build GRCh38
rs139838585 Chr.15: 42079625-42079625 on Build GRCh38
rs75210712 Chr.15: 42079444-42079444 on Build GRCh38
rs199835123 Chr.15: 42079677-42079677 on Build GRCh38
rs372294896 Chr.15: 42079620-42079620 on Build GRCh38
rs780745838 Chr.15: 42079549-42079549 on Build GRCh38
rs940021762 Chr.15: 42079877-42079877 on Build GRCh38
rs749123937 Chr.15: 42079659-42079659 on Build GRCh38
rs551295205 Chr.15: 42079834-42079834 on Build GRCh38
rs770688776 Chr.15: 42079661-42079661 on Build GRCh38
rs1047693405 Chr.15: 42079436-42079436 on Build GRCh38
rs529592283 Chr.15: 42079878-42079878 on Build GRCh38
rs576303647 Chr.15: 42079597-42079597 on Build GRCh38
rs774502025 Chr.15: 42079488-42079488 on Build GRCh38
rs774127881 Chr.15: 42079772-42079772 on Build GRCh38
rs35446774 Chr.15: 42079846-42079847 on Build GRCh38
rs760299006 Chr.15: 42079791-42079791 on Build GRCh38
rs765887048 Chr.15: 42079530-42079530 on Build GRCh38
rs752322041 Chr.15: 42079626-42079626 on Build GRCh38
rs746340363 Chr.15: 42079638-42079638 on Build GRCh38
rs765660120 Chr.15: 42079701-42079701 on Build GRCh38
rs761564302 Chr.15: 42079510-42079510 on Build GRCh38
rs147588006 Chr.15: 42079726-42079726 on Build GRCh38
rs974584943 Chr.15: 42079814-42079814 on Build GRCh38

View Full Product Details