Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23054979-23055478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM1A
Entrez Gene ID
23028
Gene Name
lysine demethylase 1A
Gene Aliases
AOF2, BHC110, CPRF, KDM1, LSD1
UniGene
Hs.591518
Gene Chromosome Location
Chr.1: 23019443-23083691 on Build GRCh38
Transcripts
NM_001009999.2 NM_015013.3 XM_005245786.2 XM_006710472.3 XM_006710473.2 XM_006710474.3 XM_017000716.1 XM_017000717.1 XM_017000718.1 XM_017000719.1 XM_017000720.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4617357 c.809T>C p.I270T Chr.1: 23055147-23055147 on Build GRCh38
905706 c.847A>G p.N283D Chr.1: 23055125-23055125 on Build GRCh38
6274391 c.829G>T p.E277* Chr.1: 23055107-23055107 on Build GRCh38
168257 c.852C>T p.F284F Chr.1: 23055130-23055130 on Build GRCh38
7190532 c.808C>T p.H270Y Chr.1: 23055086-23055086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577137998 Chr.1: 23055223-23055228 on Build GRCh38
rs979036196 Chr.1: 23055419-23055419 on Build GRCh38
rs781116924 Chr.1: 23055157-23055157 on Build GRCh38
rs952663381 Chr.1: 23055091-23055091 on Build GRCh38
rs978839477 Chr.1: 23055233-23055233 on Build GRCh38
rs759638292 Chr.1: 23055092-23055092 on Build GRCh38
rs189014649 Chr.1: 23055130-23055130 on Build GRCh38
rs895576650 Chr.1: 23055394-23055394 on Build GRCh38
rs755924133 Chr.1: 23055183-23055183 on Build GRCh38
rs538753821 Chr.1: 23055309-23055309 on Build GRCh38
rs1034467618 Chr.1: 23055429-23055429 on Build GRCh38
rs532200849 Chr.1: 23055098-23055098 on Build GRCh38
rs926929210 Chr.1: 23055306-23055306 on Build GRCh38
rs751541220 Chr.1: 23055033-23055033 on Build GRCh38
rs11380187 Chr.1: 23055302-23055303 on Build GRCh38
rs372226085 Chr.1: 23055065-23055065 on Build GRCh38
rs929777446 Chr.1: 23055301-23055302 on Build GRCh38
rs375571541 Chr.1: 23055208-23055208 on Build GRCh38
rs764506279 Chr.1: 23055128-23055128 on Build GRCh38
rs747928798 Chr.1: 23055043-23055043 on Build GRCh38
rs1057277688 Chr.1: 23055387-23055387 on Build GRCh38
rs937114921 Chr.1: 23055244-23055244 on Build GRCh38
rs533751061 Chr.1: 23055245-23055245 on Build GRCh38
rs557364184 Chr.1: 23055408-23055408 on Build GRCh38
rs981046792 Chr.1: 23055257-23055257 on Build GRCh38
rs754447616 Chr.1: 23055155-23055155 on Build GRCh38
rs905580019 Chr.1: 23055005-23055005 on Build GRCh38
rs751093595 Chr.1: 23055154-23055154 on Build GRCh38
rs754891701 Chr.1: 23055036-23055036 on Build GRCh38
rs992570812 Chr.1: 23055295-23055297 on Build GRCh38
rs377263766 Chr.1: 23055319-23055322 on Build GRCh38
rs968967251 Chr.1: 23055229-23055229 on Build GRCh38
rs552968204 Chr.1: 23055419-23055419 on Build GRCh38
rs888374013 Chr.1: 23055315-23055315 on Build GRCh38
rs779535750 Chr.1: 23055031-23055031 on Build GRCh38
rs1010371751 Chr.1: 23055418-23055419 on Build GRCh38
rs996244573 Chr.1: 23055344-23055345 on Build GRCh38
rs917118218 Chr.1: 23055299-23055299 on Build GRCh38
rs554502733 Chr.1: 23055209-23055209 on Build GRCh38
rs370886717 Chr.1: 23055207-23055207 on Build GRCh38
rs566400335 Chr.1: 23055241-23055241 on Build GRCh38
rs201973427 Chr.1: 23055203-23055203 on Build GRCh38
rs33996438 Chr.1: 23055303-23055304 on Build GRCh38
rs890514980 Chr.1: 23055407-23055407 on Build GRCh38
rs753380832 Chr.1: 23055095-23055096 on Build GRCh38
rs368796230 Chr.1: 23055061-23055061 on Build GRCh38
rs376984385 Chr.1: 23055042-23055042 on Build GRCh38
rs200154927 Chr.1: 23055021-23055021 on Build GRCh38
rs369397752 Chr.1: 23055175-23055175 on Build GRCh38
rs777589851 Chr.1: 23055184-23055184 on Build GRCh38
rs941253620 Chr.1: 23055317-23055319 on Build GRCh38
rs756940725 Chr.1: 23055205-23055206 on Build GRCh38
rs45549934 Chr.1: 23055240-23055240 on Build GRCh38
rs762638667 Chr.1: 23055136-23055136 on Build GRCh38
rs964043934 Chr.1: 23055123-23055123 on Build GRCh38
rs564982198 Chr.1: 23055018-23055018 on Build GRCh38
rs534952047 Chr.1: 23055322-23055323 on Build GRCh38
rs937031512 Chr.1: 23055342-23055342 on Build GRCh38
rs201822414 Chr.1: 23055302-23055302 on Build GRCh38
rs139626164 Chr.1: 23055103-23055103 on Build GRCh38
rs969376363 Chr.1: 23055419-23055422 on Build GRCh38
rs767305635 Chr.1: 23055007-23055007 on Build GRCh38
rs774550728 Chr.1: 23055076-23055076 on Build GRCh38
rs571410343 Chr.1: 23055280-23055280 on Build GRCh38
rs760607108 Chr.1: 23055058-23055058 on Build GRCh38
rs760962872 Chr.1: 23055115-23055115 on Build GRCh38
rs558287397 Chr.1: 23055262-23055262 on Build GRCh38
rs375167986 Chr.1: 23055147-23055147 on Build GRCh38

View Full Product Details